Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7119411-7119991 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INSR
Entrez Gene ID
3643
Gene Name
insulin receptor
Gene Aliases
CD220, HHF5
UniGene
Hs.465744
Gene Chromosome Location
Chr.19: 7112255-7294405 on Build GRCh38
Transcripts
NM_000208.3 NM_001079817.2 XM_011527988.2 XM_011527989.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5641114 c.3669C>G p.G1223G Chr.19: 7119574-7119574 on Build GRCh38
5894021 c.3660-6C>T p.? Chr.19: 7119589-7119589 on Build GRCh38
6293661 c.3680G>A p.W1227* Chr.19: 7119563-7119563 on Build GRCh38
4817360 c.3722C>G p.S1241C Chr.19: 7119521-7119521 on Build GRCh38
6983040 c.3757G>A p.G1253R Chr.19: 7119486-7119486 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs934979507 Chr.19: 7119779-7119788 on Build GRCh38
rs759260656 Chr.19: 7119505-7119505 on Build GRCh38
rs1054855507 Chr.19: 7119627-7119627 on Build GRCh38
rs143582066 Chr.19: 7119809-7119809 on Build GRCh38
rs544679202 Chr.19: 7119667-7119667 on Build GRCh38
rs1045728051 Chr.19: 7119717-7119717 on Build GRCh38
rs66692481 Chr.19: 7119844-7119847 on Build GRCh38
rs201739191 Chr.19: 7119760-7119761 on Build GRCh38
rs559000169 Chr.19: 7119662-7119662 on Build GRCh38
rs66500056 Chr.19: 7119855-7119856 on Build GRCh38
rs13306448 Chr.19: 7119647-7119647 on Build GRCh38
rs778542110 Chr.19: 7119905-7119905 on Build GRCh38
rs369102740 Chr.19: 7119468-7119468 on Build GRCh38
rs770709866 Chr.19: 7119632-7119651 on Build GRCh38
rs1028120046 Chr.19: 7119742-7119742 on Build GRCh38
rs142276524 Chr.19: 7119866-7119866 on Build GRCh38
rs12973977 Chr.19: 7119877-7119877 on Build GRCh38
rs548043043 Chr.19: 7119702-7119715 on Build GRCh38
rs67847403 Chr.19: 7119775-7119776 on Build GRCh38
rs775027642 Chr.19: 7119527-7119527 on Build GRCh38
rs942581045 Chr.19: 7119818-7119825 on Build GRCh38
rs779620267 Chr.19: 7119571-7119582 on Build GRCh38
rs145880723 Chr.19: 7119715-7119715 on Build GRCh38
rs990981450 Chr.19: 7119734-7119734 on Build GRCh38
rs1024239392 Chr.19: 7119881-7119881 on Build GRCh38
rs958959761 Chr.19: 7119865-7119865 on Build GRCh38
rs747127077 Chr.19: 7119581-7119581 on Build GRCh38
rs202117673 Chr.19: 7119834-7119834 on Build GRCh38
rs760144521 Chr.19: 7119544-7119544 on Build GRCh38
rs577057544 Chr.19: 7119645-7119645 on Build GRCh38
rs986466665 Chr.19: 7119664-7119664 on Build GRCh38
rs755555740 Chr.19: 7119718-7119718 on Build GRCh38
rs1016133125 Chr.19: 7119731-7119731 on Build GRCh38
rs869039553 Chr.19: 7119773-7119774 on Build GRCh38
rs749482249 Chr.19: 7119823-7119824 on Build GRCh38
rs530035689 Chr.19: 7119638-7119639 on Build GRCh38
rs555144543 Chr.19: 7119857-7119857 on Build GRCh38
rs948986449 Chr.19: 7119831-7119831 on Build GRCh38
rs928083858 Chr.19: 7119606-7119606 on Build GRCh38
rs745624546 Chr.19: 7119589-7119589 on Build GRCh38
rs568258415 Chr.19: 7119741-7119741 on Build GRCh38
rs772838497 Chr.19: 7119626-7119629 on Build GRCh38
rs1042557031 Chr.19: 7119670-7119670 on Build GRCh38
rs561271305 Chr.19: 7119668-7119668 on Build GRCh38
rs754327246 Chr.19: 7119558-7119558 on Build GRCh38
rs866487628 Chr.19: 7119638-7119639 on Build GRCh38
rs966799621 Chr.19: 7119777-7119777 on Build GRCh38
rs66637440 Chr.19: 7119824-7119829 on Build GRCh38
rs770750918 Chr.19: 7119490-7119490 on Build GRCh38
rs867227083 Chr.19: 7119875-7119875 on Build GRCh38
rs757664840 Chr.19: 7119569-7119569 on Build GRCh38
rs144222434 Chr.19: 7119843-7119843 on Build GRCh38
rs375179509 Chr.19: 7119779-7119779 on Build GRCh38
rs367660471 Chr.19: 7119679-7119679 on Build GRCh38
rs886465927 Chr.19: 7119844-7119844 on Build GRCh38
rs752355444 Chr.19: 7119761-7119762 on Build GRCh38
rs1043680533 Chr.19: 7119639-7119640 on Build GRCh38
rs766507353 Chr.19: 7119652-7119669 on Build GRCh38
rs560507679 Chr.19: 7119933-7119933 on Build GRCh38
rs200860415 Chr.19: 7119651-7119651 on Build GRCh38
rs149013063 Chr.19: 7119729-7119729 on Build GRCh38
rs556744836 Chr.19: 7119908-7119911 on Build GRCh38
rs1028171023 Chr.19: 7119882-7119882 on Build GRCh38
rs141200869 Chr.19: 7119772-7119773 on Build GRCh38
rs747038967 Chr.19: 7119442-7119442 on Build GRCh38
rs771749441 Chr.19: 7119608-7119608 on Build GRCh38
rs750713967 Chr.19: 7119574-7119574 on Build GRCh38
rs201814608 Chr.19: 7119653-7119653 on Build GRCh38
rs760234510 Chr.19: 7119618-7119618 on Build GRCh38
rs187956009 Chr.19: 7119642-7119642 on Build GRCh38
rs936391784 Chr.19: 7119781-7119781 on Build GRCh38
rs556403781 Chr.19: 7119826-7119826 on Build GRCh38
rs977785674 Chr.19: 7119778-7119778 on Build GRCh38
rs773962804 Chr.19: 7119495-7119495 on Build GRCh38
rs397859535 Chr.19: 7119660-7119661 on Build GRCh38
rs772119949 Chr.19: 7119436-7119436 on Build GRCh38
rs1050945347 Chr.19: 7119688-7119688 on Build GRCh38
rs371272167 Chr.19: 7119764-7119765 on Build GRCh38
rs149597782 Chr.19: 7119842-7119842 on Build GRCh38
rs574862932 Chr.19: 7119897-7119897 on Build GRCh38
rs755247502 Chr.19: 7119812-7119813 on Build GRCh38
rs72355649 Chr.19: 7119829-7119832 on Build GRCh38
rs77057545 Chr.19: 7119854-7119854 on Build GRCh38
rs143017342 Chr.19: 7119562-7119562 on Build GRCh38
rs898299432 Chr.19: 7119829-7119830 on Build GRCh38
rs916056630 Chr.19: 7119793-7119793 on Build GRCh38
rs150710908 Chr.19: 7119898-7119909 on Build GRCh38
rs939095952 Chr.19: 7119620-7119620 on Build GRCh38
rs139301762 Chr.19: 7119840-7119840 on Build GRCh38
rs113761062 Chr.19: 7119641-7119641 on Build GRCh38
rs773256337 Chr.19: 7119470-7119470 on Build GRCh38
rs763605469 Chr.19: 7119631-7119631 on Build GRCh38
rs978170706 Chr.19: 7119955-7119955 on Build GRCh38
rs62124489 Chr.19: 7119772-7119772 on Build GRCh38
rs1049124970 Chr.19: 7119838-7119838 on Build GRCh38
rs770711598 Chr.19: 7119588-7119588 on Build GRCh38
rs367614576 Chr.19: 7119625-7119626 on Build GRCh38
rs1043788178 Chr.19: 7119669-7119670 on Build GRCh38
rs775117360 Chr.19: 7119617-7119617 on Build GRCh38
rs916716318 Chr.19: 7119763-7119763 on Build GRCh38
rs777917515 Chr.19: 7119583-7119583 on Build GRCh38
rs764650383 Chr.19: 7119556-7119556 on Build GRCh38
rs953960024 Chr.19: 7119771-7119771 on Build GRCh38
rs765638025 Chr.19: 7119573-7119573 on Build GRCh38
rs398033814 Chr.19: 7119861-7119862 on Build GRCh38
rs531314011 Chr.19: 7119732-7119732 on Build GRCh38
rs121913140 Chr.19: 7119563-7119563 on Build GRCh38
rs995349044 Chr.19: 7119830-7119833 on Build GRCh38
rs539685828 Chr.19: 7119807-7119807 on Build GRCh38
rs780403712 Chr.19: 7119576-7119576 on Build GRCh38
rs4804295 Chr.19: 7119829-7119829 on Build GRCh38
rs990596637 Chr.19: 7119661-7119661 on Build GRCh38
rs547353148 Chr.19: 7119775-7119775 on Build GRCh38
rs12973088 Chr.19: 7119853-7119853 on Build GRCh38
rs960896909 Chr.19: 7119733-7119733 on Build GRCh38
rs201979105 Chr.19: 7119582-7119582 on Build GRCh38
rs66689882 Chr.19: 7119829-7119834 on Build GRCh38
rs567260236 Chr.19: 7119710-7119711 on Build GRCh38
rs979615479 Chr.19: 7119898-7119901 on Build GRCh38
rs373627875 Chr.19: 7119478-7119478 on Build GRCh38
rs1004681692 Chr.19: 7119643-7119643 on Build GRCh38
rs780312000 Chr.19: 7119441-7119441 on Build GRCh38
rs760701752 Chr.19: 7119630-7119651 on Build GRCh38
rs79631971 Chr.19: 7119856-7119856 on Build GRCh38
rs1019990571 Chr.19: 7119942-7119942 on Build GRCh38
rs1019513055 Chr.19: 7119776-7119776 on Build GRCh38
rs767023998 Chr.19: 7119514-7119514 on Build GRCh38
rs12973110 Chr.19: 7119879-7119879 on Build GRCh38
rs138211567 Chr.19: 7119722-7119722 on Build GRCh38
rs184752862 Chr.19: 7119640-7119640 on Build GRCh38
rs894740567 Chr.19: 7119713-7119713 on Build GRCh38
rs986428284 Chr.19: 7119944-7119944 on Build GRCh38
rs748736656 Chr.19: 7119601-7119603 on Build GRCh38
rs57039491 Chr.19: 7119774-7119775 on Build GRCh38
rs386806416 Chr.19: 7119640-7119642 on Build GRCh38
rs556367406 Chr.19: 7119648-7119648 on Build GRCh38
rs149069397 Chr.19: 7119561-7119561 on Build GRCh38
rs199914604 Chr.19: 7119650-7119650 on Build GRCh38
rs943351030 Chr.19: 7119829-7119854 on Build GRCh38
rs35625633 Chr.19: 7119652-7119653 on Build GRCh38
rs148200572 Chr.19: 7119593-7119593 on Build GRCh38
rs554209490 Chr.19: 7119906-7119906 on Build GRCh38
rs13306447 Chr.19: 7119680-7119680 on Build GRCh38
rs537517354 Chr.19: 7119639-7119639 on Build GRCh38
rs375291229 Chr.19: 7119913-7119924 on Build GRCh38
rs12973087 Chr.19: 7119851-7119851 on Build GRCh38
rs550695657 Chr.19: 7119826-7119829 on Build GRCh38
rs4031067 Chr.19: 7119653-7119654 on Build GRCh38
rs950723426 Chr.19: 7119724-7119727 on Build GRCh38
rs66587318 Chr.19: 7119850-7119853 on Build GRCh38
rs111513557 Chr.19: 7119855-7119855 on Build GRCh38
rs45453993 Chr.19: 7119919-7119919 on Build GRCh38
rs553828160 Chr.19: 7119644-7119644 on Build GRCh38
rs532816509 Chr.19: 7119705-7119705 on Build GRCh38
rs757646640 Chr.19: 7119568-7119568 on Build GRCh38
rs564998478 Chr.19: 7119641-7119642 on Build GRCh38
rs776866701 Chr.19: 7119632-7119633 on Build GRCh38
rs753484254 Chr.19: 7119791-7119791 on Build GRCh38
rs376773589 Chr.19: 7119523-7119523 on Build GRCh38
rs137913471 Chr.19: 7119764-7119764 on Build GRCh38
rs550892764 Chr.19: 7119943-7119943 on Build GRCh38
rs1004204888 Chr.19: 7119726-7119726 on Build GRCh38
rs41362249 Chr.19: 7119577-7119577 on Build GRCh38
rs11672032 Chr.19: 7119638-7119638 on Build GRCh38
rs150182633 Chr.19: 7119639-7119642 on Build GRCh38
rs72282488 Chr.19: 7119626-7119627 on Build GRCh38
rs1028082164 Chr.19: 7119724-7119725 on Build GRCh38
rs386806417 Chr.19: 7119762-7119764 on Build GRCh38
rs560658631 Chr.19: 7119701-7119701 on Build GRCh38
rs973898667 Chr.19: 7119646-7119646 on Build GRCh38

View Full Product Details