Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 139352503-139353078 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPS22
Entrez Gene ID
56945
Gene Name
mitochondrial ribosomal protein S22
Gene Aliases
C3orf5, COXPD5, GIBT, GK002, MRP-S22, RPMS22
UniGene
Hs.745001
Gene Chromosome Location
Chr.3: 139343994-139357129 on Build GRCh38
Transcripts
NM_020191.2 XM_005247640.2 XM_006713703.3 XM_011512995.2 XM_011512996.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6241790 c.806G>T p.G269V Chr.3: 139352720-139352720 on Build GRCh38
1727626 c.757_760delATAG p.D254fs*22 Chr.3: 139352671-139352674 on Build GRCh38
3944929 c.798A>T p.R266S Chr.3: 139352712-139352712 on Build GRCh38
4433682 c.871A>G p.R291G Chr.3: 139352785-139352785 on Build GRCh38
6393525 c.741C>A p.H247Q Chr.3: 139352655-139352655 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1001170764 Chr.3: 139352975-139352975 on Build GRCh38
rs756246802 Chr.3: 139352600-139352600 on Build GRCh38
rs540969115 Chr.3: 139352907-139352907 on Build GRCh38
rs1040419934 Chr.3: 139353051-139353051 on Build GRCh38
rs565139322 Chr.3: 139352878-139352878 on Build GRCh38
rs959378756 Chr.3: 139352881-139352881 on Build GRCh38
rs943371654 Chr.3: 139352993-139352998 on Build GRCh38
rs760807808 Chr.3: 139352848-139352848 on Build GRCh38
rs761121831 Chr.3: 139352672-139352672 on Build GRCh38
rs940926121 Chr.3: 139352948-139352948 on Build GRCh38
rs569698641 Chr.3: 139353010-139353010 on Build GRCh38
rs149674843 Chr.3: 139352681-139352681 on Build GRCh38
rs757552912 Chr.3: 139352720-139352720 on Build GRCh38
rs922090435 Chr.3: 139352605-139352605 on Build GRCh38
rs769362033 Chr.3: 139352674-139352674 on Build GRCh38
rs371823847 Chr.3: 139352817-139352817 on Build GRCh38
rs763877379 Chr.3: 139352797-139352798 on Build GRCh38
rs868695898 Chr.3: 139352671-139352674 on Build GRCh38
rs527578218 Chr.3: 139352883-139352883 on Build GRCh38
rs771057342 Chr.3: 139352631-139352631 on Build GRCh38
rs779313162 Chr.3: 139352639-139352639 on Build GRCh38
rs895539529 Chr.3: 139352981-139352981 on Build GRCh38
rs921983567 Chr.3: 139352901-139352901 on Build GRCh38
rs778869983 Chr.3: 139352531-139352532 on Build GRCh38
rs758551354 Chr.3: 139352835-139352835 on Build GRCh38
rs1000515840 Chr.3: 139352847-139352847 on Build GRCh38
rs903049773 Chr.3: 139353039-139353039 on Build GRCh38
rs1020856188 Chr.3: 139352818-139352818 on Build GRCh38
rs147739245 Chr.3: 139352701-139352701 on Build GRCh38
rs369215331 Chr.3: 139353003-139353004 on Build GRCh38
rs777662595 Chr.3: 139352609-139352609 on Build GRCh38
rs1001683797 Chr.3: 139352702-139352702 on Build GRCh38
rs1000515043 Chr.3: 139353059-139353059 on Build GRCh38
rs760887292 Chr.3: 139352793-139352794 on Build GRCh38
rs751448517 Chr.3: 139352688-139352688 on Build GRCh38
rs972216521 Chr.3: 139352964-139352964 on Build GRCh38
rs755944286 Chr.3: 139352712-139352712 on Build GRCh38
rs549585619 Chr.3: 139352958-139352958 on Build GRCh38
rs35892922 Chr.3: 139352720-139352721 on Build GRCh38
rs919494295 Chr.3: 139352974-139352975 on Build GRCh38
rs991890247 Chr.3: 139352932-139352932 on Build GRCh38
rs772611070 Chr.3: 139352660-139352660 on Build GRCh38
rs865833631 Chr.3: 139352537-139352537 on Build GRCh38
rs72982916 Chr.3: 139352584-139352584 on Build GRCh38
rs114416136 Chr.3: 139352824-139352824 on Build GRCh38
rs1051857046 Chr.3: 139352931-139352931 on Build GRCh38
rs560734613 Chr.3: 139352925-139352925 on Build GRCh38
rs767967295 Chr.3: 139352655-139352656 on Build GRCh38
rs146694653 Chr.3: 139352786-139352786 on Build GRCh38
rs772835214 Chr.3: 139352779-139352779 on Build GRCh38
rs759211223 Chr.3: 139352788-139352788 on Build GRCh38
rs767431419 Chr.3: 139352791-139352791 on Build GRCh38
rs1037990416 Chr.3: 139352957-139352957 on Build GRCh38
rs35952021 Chr.3: 139352699-139352700 on Build GRCh38
rs367552247 Chr.3: 139352733-139352733 on Build GRCh38
rs749534978 Chr.3: 139352620-139352620 on Build GRCh38
rs750734760 Chr.3: 139353048-139353048 on Build GRCh38
rs757316626 Chr.3: 139352820-139352820 on Build GRCh38
rs367924368 Chr.3: 139352680-139352680 on Build GRCh38
rs759370599 Chr.3: 139352691-139352691 on Build GRCh38
rs765934394 Chr.3: 139352687-139352687 on Build GRCh38
rs767992368 Chr.3: 139352982-139352982 on Build GRCh38
rs529866896 Chr.3: 139352934-139352934 on Build GRCh38
rs760001673 Chr.3: 139352602-139352604 on Build GRCh38
rs933450038 Chr.3: 139352638-139352638 on Build GRCh38
rs372605627 Chr.3: 139352749-139352749 on Build GRCh38
rs142654059 Chr.3: 139352760-139352760 on Build GRCh38
rs750657539 Chr.3: 139352831-139352831 on Build GRCh38
rs764689086 Chr.3: 139352802-139352802 on Build GRCh38
rs762299963 Chr.3: 139352713-139352713 on Build GRCh38
rs908073434 Chr.3: 139352945-139352945 on Build GRCh38
rs779079851 Chr.3: 139352725-139352725 on Build GRCh38
rs775852045 Chr.3: 139352783-139352784 on Build GRCh38
rs562973489 Chr.3: 139352664-139352664 on Build GRCh38
rs780462576 Chr.3: 139352839-139352839 on Build GRCh38
rs975149395 Chr.3: 139352887-139352887 on Build GRCh38
rs780340192 Chr.3: 139352753-139352753 on Build GRCh38
rs529406629 Chr.3: 139352782-139352782 on Build GRCh38
rs775354826 Chr.3: 139352793-139352793 on Build GRCh38
rs140631494 Chr.3: 139352655-139352655 on Build GRCh38
rs1055875463 Chr.3: 139352976-139352976 on Build GRCh38
rs769037679 Chr.3: 139352767-139352767 on Build GRCh38
rs752701558 Chr.3: 139352707-139352707 on Build GRCh38
rs760726711 Chr.3: 139352797-139352797 on Build GRCh38
rs576476643 Chr.3: 139352783-139352783 on Build GRCh38
rs933417491 Chr.3: 139352930-139352930 on Build GRCh38
rs863224077 Chr.3: 139352682-139352683 on Build GRCh38
rs748724919 Chr.3: 139352781-139352781 on Build GRCh38
rs530849638 Chr.3: 139352531-139352531 on Build GRCh38

View Full Product Details