Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 45532763-45533293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MARCH8
Entrez Gene ID
220972
Gene Name
membrane associated ring-CH-type finger 8
Gene Aliases
CMIR, MARCH-VIII, MIR, RNF178, c-MIR
UniGene
Hs.499489
Gene Chromosome Location
Chr.10: 45454585-45594907 on Build GRCh38
Transcripts
NM_001002266.2 NM_001282866.1 NM_145021.5 XM_005271804.2 XM_006717704.2 XM_011539492.2 XM_011539493.2 XM_011539494.2 XM_011539495.1 XM_017015894.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6708266 c.7A>G p.M3V Chr.10: 45533205-45533205 on Build GRCh38
5786331 c.47C>G p.A16G Chr.10: 45533165-45533165 on Build GRCh38
1347878 c.5G>A p.S2N Chr.10: 45533207-45533207 on Build GRCh38
4489607 c.34C>T p.P12S Chr.10: 45533178-45533178 on Build GRCh38
1243789 c.20A>G p.Q7R Chr.10: 45533192-45533192 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs894638098 Chr.10: 45532897-45532897 on Build GRCh38
rs759277438 Chr.10: 45532989-45532989 on Build GRCh38
rs761950497 Chr.10: 45533236-45533236 on Build GRCh38
rs764422518 Chr.10: 45533106-45533106 on Build GRCh38
rs945546340 Chr.10: 45532887-45532887 on Build GRCh38
rs902861498 Chr.10: 45532852-45532852 on Build GRCh38
rs757454430 Chr.10: 45533115-45533115 on Build GRCh38
rs918156679 Chr.10: 45533112-45533112 on Build GRCh38
rs927093968 Chr.10: 45533063-45533063 on Build GRCh38
rs765496356 Chr.10: 45533243-45533243 on Build GRCh38
rs775326712 Chr.10: 45533103-45533103 on Build GRCh38
rs776431896 Chr.10: 45533234-45533234 on Build GRCh38
rs557096388 Chr.10: 45533204-45533204 on Build GRCh38
rs941725105 Chr.10: 45532795-45532795 on Build GRCh38
rs759482644 Chr.10: 45533081-45533081 on Build GRCh38
rs778399129 Chr.10: 45533199-45533199 on Build GRCh38
rs538999551 Chr.10: 45533190-45533190 on Build GRCh38
rs945598583 Chr.10: 45533133-45533133 on Build GRCh38
rs938375346 Chr.10: 45532831-45532831 on Build GRCh38
rs142329756 Chr.10: 45533188-45533188 on Build GRCh38
rs1016064066 Chr.10: 45533009-45533009 on Build GRCh38
rs979820661 Chr.10: 45533062-45533062 on Build GRCh38
rs1039510579 Chr.10: 45533205-45533205 on Build GRCh38
rs201435975 Chr.10: 45533162-45533162 on Build GRCh38
rs538819401 Chr.10: 45532986-45532986 on Build GRCh38
rs935749503 Chr.10: 45532876-45532876 on Build GRCh38
rs1006303186 Chr.10: 45532992-45532992 on Build GRCh38
rs759044524 Chr.10: 45533147-45533147 on Build GRCh38
rs548327042 Chr.10: 45532940-45532940 on Build GRCh38
rs774055028 Chr.10: 45533248-45533248 on Build GRCh38
rs182679373 Chr.10: 45532899-45532899 on Build GRCh38
rs922478952 Chr.10: 45532798-45532798 on Build GRCh38
rs1054654559 Chr.10: 45532893-45532893 on Build GRCh38
rs975347641 Chr.10: 45532796-45532796 on Build GRCh38
rs997550193 Chr.10: 45532988-45532988 on Build GRCh38
rs111936295 Chr.10: 45533235-45533235 on Build GRCh38
rs1041380514 Chr.10: 45532809-45532809 on Build GRCh38
rs552045759 Chr.10: 45533064-45533064 on Build GRCh38
rs776534447 Chr.10: 45533245-45533246 on Build GRCh38
rs368170309 Chr.10: 45533098-45533098 on Build GRCh38
rs748765552 Chr.10: 45533130-45533130 on Build GRCh38
rs562576446 Chr.10: 45532892-45532892 on Build GRCh38
rs908951545 Chr.10: 45532793-45532793 on Build GRCh38
rs1000867834 Chr.10: 45532910-45532910 on Build GRCh38
rs760445287 Chr.10: 45533105-45533105 on Build GRCh38
rs935701414 Chr.10: 45533229-45533229 on Build GRCh38
rs371554606 Chr.10: 45533217-45533217 on Build GRCh38
rs768361217 Chr.10: 45533231-45533231 on Build GRCh38
rs1057107701 Chr.10: 45532843-45532843 on Build GRCh38
rs765248479 Chr.10: 45533127-45533127 on Build GRCh38
rs993724631 Chr.10: 45532962-45532962 on Build GRCh38
rs907441925 Chr.10: 45532948-45532948 on Build GRCh38
rs77833583 Chr.10: 45533109-45533109 on Build GRCh38
rs748913191 Chr.10: 45533195-45533195 on Build GRCh38
rs1018139487 Chr.10: 45533002-45533002 on Build GRCh38
rs771474686 Chr.10: 45533088-45533088 on Build GRCh38
rs760500344 Chr.10: 45533230-45533230 on Build GRCh38
rs140989641 Chr.10: 45533151-45533151 on Build GRCh38
rs147834018 Chr.10: 45533017-45533017 on Build GRCh38
rs188118859 Chr.10: 45533004-45533004 on Build GRCh38
rs58437785 Chr.10: 45533250-45533250 on Build GRCh38
rs766960475 Chr.10: 45533056-45533066 on Build GRCh38
rs376819028 Chr.10: 45532807-45532807 on Build GRCh38
rs752143757 Chr.10: 45533261-45533261 on Build GRCh38
rs770557064 Chr.10: 45533196-45533196 on Build GRCh38
rs1026260662 Chr.10: 45532974-45532974 on Build GRCh38
rs775381719 Chr.10: 45533228-45533228 on Build GRCh38
rs562055549 Chr.10: 45533088-45533089 on Build GRCh38

View Full Product Details