Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 105029505-105030020 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP1
Entrez Gene ID
834
Gene Name
caspase 1
Gene Aliases
ICE, IL1BC, P45
UniGene
Hs.2490
Gene Chromosome Location
Chr.11: 105025508-105035591 on Build GRCh38
Transcripts
NM_001223.4 NM_001257118.2 NM_001257119.2 NM_033292.3 NM_033293.3 NM_033294.3 NM_033295.3 XM_017018393.1 XM_017018394.1 XM_017018395.1 XM_017018396.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7099439 c.667G>A p.E223K Chr.11: 105029860-105029860 on Build GRCh38
2109178 c.689C>T p.T230M Chr.11: 105029838-105029838 on Build GRCh38
2109175 c.771A>C p.Q257H Chr.11: 105029756-105029756 on Build GRCh38
4017427 c.707C>T p.S236F Chr.11: 105029820-105029820 on Build GRCh38
5896998 c.761A>T p.D254V Chr.11: 105029766-105029766 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1028235018 Chr.11: 105029716-105029716 on Build GRCh38
rs755460067 Chr.11: 105029729-105029729 on Build GRCh38
rs775065436 Chr.11: 105029771-105029771 on Build GRCh38
rs150630061 Chr.11: 105029639-105029639 on Build GRCh38
rs773709056 Chr.11: 105029659-105029659 on Build GRCh38
rs532657275 Chr.11: 105029868-105029868 on Build GRCh38
rs746553833 Chr.11: 105029946-105029946 on Build GRCh38
rs889690431 Chr.11: 105029727-105029727 on Build GRCh38
rs768758243 Chr.11: 105029611-105029611 on Build GRCh38
rs781661571 Chr.11: 105029735-105029735 on Build GRCh38
rs751791304 Chr.11: 105029725-105029725 on Build GRCh38
rs368643201 Chr.11: 105030000-105030000 on Build GRCh38
rs760403802 Chr.11: 105029668-105029668 on Build GRCh38
rs927529135 Chr.11: 105029574-105029574 on Build GRCh38
rs533869400 Chr.11: 105029588-105029588 on Build GRCh38
rs771556294 Chr.11: 105029762-105029762 on Build GRCh38
rs202211771 Chr.11: 105029723-105029723 on Build GRCh38
rs760320450 Chr.11: 105029780-105029780 on Build GRCh38
rs759033477 Chr.11: 105029665-105029665 on Build GRCh38
rs756607916 Chr.11: 105029924-105029924 on Build GRCh38
rs192726653 Chr.11: 105029912-105029912 on Build GRCh38
rs371852986 Chr.11: 105029650-105029650 on Build GRCh38
rs749826539 Chr.11: 105029930-105029930 on Build GRCh38
rs1044697092 Chr.11: 105029565-105029565 on Build GRCh38
rs370041750 Chr.11: 105029824-105029824 on Build GRCh38
rs501192 Chr.11: 105029658-105029658 on Build GRCh38
rs753843714 Chr.11: 105029671-105029671 on Build GRCh38
rs940071727 Chr.11: 105029558-105029558 on Build GRCh38
rs762916735 Chr.11: 105029891-105029891 on Build GRCh38
rs151040610 Chr.11: 105029866-105029866 on Build GRCh38
rs750491910 Chr.11: 105029707-105029707 on Build GRCh38
rs553049622 Chr.11: 105029597-105029597 on Build GRCh38
rs372432084 Chr.11: 105029715-105029715 on Build GRCh38
rs580253 Chr.11: 105029761-105029761 on Build GRCh38
rs938484400 Chr.11: 105029617-105029617 on Build GRCh38
rs375629927 Chr.11: 105029750-105029750 on Build GRCh38
rs764518347 Chr.11: 105029926-105029926 on Build GRCh38
rs936197968 Chr.11: 105029828-105029828 on Build GRCh38
rs773136863 Chr.11: 105029816-105029816 on Build GRCh38
rs139695105 Chr.11: 105029739-105029739 on Build GRCh38
rs375910475 Chr.11: 105029996-105029996 on Build GRCh38
rs754346383 Chr.11: 105029927-105029927 on Build GRCh38
rs769083128 Chr.11: 105029648-105029648 on Build GRCh38
rs980259586 Chr.11: 105029778-105029778 on Build GRCh38
rs200393683 Chr.11: 105029942-105029942 on Build GRCh38
rs1036217986 Chr.11: 105029592-105029592 on Build GRCh38
rs763315917 Chr.11: 105029600-105029600 on Build GRCh38
rs776388930 Chr.11: 105029798-105029798 on Build GRCh38
rs770442760 Chr.11: 105029748-105029748 on Build GRCh38
rs913198275 Chr.11: 105029853-105029853 on Build GRCh38
rs148534697 Chr.11: 105029837-105029837 on Build GRCh38
rs768484046 Chr.11: 105029793-105029793 on Build GRCh38
rs568182657 Chr.11: 105029965-105029965 on Build GRCh38
rs147332704 Chr.11: 105029666-105029666 on Build GRCh38
rs773089353 Chr.11: 105029890-105029890 on Build GRCh38
rs867394035 Chr.11: 105029851-105029851 on Build GRCh38
rs777664723 Chr.11: 105029589-105029589 on Build GRCh38
rs142765993 Chr.11: 105029865-105029865 on Build GRCh38
rs917379947 Chr.11: 105029526-105029526 on Build GRCh38
rs878955041 Chr.11: 105029838-105029838 on Build GRCh38
rs762999654 Chr.11: 105029822-105029822 on Build GRCh38
rs971596037 Chr.11: 105029772-105029772 on Build GRCh38
rs767853360 Chr.11: 105029914-105029914 on Build GRCh38
rs112668107 Chr.11: 105029714-105029714 on Build GRCh38
rs972865406 Chr.11: 105029763-105029763 on Build GRCh38
rs548263477 Chr.11: 105029957-105029957 on Build GRCh38
rs184122618 Chr.11: 105029932-105029932 on Build GRCh38
rs753761002 Chr.11: 105029858-105029858 on Build GRCh38
rs774591643 Chr.11: 105029896-105029896 on Build GRCh38
rs761710665 Chr.11: 105029682-105029682 on Build GRCh38
rs765350445 Chr.11: 105029697-105029697 on Build GRCh38
rs572084223 Chr.11: 105029823-105029823 on Build GRCh38
rs778152581 Chr.11: 105029863-105029863 on Build GRCh38
rs758527710 Chr.11: 105029615-105029615 on Build GRCh38
rs372270304 Chr.11: 105029892-105029892 on Build GRCh38
rs901043353 Chr.11: 105029660-105029660 on Build GRCh38
rs950291452 Chr.11: 105029554-105029554 on Build GRCh38
rs756248172 Chr.11: 105029766-105029766 on Build GRCh38
rs374686096 Chr.11: 105029931-105029931 on Build GRCh38
rs539628873 Chr.11: 105029667-105029667 on Build GRCh38
rs766419133 Chr.11: 105029895-105029895 on Build GRCh38
rs752907821 Chr.11: 105029917-105029917 on Build GRCh38
rs368177280 Chr.11: 105029670-105029670 on Build GRCh38
rs1057364086 Chr.11: 105029992-105029992 on Build GRCh38
rs747943375 Chr.11: 105029884-105029884 on Build GRCh38
rs45617533 Chr.11: 105029808-105029808 on Build GRCh38
rs780276129 Chr.11: 105029634-105029634 on Build GRCh38
rs769615860 Chr.11: 105029887-105029887 on Build GRCh38
rs776226913 Chr.11: 105029882-105029882 on Build GRCh38
rs891884502 Chr.11: 105029640-105029640 on Build GRCh38
rs756477465 Chr.11: 105029860-105029860 on Build GRCh38
rs748660195 Chr.11: 105029653-105029653 on Build GRCh38
rs761547072 Chr.11: 105029800-105029800 on Build GRCh38
rs778717062 Chr.11: 105029878-105029878 on Build GRCh38
rs527708384 Chr.11: 105029988-105029988 on Build GRCh38
rs367752178 Chr.11: 105029614-105029614 on Build GRCh38

View Full Product Details