Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 51063259-51063852 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSRNP2
Entrez Gene ID
81566
Gene Name
cysteine and serine rich nuclear protein 2
Gene Aliases
C12orf2, C12orf22, FAM130A1, PPP1R72, TAIP-12
UniGene
Hs.524425
Gene Chromosome Location
Chr.12: 51061205-51083671 on Build GRCh38
Transcripts
NM_030809.2 NR_045072.1 XM_006719621.2 XM_017019991.1 XM_011538163.2 XM_017019168.1

Gene Symbol
LETMD1
Entrez Gene ID
25875
Gene Name
LETM1 domain containing 1
Gene Aliases
1110019O13Rik, HCCR-1, HCCR-2, HCCR1, HCRR-2
UniGene
Hs.655272
Gene Chromosome Location
Chr.12: 51048230-51068984 on Build GRCh38
Transcripts
NM_030809.2 NR_045072.1 XM_006719621.2 XM_017019991.1 XM_011538163.2 XM_017019168.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4938988 c.1536G>T p.T512T Chr.12: 51063842-51063842 on Build GRCh38
4965994 c.1618C>T p.P540S Chr.12: 51063760-51063760 on Build GRCh38
310376 c.1532G>A p.R511H Chr.12: 51063846-51063846 on Build GRCh38
6573403 c.1597G>A p.E533K Chr.12: 51063781-51063781 on Build GRCh38
4469732 c.1617C>T p.L539L Chr.12: 51063761-51063761 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202000571 Chr.12: 51063740-51063740 on Build GRCh38
rs778785019 Chr.12: 51063736-51063736 on Build GRCh38
rs911083388 Chr.12: 51063310-51063310 on Build GRCh38
rs899892181 Chr.12: 51063321-51063321 on Build GRCh38
rs17229232 Chr.12: 51063451-51063451 on Build GRCh38
rs757512573 Chr.12: 51063759-51063759 on Build GRCh38
rs971283083 Chr.12: 51063635-51063635 on Build GRCh38
rs372354834 Chr.12: 51063741-51063741 on Build GRCh38
rs773313295 Chr.12: 51063660-51063660 on Build GRCh38
rs768574777 Chr.12: 51063563-51063563 on Build GRCh38
rs796949786 Chr.12: 51063516-51063516 on Build GRCh38
rs886436875 Chr.12: 51063338-51063338 on Build GRCh38
rs982609791 Chr.12: 51063799-51063799 on Build GRCh38
rs555295590 Chr.12: 51063409-51063409 on Build GRCh38
rs754791827 Chr.12: 51063768-51063771 on Build GRCh38
rs112998981 Chr.12: 51063631-51063631 on Build GRCh38
rs1033316949 Chr.12: 51063684-51063684 on Build GRCh38
rs746150411 Chr.12: 51063785-51063785 on Build GRCh38
rs377135259 Chr.12: 51063797-51063797 on Build GRCh38
rs895404272 Chr.12: 51063598-51063599 on Build GRCh38
rs755010963 Chr.12: 51063726-51063726 on Build GRCh38
rs967943988 Chr.12: 51063749-51063749 on Build GRCh38
rs957751884 Chr.12: 51063410-51063410 on Build GRCh38
rs977792177 Chr.12: 51063282-51063282 on Build GRCh38
rs933459627 Chr.12: 51063557-51063557 on Build GRCh38
rs1024665990 Chr.12: 51063346-51063346 on Build GRCh38
rs200273456 Chr.12: 51063790-51063790 on Build GRCh38
rs923593203 Chr.12: 51063286-51063286 on Build GRCh38
rs989076969 Chr.12: 51063494-51063494 on Build GRCh38
rs922421150 Chr.12: 51063552-51063552 on Build GRCh38
rs142747009 Chr.12: 51063606-51063606 on Build GRCh38
rs980401482 Chr.12: 51063402-51063402 on Build GRCh38
rs574118624 Chr.12: 51063694-51063694 on Build GRCh38
rs546362351 Chr.12: 51063580-51063580 on Build GRCh38
rs150504099 Chr.12: 51063525-51063525 on Build GRCh38
rs141806451 Chr.12: 51063483-51063483 on Build GRCh38
rs914714328 Chr.12: 51063475-51063475 on Build GRCh38
rs767181790 Chr.12: 51063709-51063709 on Build GRCh38
rs879229864 Chr.12: 51063772-51063772 on Build GRCh38
rs978075838 Chr.12: 51063479-51063479 on Build GRCh38
rs539172699 Chr.12: 51063323-51063323 on Build GRCh38
rs769745834 Chr.12: 51063789-51063789 on Build GRCh38
rs1004170859 Chr.12: 51063605-51063605 on Build GRCh38
rs761401053 Chr.12: 51063703-51063703 on Build GRCh38
rs911155449 Chr.12: 51063504-51063504 on Build GRCh38
rs747136985 Chr.12: 51063778-51063778 on Build GRCh38
rs541292479 Chr.12: 51063470-51063470 on Build GRCh38
rs765963536 Chr.12: 51063720-51063720 on Build GRCh38
rs749899593 Chr.12: 51063717-51063717 on Build GRCh38
rs942549603 Chr.12: 51063311-51063311 on Build GRCh38
rs1037683400 Chr.12: 51063598-51063601 on Build GRCh38
rs989107947 Chr.12: 51063735-51063735 on Build GRCh38
rs970378014 Chr.12: 51063366-51063366 on Build GRCh38
rs936639436 Chr.12: 51063289-51063289 on Build GRCh38
rs769161467 Chr.12: 51063807-51063807 on Build GRCh38
rs944267888 Chr.12: 51063771-51063771 on Build GRCh38
rs527327224 Chr.12: 51063339-51063339 on Build GRCh38
rs1040831204 Chr.12: 51063319-51063319 on Build GRCh38
rs1053609559 Chr.12: 51063309-51063309 on Build GRCh38
rs886922620 Chr.12: 51063602-51063602 on Build GRCh38
rs774653334 Chr.12: 51063817-51063817 on Build GRCh38
rs944048463 Chr.12: 51063484-51063484 on Build GRCh38
rs776482515 Chr.12: 51063615-51063615 on Build GRCh38
rs12304202 Chr.12: 51063593-51063593 on Build GRCh38
rs1006242202 Chr.12: 51063595-51063598 on Build GRCh38
rs148064219 Chr.12: 51063329-51063329 on Build GRCh38
rs199690668 Chr.12: 51063823-51063823 on Build GRCh38
rs541378088 Chr.12: 51063721-51063721 on Build GRCh38
rs749688709 Chr.12: 51063793-51063793 on Build GRCh38
rs1039637936 Chr.12: 51063506-51063506 on Build GRCh38
rs1012441339 Chr.12: 51063416-51063416 on Build GRCh38
rs577113120 Chr.12: 51063340-51063340 on Build GRCh38
rs946346128 Chr.12: 51063476-51063476 on Build GRCh38
rs970536001 Chr.12: 51063464-51063464 on Build GRCh38
rs1048259528 Chr.12: 51063577-51063577 on Build GRCh38
rs959585220 Chr.12: 51063398-51063398 on Build GRCh38
rs139339223 Chr.12: 51063766-51063766 on Build GRCh38
rs1001888788 Chr.12: 51063651-51063651 on Build GRCh38
rs12317980 Chr.12: 51063719-51063719 on Build GRCh38
rs906410209 Chr.12: 51063630-51063630 on Build GRCh38

View Full Product Details