Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 30737326-30737895 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LAPTM5
Entrez Gene ID
7805
Gene Name
lysosomal protein transmembrane 5
Gene Aliases
CLAST6
UniGene
Hs.371021
Gene Chromosome Location
Chr.1: 30732468-30757840 on Build GRCh38
Transcripts
NM_006762.2 XM_011542098.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6126141 c.558G>C p.K186N Chr.1: 30737652-30737652 on Build GRCh38
908106 c.543T>A p.H181Q Chr.1: 30737667-30737667 on Build GRCh38
908105 c.558G>T p.K186N Chr.1: 30737652-30737652 on Build GRCh38
185204 c.561G>T p.M187I Chr.1: 30737649-30737649 on Build GRCh38
6309750 c.555C>A p.I185I Chr.1: 30737655-30737655 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1028386541 Chr.1: 30737509-30737509 on Build GRCh38
rs531767765 Chr.1: 30737768-30737768 on Build GRCh38
rs552879897 Chr.1: 30737493-30737493 on Build GRCh38
rs772801469 Chr.1: 30737649-30737649 on Build GRCh38
rs981110839 Chr.1: 30737405-30737405 on Build GRCh38
rs972139218 Chr.1: 30737382-30737382 on Build GRCh38
rs776216936 Chr.1: 30737611-30737611 on Build GRCh38
rs964519907 Chr.1: 30737400-30737400 on Build GRCh38
rs898009174 Chr.1: 30737832-30737832 on Build GRCh38
rs768726812 Chr.1: 30737570-30737570 on Build GRCh38
rs540518918 Chr.1: 30737582-30737582 on Build GRCh38
rs11541206 Chr.1: 30737604-30737604 on Build GRCh38
rs758807421 Chr.1: 30737664-30737664 on Build GRCh38
rs961009360 Chr.1: 30737459-30737459 on Build GRCh38
rs539370764 Chr.1: 30737835-30737835 on Build GRCh38
rs186250146 Chr.1: 30737767-30737767 on Build GRCh38
rs548457500 Chr.1: 30737363-30737363 on Build GRCh38
rs144620246 Chr.1: 30737631-30737631 on Build GRCh38
rs766873943 Chr.1: 30737669-30737669 on Build GRCh38
rs562081310 Chr.1: 30737800-30737800 on Build GRCh38
rs760302369 Chr.1: 30737706-30737706 on Build GRCh38
rs140232148 Chr.1: 30737595-30737595 on Build GRCh38
rs181459031 Chr.1: 30737425-30737425 on Build GRCh38
rs373967290 Chr.1: 30737717-30737717 on Build GRCh38
rs969760230 Chr.1: 30737836-30737836 on Build GRCh38
rs553922714 Chr.1: 30737396-30737396 on Build GRCh38
rs891480925 Chr.1: 30737381-30737381 on Build GRCh38
rs780226089 Chr.1: 30737819-30737819 on Build GRCh38
rs34359099 Chr.1: 30737705-30737706 on Build GRCh38
rs971155700 Chr.1: 30737634-30737634 on Build GRCh38
rs189831594 Chr.1: 30737806-30737806 on Build GRCh38
rs756332410 Chr.1: 30737730-30737730 on Build GRCh38
rs1004986533 Chr.1: 30737805-30737805 on Build GRCh38
rs979524466 Chr.1: 30737864-30737864 on Build GRCh38
rs1188355 Chr.1: 30737820-30737820 on Build GRCh38
rs925334045 Chr.1: 30737865-30737865 on Build GRCh38
rs763687921 Chr.1: 30737605-30737605 on Build GRCh38
rs111985890 Chr.1: 30737702-30737702 on Build GRCh38
rs996500017 Chr.1: 30737505-30737505 on Build GRCh38
rs996319658 Chr.1: 30737404-30737404 on Build GRCh38
rs758207717 Chr.1: 30737815-30737815 on Build GRCh38
rs764721370 Chr.1: 30737642-30737642 on Build GRCh38
rs1027328319 Chr.1: 30737780-30737780 on Build GRCh38
rs111952312 Chr.1: 30737345-30737345 on Build GRCh38
rs375995678 Chr.1: 30737672-30737672 on Build GRCh38
rs374760602 Chr.1: 30737458-30737458 on Build GRCh38
rs777988961 Chr.1: 30737742-30737742 on Build GRCh38
rs375929465 Chr.1: 30737586-30737586 on Build GRCh38
rs376132681 Chr.1: 30737449-30737450 on Build GRCh38
rs900845640 Chr.1: 30737496-30737496 on Build GRCh38
rs759546359 Chr.1: 30737593-30737593 on Build GRCh38
rs945170069 Chr.1: 30737436-30737436 on Build GRCh38
rs568375383 Chr.1: 30737380-30737380 on Build GRCh38
rs891738962 Chr.1: 30737703-30737703 on Build GRCh38
rs12405718 Chr.1: 30737365-30737365 on Build GRCh38
rs754777044 Chr.1: 30737721-30737722 on Build GRCh38
rs936568111 Chr.1: 30737422-30737422 on Build GRCh38
rs962165434 Chr.1: 30737600-30737600 on Build GRCh38
rs900163148 Chr.1: 30737759-30737759 on Build GRCh38
rs56296001 Chr.1: 30737809-30737809 on Build GRCh38
rs201201887 Chr.1: 30737688-30737688 on Build GRCh38
rs575605013 Chr.1: 30737439-30737439 on Build GRCh38
rs369299830 Chr.1: 30737681-30737681 on Build GRCh38
rs570660561 Chr.1: 30737416-30737416 on Build GRCh38
rs548586906 Chr.1: 30737799-30737799 on Build GRCh38
rs371054508 Chr.1: 30737720-30737720 on Build GRCh38
rs572099767 Chr.1: 30737562-30737562 on Build GRCh38
rs892294483 Chr.1: 30737435-30737435 on Build GRCh38
rs115211698 Chr.1: 30737353-30737353 on Build GRCh38
rs374364535 Chr.1: 30737727-30737727 on Build GRCh38
rs539309193 Chr.1: 30737417-30737417 on Build GRCh38
rs546482766 Chr.1: 30737624-30737624 on Build GRCh38
rs556071686 Chr.1: 30737447-30737448 on Build GRCh38
rs1008537827 Chr.1: 30737384-30737384 on Build GRCh38
rs748257104 Chr.1: 30737583-30737583 on Build GRCh38
rs527438194 Chr.1: 30737544-30737544 on Build GRCh38
rs767288360 Chr.1: 30737588-30737588 on Build GRCh38
rs866914981 Chr.1: 30737678-30737678 on Build GRCh38
rs993589876 Chr.1: 30737630-30737630 on Build GRCh38
rs535063622 Chr.1: 30737464-30737464 on Build GRCh38
rs1052989104 Chr.1: 30737374-30737374 on Build GRCh38
rs1040071905 Chr.1: 30737757-30737757 on Build GRCh38
rs200376170 Chr.1: 30737674-30737674 on Build GRCh38
rs372654193 Chr.1: 30737671-30737671 on Build GRCh38
rs140031902 Chr.1: 30737829-30737830 on Build GRCh38
rs948118046 Chr.1: 30737351-30737351 on Build GRCh38

View Full Product Details