Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 78615713-78616307 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF219
Entrez Gene ID
79596
Gene Name
ring finger protein 219
Gene Aliases
C13orf7
UniGene
Hs.567576
Gene Chromosome Location
Chr.13: 78614286-78659179 on Build GRCh38
Transcripts
NM_024546.3 XM_011535225.1 XM_017020743.1 NR_047001.1

Gene Symbol
RNF219-AS1
Entrez Gene ID
100874222
Gene Name
RNF219 antisense RNA 1
Gene Aliases
POU4F1-AS1
UniGene
Hs.493062
Gene Chromosome Location
Chr.13: 78054855-78617325 on Build GRCh38
Transcripts
NM_024546.3 XM_011535225.1 XM_017020743.1 NR_047001.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1223986 c.1939C>T p.Q647* Chr.13: 78615822-78615822 on Build GRCh38
6767829 c.1464A>G p.I488M Chr.13: 78616297-78616297 on Build GRCh38
948752 c.1508C>G p.S503* Chr.13: 78616253-78616253 on Build GRCh38
948751 c.1538C>A p.S513Y Chr.13: 78616223-78616223 on Build GRCh38
5774189 c.1593T>C p.D531D Chr.13: 78616168-78616168 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1053820980 Chr.13: 78615940-78615940 on Build GRCh38
rs376347500 Chr.13: 78615919-78615919 on Build GRCh38
rs758938602 Chr.13: 78615767-78615767 on Build GRCh38
rs750489768 Chr.13: 78616061-78616061 on Build GRCh38
rs546639817 Chr.13: 78616042-78616042 on Build GRCh38
rs934223888 Chr.13: 78616183-78616183 on Build GRCh38
rs77899903 Chr.13: 78616196-78616196 on Build GRCh38
rs758742696 Chr.13: 78615862-78615862 on Build GRCh38
rs141239893 Chr.13: 78615744-78615744 on Build GRCh38
rs750012541 Chr.13: 78615749-78615749 on Build GRCh38
rs775183645 Chr.13: 78616263-78616263 on Build GRCh38
rs199596705 Chr.13: 78616200-78616200 on Build GRCh38
rs761617267 Chr.13: 78615814-78615815 on Build GRCh38
rs149986119 Chr.13: 78615944-78615944 on Build GRCh38
rs976409668 Chr.13: 78615789-78615789 on Build GRCh38
rs769997046 Chr.13: 78615805-78615805 on Build GRCh38
rs147894314 Chr.13: 78616084-78616084 on Build GRCh38
rs762122555 Chr.13: 78616264-78616264 on Build GRCh38
rs778296738 Chr.13: 78615877-78615877 on Build GRCh38
rs370680484 Chr.13: 78616184-78616184 on Build GRCh38
rs776653573 Chr.13: 78615807-78615807 on Build GRCh38
rs762186632 Chr.13: 78616175-78616175 on Build GRCh38
rs750288465 Chr.13: 78616103-78616103 on Build GRCh38
rs776119609 Chr.13: 78616245-78616245 on Build GRCh38
rs776327321 Chr.13: 78616122-78616122 on Build GRCh38
rs746061496 Chr.13: 78615790-78615790 on Build GRCh38
rs747936802 Chr.13: 78616089-78616089 on Build GRCh38
rs770889443 Chr.13: 78615780-78615780 on Build GRCh38
rs766072716 Chr.13: 78615996-78615996 on Build GRCh38
rs780343005 Chr.13: 78615968-78615971 on Build GRCh38
rs906797568 Chr.13: 78616004-78616004 on Build GRCh38
rs1005722181 Chr.13: 78616038-78616038 on Build GRCh38
rs957240632 Chr.13: 78616242-78616242 on Build GRCh38
rs910266944 Chr.13: 78616063-78616063 on Build GRCh38
rs954109961 Chr.13: 78616171-78616171 on Build GRCh38
rs747144084 Chr.13: 78615772-78615772 on Build GRCh38
rs753085246 Chr.13: 78615861-78615861 on Build GRCh38
rs1010991381 Chr.13: 78615916-78615916 on Build GRCh38
rs550613447 Chr.13: 78616277-78616277 on Build GRCh38
rs774607490 Chr.13: 78616174-78616174 on Build GRCh38
rs370212589 Chr.13: 78616034-78616034 on Build GRCh38
rs770792919 Chr.13: 78616239-78616239 on Build GRCh38
rs181675982 Chr.13: 78615841-78615841 on Build GRCh38
rs147666396 Chr.13: 78615997-78615997 on Build GRCh38
rs267603860 Chr.13: 78615838-78615838 on Build GRCh38
rs779160626 Chr.13: 78615754-78615754 on Build GRCh38
rs766553709 Chr.13: 78616197-78616197 on Build GRCh38
rs140845041 Chr.13: 78616002-78616002 on Build GRCh38
rs373484257 Chr.13: 78615782-78615782 on Build GRCh38
rs761426697 Chr.13: 78615842-78615842 on Build GRCh38
rs376608775 Chr.13: 78616079-78616079 on Build GRCh38
rs200737834 Chr.13: 78616199-78616199 on Build GRCh38
rs769747677 Chr.13: 78615828-78615828 on Build GRCh38
rs780373018 Chr.13: 78615929-78615929 on Build GRCh38
rs750909505 Chr.13: 78616194-78616194 on Build GRCh38
rs756195575 Chr.13: 78616065-78616065 on Build GRCh38
rs533232798 Chr.13: 78616221-78616221 on Build GRCh38
rs771979509 Chr.13: 78615965-78615965 on Build GRCh38
rs760250533 Chr.13: 78615854-78615854 on Build GRCh38
rs772630135 Chr.13: 78615977-78615977 on Build GRCh38
rs1029305297 Chr.13: 78616126-78616126 on Build GRCh38
rs749517299 Chr.13: 78616067-78616067 on Build GRCh38
rs774407819 Chr.13: 78615837-78615837 on Build GRCh38
rs369495978 Chr.13: 78616019-78616019 on Build GRCh38
rs577036423 Chr.13: 78615834-78615834 on Build GRCh38
rs759196346 Chr.13: 78616131-78616131 on Build GRCh38
rs761670452 Chr.13: 78615952-78615952 on Build GRCh38
rs267603861 Chr.13: 78615839-78615839 on Build GRCh38
rs749879342 Chr.13: 78616088-78616088 on Build GRCh38
rs150803862 Chr.13: 78615851-78615851 on Build GRCh38
rs202005539 Chr.13: 78616195-78616195 on Build GRCh38
rs752061166 Chr.13: 78615892-78615892 on Build GRCh38
rs572904589 Chr.13: 78615857-78615857 on Build GRCh38
rs200280997 Chr.13: 78615852-78615852 on Build GRCh38
rs35223649 Chr.13: 78616262-78616262 on Build GRCh38
rs761268616 Chr.13: 78616281-78616281 on Build GRCh38
rs764799627 Chr.13: 78616145-78616145 on Build GRCh38
rs934255111 Chr.13: 78615823-78615824 on Build GRCh38
rs747944263 Chr.13: 78616206-78616206 on Build GRCh38
rs746544043 Chr.13: 78616111-78616111 on Build GRCh38
rs575630386 Chr.13: 78615984-78615984 on Build GRCh38
rs748446637 Chr.13: 78615755-78615755 on Build GRCh38
rs768455285 Chr.13: 78616147-78616147 on Build GRCh38
rs199656501 Chr.13: 78615922-78615922 on Build GRCh38
rs745338657 Chr.13: 78616260-78616260 on Build GRCh38
rs767919065 Chr.13: 78616272-78616272 on Build GRCh38
rs755427894 Chr.13: 78615752-78615752 on Build GRCh38
rs139175947 Chr.13: 78615943-78615943 on Build GRCh38
rs1002431007 Chr.13: 78616011-78616011 on Build GRCh38
rs558104809 Chr.13: 78615947-78615947 on Build GRCh38
rs778207104 Chr.13: 78615769-78615769 on Build GRCh38
rs144703781 Chr.13: 78616060-78616060 on Build GRCh38
rs773181890 Chr.13: 78616105-78616105 on Build GRCh38
rs557120118 Chr.13: 78615833-78615833 on Build GRCh38
rs537026913 Chr.13: 78615738-78615738 on Build GRCh38
rs529259808 Chr.13: 78616044-78616044 on Build GRCh38
rs760239074 Chr.13: 78615953-78615953 on Build GRCh38
rs865795942 Chr.13: 78616030-78616033 on Build GRCh38
rs758184483 Chr.13: 78616217-78616217 on Build GRCh38
rs763472399 Chr.13: 78616028-78616028 on Build GRCh38
rs757411445 Chr.13: 78615910-78615910 on Build GRCh38
rs905284196 Chr.13: 78615930-78615930 on Build GRCh38
rs770427159 Chr.13: 78616112-78616112 on Build GRCh38
rs200093039 Chr.13: 78616205-78616205 on Build GRCh38
rs771927875 Chr.13: 78616090-78616090 on Build GRCh38
rs762794710 Chr.13: 78615817-78615817 on Build GRCh38
rs769938395 Chr.13: 78615923-78615923 on Build GRCh38
rs775428065 Chr.13: 78615808-78615808 on Build GRCh38
rs777642340 Chr.13: 78616218-78616218 on Build GRCh38
rs998265436 Chr.13: 78616118-78616118 on Build GRCh38

View Full Product Details