Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6634056-6634641 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC11
Entrez Gene ID
55735
Gene Name
DnaJ heat shock protein family (Hsp40) member C11
Gene Aliases
dJ126A5.1
UniGene
Hs.462640
Gene Chromosome Location
Chr.1: 6634168-6701906 on Build GRCh38
Transcripts
NM_018198.3 XM_017001745.1 NM_138350.3 XM_005263532.3 XM_011542400.2 XM_017002761.1

Gene Symbol
THAP3
Entrez Gene ID
90326
Gene Name
THAP domain containing 3
Gene Aliases
-
UniGene
Hs.689006
Gene Chromosome Location
Chr.1: 6624645-6635586 on Build GRCh38
Transcripts
NM_018198.3 XM_017001745.1 NM_138350.3 XM_005263532.3 XM_011542400.2 XM_017002761.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7133941 c.970C>T p.R324W Chr.1: 6634555-6634555 on Build GRCh38
4144164 c.505G>T p.A169S Chr.1: 6634065-6634065 on Build GRCh38
6276105 c.520G>T p.E174* Chr.1: 6634080-6634080 on Build GRCh38
6293084 c.500G>A p.R167H Chr.1: 6634060-6634060 on Build GRCh38
7267020 c.979C>T p.R327C Chr.1: 6634546-6634546 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761925029 Chr.1: 6634087-6634089 on Build GRCh38
rs17692 Chr.1: 6634454-6634454 on Build GRCh38
rs1018895500 Chr.1: 6634385-6634385 on Build GRCh38
rs764327678 Chr.1: 6634522-6634522 on Build GRCh38
rs1025383676 Chr.1: 6634229-6634229 on Build GRCh38
rs767361765 Chr.1: 6634096-6634096 on Build GRCh38
rs760352886 Chr.1: 6634567-6634567 on Build GRCh38
rs949927083 Chr.1: 6634299-6634299 on Build GRCh38
rs772997373 Chr.1: 6634622-6634622 on Build GRCh38
rs138994365 Chr.1: 6634458-6634458 on Build GRCh38
rs775558318 Chr.1: 6634575-6634575 on Build GRCh38
rs774263533 Chr.1: 6634506-6634506 on Build GRCh38
rs145157732 Chr.1: 6634457-6634457 on Build GRCh38
rs773018245 Chr.1: 6634565-6634565 on Build GRCh38
rs756021835 Chr.1: 6634114-6634115 on Build GRCh38
rs891822833 Chr.1: 6634319-6634319 on Build GRCh38
rs11553408 Chr.1: 6634277-6634277 on Build GRCh38
rs954909705 Chr.1: 6634425-6634425 on Build GRCh38
rs543786044 Chr.1: 6634189-6634189 on Build GRCh38
rs368884220 Chr.1: 6634302-6634302 on Build GRCh38
rs527251312 Chr.1: 6634281-6634281 on Build GRCh38
rs988740881 Chr.1: 6634290-6634290 on Build GRCh38
rs373662285 Chr.1: 6634251-6634251 on Build GRCh38
rs1010306320 Chr.1: 6634182-6634182 on Build GRCh38
rs545120514 Chr.1: 6634215-6634215 on Build GRCh38
rs891256885 Chr.1: 6634325-6634325 on Build GRCh38
rs12727844 Chr.1: 6634147-6634147 on Build GRCh38
rs146601847 Chr.1: 6634197-6634197 on Build GRCh38
rs986088796 Chr.1: 6634214-6634214 on Build GRCh38
rs969052560 Chr.1: 6634272-6634272 on Build GRCh38
rs548979382 Chr.1: 6634324-6634324 on Build GRCh38
rs755163069 Chr.1: 6634102-6634102 on Build GRCh38
rs1021510611 Chr.1: 6634415-6634415 on Build GRCh38
rs757022375 Chr.1: 6634268-6634268 on Build GRCh38
rs1043706 Chr.1: 6634434-6634434 on Build GRCh38
rs778348115 Chr.1: 6634615-6634615 on Build GRCh38
rs113571259 Chr.1: 6634574-6634574 on Build GRCh38
rs755371320 Chr.1: 6634437-6634437 on Build GRCh38
rs370946581 Chr.1: 6634546-6634546 on Build GRCh38
rs763333669 Chr.1: 6634588-6634588 on Build GRCh38
rs750202031 Chr.1: 6634512-6634512 on Build GRCh38
rs956116193 Chr.1: 6634169-6634169 on Build GRCh38
rs780539536 Chr.1: 6634612-6634612 on Build GRCh38
rs935955156 Chr.1: 6634306-6634306 on Build GRCh38
rs761573686 Chr.1: 6634087-6634087 on Build GRCh38
rs774538944 Chr.1: 6634080-6634082 on Build GRCh38
rs969042729 Chr.1: 6634336-6634336 on Build GRCh38
rs905783159 Chr.1: 6634305-6634305 on Build GRCh38
rs1039594031 Chr.1: 6634312-6634312 on Build GRCh38
rs14747 Chr.1: 6634267-6634267 on Build GRCh38
rs962945995 Chr.1: 6634419-6634419 on Build GRCh38
rs758525463 Chr.1: 6634516-6634516 on Build GRCh38
rs567421358 Chr.1: 6634332-6634332 on Build GRCh38
rs958297266 Chr.1: 6634387-6634387 on Build GRCh38
rs1054373260 Chr.1: 6634313-6634313 on Build GRCh38
rs917046261 Chr.1: 6634295-6634295 on Build GRCh38
rs781731643 Chr.1: 6634494-6634494 on Build GRCh38
rs1018538904 Chr.1: 6634216-6634216 on Build GRCh38
rs6804 Chr.1: 6634514-6634514 on Build GRCh38
rs757801172 Chr.1: 6634598-6634598 on Build GRCh38
rs532794371 Chr.1: 6634208-6634208 on Build GRCh38
rs751538968 Chr.1: 6634082-6634082 on Build GRCh38
rs750988686 Chr.1: 6634600-6634600 on Build GRCh38
rs1043703 Chr.1: 6634453-6634453 on Build GRCh38
rs988427134 Chr.1: 6634404-6634404 on Build GRCh38
rs759752759 Chr.1: 6634513-6634513 on Build GRCh38
rs1010255317 Chr.1: 6634321-6634321 on Build GRCh38
rs767236783 Chr.1: 6634509-6634509 on Build GRCh38
rs767526516 Chr.1: 6634096-6634098 on Build GRCh38
rs766275053 Chr.1: 6634508-6634508 on Build GRCh38
rs1046980930 Chr.1: 6634504-6634504 on Build GRCh38
rs775026315 Chr.1: 6634497-6634497 on Build GRCh38
rs898451957 Chr.1: 6634314-6634314 on Build GRCh38
rs964693226 Chr.1: 6634236-6634236 on Build GRCh38
rs1032585065 Chr.1: 6634375-6634375 on Build GRCh38
rs761808804 Chr.1: 6634508-6634508 on Build GRCh38
rs1029761541 Chr.1: 6634323-6634323 on Build GRCh38
rs758930243 Chr.1: 6634478-6634478 on Build GRCh38
rs925567707 Chr.1: 6634278-6634278 on Build GRCh38
rs748998390 Chr.1: 6634553-6634553 on Build GRCh38
rs201689224 Chr.1: 6634452-6634453 on Build GRCh38
rs1033145109 Chr.1: 6634161-6634161 on Build GRCh38
rs748875908 Chr.1: 6634614-6634614 on Build GRCh38
rs1031989405 Chr.1: 6634609-6634609 on Build GRCh38
rs914010678 Chr.1: 6634112-6634112 on Build GRCh38
rs376340625 Chr.1: 6634555-6634555 on Build GRCh38
rs771650199 Chr.1: 6634617-6634617 on Build GRCh38
rs374400737 Chr.1: 6634121-6634121 on Build GRCh38
rs79312390 Chr.1: 6634160-6634160 on Build GRCh38
rs895677977 Chr.1: 6634097-6634097 on Build GRCh38
rs910517195 Chr.1: 6634303-6634303 on Build GRCh38
rs752106891 Chr.1: 6634530-6634530 on Build GRCh38
rs753742546 Chr.1: 6634507-6634508 on Build GRCh38
rs772021526 Chr.1: 6634559-6634559 on Build GRCh38
rs746251740 Chr.1: 6634545-6634545 on Build GRCh38
rs778748238 Chr.1: 6634556-6634557 on Build GRCh38
rs762339154 Chr.1: 6634504-6634504 on Build GRCh38
rs747833659 Chr.1: 6634599-6634600 on Build GRCh38
rs59818822 Chr.1: 6634496-6634496 on Build GRCh38
rs531613111 Chr.1: 6634430-6634430 on Build GRCh38
rs781561069 Chr.1: 6634542-6634542 on Build GRCh38
rs754090272 Chr.1: 6634101-6634101 on Build GRCh38
rs1000160269 Chr.1: 6634153-6634153 on Build GRCh38
rs530378430 Chr.1: 6634515-6634515 on Build GRCh38
rs1013355408 Chr.1: 6634262-6634262 on Build GRCh38
rs778390365 Chr.1: 6634486-6634486 on Build GRCh38
rs929025869 Chr.1: 6634271-6634271 on Build GRCh38
rs754796616 Chr.1: 6634530-6634536 on Build GRCh38
rs1001906718 Chr.1: 6634369-6634369 on Build GRCh38
rs905830970 Chr.1: 6634152-6634152 on Build GRCh38
rs566176860 Chr.1: 6634493-6634493 on Build GRCh38
rs756421137 Chr.1: 6634605-6634605 on Build GRCh38
rs757842300 Chr.1: 6634532-6634532 on Build GRCh38
rs779551609 Chr.1: 6634548-6634548 on Build GRCh38
rs995281686 Chr.1: 6634320-6634320 on Build GRCh38
rs767880909 Chr.1: 6634599-6634599 on Build GRCh38
rs751700525 Chr.1: 6634595-6634595 on Build GRCh38
rs935903349 Chr.1: 6634501-6634501 on Build GRCh38
rs752739023 Chr.1: 6634130-6634130 on Build GRCh38
rs764201137 Chr.1: 6634590-6634590 on Build GRCh38
rs766242435 Chr.1: 6634570-6634570 on Build GRCh38
rs750468281 Chr.1: 6634109-6634109 on Build GRCh38
rs768155061 Chr.1: 6634554-6634554 on Build GRCh38

View Full Product Details