Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 50977464-50977902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLK7
Entrez Gene ID
5650
Gene Name
kallikrein related peptidase 7
Gene Aliases
PRSS6, SCCE, hK7
UniGene
Hs.151254
Gene Chromosome Location
Chr.19: 50976479-50984064 on Build GRCh38
Transcripts
NM_001207053.1 NM_001243126.1 NM_005046.3 NM_139277.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
166630 c.646C>T p.Q216* Chr.19: 50977652-50977652 on Build GRCh38
6883584 c.649G>A p.G217S Chr.19: 50977649-50977649 on Build GRCh38
4696645 c.752delA p.K251fs*>3 Chr.19: 50977546-50977546 on Build GRCh38
6696997 c.618G>C p.G206G Chr.19: 50977680-50977680 on Build GRCh38
3716507 c.697G>A p.G233R Chr.19: 50977601-50977601 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1654528 Chr.19: 50977584-50977584 on Build GRCh38
rs1027053305 Chr.19: 50977812-50977812 on Build GRCh38
rs375837932 Chr.19: 50977552-50977552 on Build GRCh38
rs751551701 Chr.19: 50977702-50977702 on Build GRCh38
rs777493571 Chr.19: 50977725-50977725 on Build GRCh38
rs758335462 Chr.19: 50977617-50977617 on Build GRCh38
rs17855561 Chr.19: 50977620-50977620 on Build GRCh38
rs776067181 Chr.19: 50977660-50977660 on Build GRCh38
rs774830711 Chr.19: 50977556-50977556 on Build GRCh38
rs778638425 Chr.19: 50977522-50977522 on Build GRCh38
rs113446404 Chr.19: 50977769-50977769 on Build GRCh38
rs374033717 Chr.19: 50977639-50977639 on Build GRCh38
rs769536810 Chr.19: 50977644-50977644 on Build GRCh38
rs766024916 Chr.19: 50977680-50977680 on Build GRCh38
rs759999024 Chr.19: 50977564-50977564 on Build GRCh38
rs113580433 Chr.19: 50977704-50977704 on Build GRCh38
rs753999943 Chr.19: 50977502-50977502 on Build GRCh38
rs199661764 Chr.19: 50977546-50977546 on Build GRCh38
rs150542901 Chr.19: 50977535-50977535 on Build GRCh38
rs866275891 Chr.19: 50977551-50977551 on Build GRCh38
rs73932697 Chr.19: 50977674-50977674 on Build GRCh38
rs150314873 Chr.19: 50977836-50977836 on Build GRCh38
rs866607709 Chr.19: 50977655-50977655 on Build GRCh38
rs571182395 Chr.19: 50977854-50977854 on Build GRCh38
rs200480032 Chr.19: 50977541-50977541 on Build GRCh38
rs185303262 Chr.19: 50977860-50977860 on Build GRCh38
rs142918110 Chr.19: 50977548-50977548 on Build GRCh38
rs1009889045 Chr.19: 50977505-50977505 on Build GRCh38
rs761045320 Chr.19: 50977664-50977664 on Build GRCh38
rs774919023 Chr.19: 50977647-50977647 on Build GRCh38
rs371407092 Chr.19: 50977709-50977709 on Build GRCh38
rs772516769 Chr.19: 50977728-50977728 on Build GRCh38
rs777957190 Chr.19: 50977501-50977501 on Build GRCh38
rs780812760 Chr.19: 50977631-50977631 on Build GRCh38
rs750203731 Chr.19: 50977593-50977593 on Build GRCh38
rs112736390 Chr.19: 50977573-50977573 on Build GRCh38
rs377551989 Chr.19: 50977581-50977581 on Build GRCh38
rs1654527 Chr.19: 50977726-50977726 on Build GRCh38
rs559982577 Chr.19: 50977835-50977835 on Build GRCh38
rs757050585 Chr.19: 50977516-50977516 on Build GRCh38
rs764180281 Chr.19: 50977671-50977671 on Build GRCh38
rs776670805 Chr.19: 50977673-50977673 on Build GRCh38
rs552179062 Chr.19: 50977852-50977852 on Build GRCh38
rs944298885 Chr.19: 50977649-50977649 on Build GRCh38
rs200130750 Chr.19: 50977708-50977708 on Build GRCh38
rs139349638 Chr.19: 50977540-50977540 on Build GRCh38
rs148515610 Chr.19: 50977675-50977675 on Build GRCh38
rs1000622042 Chr.19: 50977801-50977801 on Build GRCh38
rs572447388 Chr.19: 50977583-50977583 on Build GRCh38
rs181437064 Chr.19: 50977619-50977619 on Build GRCh38
rs112212984 Chr.19: 50977815-50977815 on Build GRCh38
rs760820716 Chr.19: 50977578-50977578 on Build GRCh38
rs772265162 Chr.19: 50977652-50977652 on Build GRCh38
rs552875279 Chr.19: 50977494-50977494 on Build GRCh38
rs760425345 Chr.19: 50977832-50977832 on Build GRCh38
rs112139149 Chr.19: 50977534-50977534 on Build GRCh38
rs189779210 Chr.19: 50977603-50977603 on Build GRCh38
rs758229547 Chr.19: 50977679-50977679 on Build GRCh38
rs749971315 Chr.19: 50977768-50977768 on Build GRCh38
rs140207475 Chr.19: 50977767-50977767 on Build GRCh38
rs1009875122 Chr.19: 50977819-50977819 on Build GRCh38
rs774832270 Chr.19: 50977761-50977761 on Build GRCh38
rs754525282 Chr.19: 50977621-50977621 on Build GRCh38
rs544020708 Chr.19: 50977707-50977707 on Build GRCh38
rs754089866 Chr.19: 50977582-50977582 on Build GRCh38
rs768252949 Chr.19: 50977545-50977545 on Build GRCh38
rs746228119 Chr.19: 50977648-50977648 on Build GRCh38
rs973962058 Chr.19: 50977790-50977790 on Build GRCh38

View Full Product Details