Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 55430951-55431473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LANCL2
Entrez Gene ID
55915
Gene Name
LanC like 2
Gene Aliases
GPR69B, TASP
UniGene
Hs.595384
Gene Chromosome Location
Chr.7: 55365448-55433742 on Build GRCh38
Transcripts
NM_018697.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6874172 c.1267G>A p.G423S Chr.7: 55431234-55431234 on Build GRCh38
1734918 c.1310C>T p.S437L Chr.7: 55431277-55431277 on Build GRCh38
4961666 c.1269C>T p.G423G Chr.7: 55431236-55431236 on Build GRCh38
1090908 c.1341G>A p.S447S Chr.7: 55431308-55431308 on Build GRCh38
5432394 c.1288G>T p.D430Y Chr.7: 55431255-55431255 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs993224818 Chr.7: 55430985-55430985 on Build GRCh38
rs201881002 Chr.7: 55431308-55431308 on Build GRCh38
rs1019746072 Chr.7: 55431405-55431405 on Build GRCh38
rs964561827 Chr.7: 55431424-55431424 on Build GRCh38
rs202032634 Chr.7: 55431309-55431309 on Build GRCh38
rs745949849 Chr.7: 55431181-55431181 on Build GRCh38
rs767444624 Chr.7: 55431220-55431220 on Build GRCh38
rs951617943 Chr.7: 55431066-55431066 on Build GRCh38
rs545699675 Chr.7: 55431314-55431314 on Build GRCh38
rs1044068538 Chr.7: 55431134-55431134 on Build GRCh38
rs112308825 Chr.7: 55430978-55430978 on Build GRCh38
rs909088676 Chr.7: 55431432-55431432 on Build GRCh38
rs528568663 Chr.7: 55431408-55431408 on Build GRCh38
rs541122906 Chr.7: 55431279-55431279 on Build GRCh38
rs748937226 Chr.7: 55431215-55431215 on Build GRCh38
rs549007503 Chr.7: 55431371-55431371 on Build GRCh38
rs984638789 Chr.7: 55431418-55431418 on Build GRCh38
rs180926854 Chr.7: 55430979-55430979 on Build GRCh38
rs765581027 Chr.7: 55431265-55431265 on Build GRCh38
rs781066545 Chr.7: 55431428-55431428 on Build GRCh38
rs375173086 Chr.7: 55431175-55431175 on Build GRCh38
rs140816570 Chr.7: 55431280-55431280 on Build GRCh38
rs2293322 Chr.7: 55431062-55431062 on Build GRCh38
rs775122295 Chr.7: 55431364-55431364 on Build GRCh38
rs898805558 Chr.7: 55431341-55431341 on Build GRCh38
rs747591612 Chr.7: 55431200-55431200 on Build GRCh38
rs750476655 Chr.7: 55431266-55431266 on Build GRCh38
rs145947925 Chr.7: 55431237-55431237 on Build GRCh38
rs1021844827 Chr.7: 55431434-55431434 on Build GRCh38
rs758987037 Chr.7: 55431219-55431219 on Build GRCh38
rs987011507 Chr.7: 55431109-55431109 on Build GRCh38
rs912805008 Chr.7: 55431117-55431117 on Build GRCh38
rs773850906 Chr.7: 55431218-55431218 on Build GRCh38
rs375694504 Chr.7: 55431368-55431368 on Build GRCh38
rs573818801 Chr.7: 55431268-55431268 on Build GRCh38
rs815988 Chr.7: 55431365-55431365 on Build GRCh38
rs576012950 Chr.7: 55431178-55431178 on Build GRCh38
rs34827381 Chr.7: 55431004-55431005 on Build GRCh38
rs914535147 Chr.7: 55431155-55431155 on Build GRCh38
rs538831688 Chr.7: 55431190-55431190 on Build GRCh38
rs948362800 Chr.7: 55431133-55431133 on Build GRCh38
rs768989818 Chr.7: 55431209-55431209 on Build GRCh38
rs75177017 Chr.7: 55431386-55431386 on Build GRCh38
rs184954711 Chr.7: 55431148-55431148 on Build GRCh38
rs760716022 Chr.7: 55431236-55431236 on Build GRCh38
rs369730124 Chr.7: 55431211-55431211 on Build GRCh38
rs78231031 Chr.7: 55431452-55431452 on Build GRCh38
rs190150971 Chr.7: 55431260-55431260 on Build GRCh38
rs1035134784 Chr.7: 55431203-55431203 on Build GRCh38
rs745570031 Chr.7: 55431338-55431338 on Build GRCh38
rs964360094 Chr.7: 55431138-55431138 on Build GRCh38
rs919129927 Chr.7: 55431081-55431081 on Build GRCh38
rs201472149 Chr.7: 55431349-55431349 on Build GRCh38
rs142198607 Chr.7: 55431085-55431085 on Build GRCh38
rs775255624 Chr.7: 55431223-55431223 on Build GRCh38
rs866208013 Chr.7: 55431313-55431313 on Build GRCh38
rs770578092 Chr.7: 55431216-55431216 on Build GRCh38
rs541367026 Chr.7: 55431429-55431429 on Build GRCh38
rs765755881 Chr.7: 55431232-55431232 on Build GRCh38
rs777745347 Chr.7: 55431284-55431284 on Build GRCh38
rs780457501 Chr.7: 55431191-55431191 on Build GRCh38
rs929094538 Chr.7: 55431390-55431390 on Build GRCh38
rs372359047 Chr.7: 55431307-55431307 on Build GRCh38

View Full Product Details