Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 192660134-192660479 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RGS13
Entrez Gene ID
6003
Gene Name
regulator of G-protein signaling 13
Gene Aliases
-
UniGene
Hs.497220
Gene Chromosome Location
Chr.1: 192636138-192660311 on Build GRCh38
Transcripts
NM_002927.4 NM_144766.2

SNP Detail

SNP ID Chromosome Location
rs567589875 Chr.1: 192660443-192660443 on Build GRCh38
rs940758674 Chr.1: 192660236-192660236 on Build GRCh38
rs1055341664 Chr.1: 192660355-192660355 on Build GRCh38
rs1038236168 Chr.1: 192660279-192660279 on Build GRCh38
rs772648844 Chr.1: 192660368-192660368 on Build GRCh38
rs995331973 Chr.1: 192660353-192660353 on Build GRCh38
rs867847594 Chr.1: 192660271-192660271 on Build GRCh38
rs1047838385 Chr.1: 192660371-192660371 on Build GRCh38
rs113632160 Chr.1: 192660249-192660249 on Build GRCh38
rs986449910 Chr.1: 192660229-192660229 on Build GRCh38
rs536220080 Chr.1: 192660447-192660447 on Build GRCh38
rs764446252 Chr.1: 192660352-192660352 on Build GRCh38
rs79868199 Chr.1: 192660195-192660195 on Build GRCh38
rs895079496 Chr.1: 192660360-192660360 on Build GRCh38
rs898321393 Chr.1: 192660316-192660316 on Build GRCh38
rs748580496 Chr.1: 192660262-192660265 on Build GRCh38
rs922190901 Chr.1: 192660164-192660164 on Build GRCh38
rs571395394 Chr.1: 192660268-192660268 on Build GRCh38
rs966345448 Chr.1: 192660412-192660412 on Build GRCh38
rs906128323 Chr.1: 192660454-192660454 on Build GRCh38
rs933654921 Chr.1: 192660223-192660223 on Build GRCh38
rs557249090 Chr.1: 192660424-192660424 on Build GRCh38
rs933538669 Chr.1: 192660169-192660169 on Build GRCh38
rs886484899 Chr.1: 192660374-192660374 on Build GRCh38
rs1032080697 Chr.1: 192660435-192660435 on Build GRCh38
rs942610974 Chr.1: 192660267-192660267 on Build GRCh38
rs117860365 Chr.1: 192660376-192660376 on Build GRCh38

View Full Product Details