Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 41888661-41889231 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2CD2
Entrez Gene ID
25966
Gene Name
C2 calcium dependent domain containing 2
Gene Aliases
C21orf25, C21orf258, TMEM24L
UniGene
Hs.473894
Gene Chromosome Location
Chr.21: 41885110-41953957 on Build GRCh38
Transcripts
NM_015500.1 NM_199050.2 XM_005261109.4 XM_011529522.1 XM_011529523.2 XM_017028315.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1714021 c.2081C>T p.P694L Chr.21: 41889134-41889134 on Build GRCh38
5669128 c.1988G>C p.R663P Chr.21: 41889227-41889227 on Build GRCh38
4476438 c.2069C>T p.A690V Chr.21: 41889146-41889146 on Build GRCh38
324884 c.2023A>G p.K675E Chr.21: 41889192-41889192 on Build GRCh38
3551001 c.2071C>T p.P691S Chr.21: 41889144-41889144 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375297943 Chr.21: 41889088-41889088 on Build GRCh38
rs901723393 Chr.21: 41888731-41888731 on Build GRCh38
rs368007922 Chr.21: 41888869-41888869 on Build GRCh38
rs199638205 Chr.21: 41889150-41889150 on Build GRCh38
rs1024652884 Chr.21: 41888949-41888949 on Build GRCh38
rs202167715 Chr.21: 41889146-41889146 on Build GRCh38
rs1036886787 Chr.21: 41889018-41889018 on Build GRCh38
rs1038295526 Chr.21: 41888922-41888922 on Build GRCh38
rs746198554 Chr.21: 41889191-41889191 on Build GRCh38
rs750388485 Chr.21: 41888993-41888993 on Build GRCh38
rs1039419605 Chr.21: 41888972-41888972 on Build GRCh38
rs984975472 Chr.21: 41889013-41889013 on Build GRCh38
rs759467044 Chr.21: 41889151-41889151 on Build GRCh38
rs958847567 Chr.21: 41888994-41888994 on Build GRCh38
rs962242190 Chr.21: 41889189-41889189 on Build GRCh38
rs746631048 Chr.21: 41889212-41889212 on Build GRCh38
rs771512557 Chr.21: 41889207-41889207 on Build GRCh38
rs538794615 Chr.21: 41888971-41888971 on Build GRCh38
rs184522600 Chr.21: 41888862-41888862 on Build GRCh38
rs972154421 Chr.21: 41889190-41889190 on Build GRCh38
rs117899230 Chr.21: 41888807-41888807 on Build GRCh38
rs139288725 Chr.21: 41889142-41889142 on Build GRCh38
rs746346937 Chr.21: 41889100-41889100 on Build GRCh38
rs772713203 Chr.21: 41889209-41889209 on Build GRCh38
rs758543547 Chr.21: 41889160-41889160 on Build GRCh38
rs536797013 Chr.21: 41888826-41888826 on Build GRCh38
rs55907107 Chr.21: 41889081-41889081 on Build GRCh38
rs757455763 Chr.21: 41889185-41889185 on Build GRCh38
rs892183259 Chr.21: 41889089-41889089 on Build GRCh38
rs766127607 Chr.21: 41889147-41889147 on Build GRCh38
rs1017919450 Chr.21: 41888732-41888732 on Build GRCh38
rs373762862 Chr.21: 41889201-41889201 on Build GRCh38
rs781573696 Chr.21: 41889188-41889188 on Build GRCh38
rs796559808 Chr.21: 41889081-41889082 on Build GRCh38
rs1036367593 Chr.21: 41888725-41888725 on Build GRCh38
rs772837893 Chr.21: 41889141-41889141 on Build GRCh38
rs889042686 Chr.21: 41889046-41889046 on Build GRCh38
rs1001106319 Chr.21: 41888757-41888757 on Build GRCh38
rs377599432 Chr.21: 41889091-41889091 on Build GRCh38
rs2839403 Chr.21: 41889082-41889082 on Build GRCh38
rs956913401 Chr.21: 41888784-41888784 on Build GRCh38
rs780015618 Chr.21: 41889033-41889033 on Build GRCh38
rs1051396976 Chr.21: 41888959-41888959 on Build GRCh38
rs923292376 Chr.21: 41889029-41889029 on Build GRCh38
rs981715539 Chr.21: 41888966-41888966 on Build GRCh38
rs758279179 Chr.21: 41889017-41889017 on Build GRCh38
rs890838541 Chr.21: 41888711-41888711 on Build GRCh38
rs780531926 Chr.21: 41889199-41889199 on Build GRCh38
rs376773156 Chr.21: 41888827-41888827 on Build GRCh38
rs1025185132 Chr.21: 41889110-41889110 on Build GRCh38
rs780768368 Chr.21: 41889130-41889130 on Build GRCh38
rs941977859 Chr.21: 41888899-41888899 on Build GRCh38
rs752752497 Chr.21: 41889157-41889157 on Build GRCh38
rs1050349953 Chr.21: 41889042-41889042 on Build GRCh38
rs929837321 Chr.21: 41888950-41888950 on Build GRCh38
rs760260238 Chr.21: 41889205-41889205 on Build GRCh38
rs771636879 Chr.21: 41889128-41889128 on Build GRCh38
rs918482398 Chr.21: 41888943-41888943 on Build GRCh38
rs1031175147 Chr.21: 41888761-41888762 on Build GRCh38
rs770227317 Chr.21: 41889107-41889107 on Build GRCh38
rs1009150065 Chr.21: 41888937-41888937 on Build GRCh38
rs574263830 Chr.21: 41889181-41889181 on Build GRCh38
rs558890242 Chr.21: 41889016-41889016 on Build GRCh38
rs956475336 Chr.21: 41888875-41888875 on Build GRCh38
rs547737358 Chr.21: 41888787-41888787 on Build GRCh38
rs776023654 Chr.21: 41889127-41889127 on Build GRCh38
rs756539158 Chr.21: 41889193-41889193 on Build GRCh38
rs374271319 Chr.21: 41889104-41889104 on Build GRCh38
rs997433994 Chr.21: 41888736-41888736 on Build GRCh38
rs970835601 Chr.21: 41888954-41888954 on Build GRCh38
rs768211161 Chr.21: 41889092-41889092 on Build GRCh38
rs1050882082 Chr.21: 41888699-41888699 on Build GRCh38
rs765056633 Chr.21: 41889154-41889154 on Build GRCh38

View Full Product Details