Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 28932212-28932776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD19
Entrez Gene ID
930
Gene Name
CD19 molecule
Gene Aliases
B4, CVID3
UniGene
Hs.652262
Gene Chromosome Location
Chr.16: 28931735-28939347 on Build GRCh38
Transcripts
NM_001178098.1 NM_001770.5 XM_006721103.3 XM_011545981.1 XM_017023893.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4059902 c.348G>A p.E116E Chr.16: 28932605-28932605 on Build GRCh38
5384671 c.137C>T p.P46L Chr.16: 28932394-28932394 on Build GRCh38
7232360 c.150G>A p.L50L Chr.16: 28932407-28932407 on Build GRCh38
212712 c.91G>C p.G31R Chr.16: 28932348-28932348 on Build GRCh38
4059901 c.101C>T p.A34V Chr.16: 28932358-28932358 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34059038 Chr.16: 28932496-28932497 on Build GRCh38
rs886037920 Chr.16: 28932413-28932413 on Build GRCh38
rs756836129 Chr.16: 28932545-28932545 on Build GRCh38
rs775942768 Chr.16: 28932335-28932335 on Build GRCh38
rs149157381 Chr.16: 28932468-28932468 on Build GRCh38
rs751791834 Chr.16: 28932543-28932543 on Build GRCh38
rs367989100 Chr.16: 28932328-28932328 on Build GRCh38
rs750521248 Chr.16: 28932530-28932530 on Build GRCh38
rs761387768 Chr.16: 28932338-28932338 on Build GRCh38
rs906793959 Chr.16: 28932376-28932376 on Build GRCh38
rs775660739 Chr.16: 28932488-28932488 on Build GRCh38
rs758555433 Chr.16: 28932531-28932531 on Build GRCh38
rs560539055 Chr.16: 28932258-28932258 on Build GRCh38
rs367858870 Chr.16: 28932573-28932573 on Build GRCh38
rs77213850 Chr.16: 28932263-28932263 on Build GRCh38
rs753138134 Chr.16: 28932367-28932367 on Build GRCh38
rs766423390 Chr.16: 28932621-28932621 on Build GRCh38
rs371575919 Chr.16: 28932611-28932611 on Build GRCh38
rs374119720 Chr.16: 28932313-28932313 on Build GRCh38
rs757328326 Chr.16: 28932401-28932401 on Build GRCh38
rs934181880 Chr.16: 28932683-28932683 on Build GRCh38
rs777186183 Chr.16: 28932515-28932515 on Build GRCh38
rs150438147 Chr.16: 28932418-28932418 on Build GRCh38
rs556621561 Chr.16: 28932278-28932278 on Build GRCh38
rs748821637 Chr.16: 28932560-28932560 on Build GRCh38
rs940100916 Chr.16: 28932293-28932293 on Build GRCh38
rs140445039 Chr.16: 28932404-28932404 on Build GRCh38
rs376909352 Chr.16: 28932626-28932626 on Build GRCh38
rs533648795 Chr.16: 28932582-28932582 on Build GRCh38
rs756779230 Chr.16: 28932644-28932644 on Build GRCh38
rs201308929 Chr.16: 28932516-28932516 on Build GRCh38
rs769405929 Chr.16: 28932464-28932464 on Build GRCh38
rs762741803 Chr.16: 28932355-28932355 on Build GRCh38
rs765272658 Chr.16: 28932526-28932526 on Build GRCh38
rs1003734209 Chr.16: 28932684-28932684 on Build GRCh38
rs756682700 Chr.16: 28932296-28932296 on Build GRCh38
rs773206812 Chr.16: 28932610-28932610 on Build GRCh38
rs761106054 Chr.16: 28932494-28932494 on Build GRCh38
rs566183838 Chr.16: 28932558-28932558 on Build GRCh38
rs145255205 Chr.16: 28932451-28932451 on Build GRCh38
rs922692076 Chr.16: 28932680-28932680 on Build GRCh38
rs756059522 Chr.16: 28932634-28932634 on Build GRCh38
rs151258764 Chr.16: 28932351-28932351 on Build GRCh38
rs767961417 Chr.16: 28932356-28932356 on Build GRCh38
rs556158133 Chr.16: 28932676-28932676 on Build GRCh38
rs934151410 Chr.16: 28932365-28932365 on Build GRCh38
rs777859865 Chr.16: 28932636-28932636 on Build GRCh38
rs902191638 Chr.16: 28932730-28932730 on Build GRCh38
rs199709601 Chr.16: 28932302-28932302 on Build GRCh38
rs537910426 Chr.16: 28932231-28932231 on Build GRCh38
rs757413867 Chr.16: 28932306-28932306 on Build GRCh38
rs570540084 Chr.16: 28932638-28932638 on Build GRCh38
rs35210516 Chr.16: 28932493-28932493 on Build GRCh38
rs558660062 Chr.16: 28932598-28932598 on Build GRCh38
rs754166542 Chr.16: 28932303-28932303 on Build GRCh38
rs902138712 Chr.16: 28932452-28932452 on Build GRCh38
rs779619922 Chr.16: 28932695-28932695 on Build GRCh38
rs764433136 Chr.16: 28932512-28932512 on Build GRCh38
rs112625011 Chr.16: 28932287-28932287 on Build GRCh38
rs375965055 Chr.16: 28932397-28932397 on Build GRCh38
rs746831063 Chr.16: 28932425-28932425 on Build GRCh38
rs777371854 Chr.16: 28932559-28932559 on Build GRCh38
rs770731258 Chr.16: 28932481-28932481 on Build GRCh38
rs777677017 Chr.16: 28932298-28932298 on Build GRCh38
rs769495907 Chr.16: 28932347-28932347 on Build GRCh38
rs761039208 Chr.16: 28932387-28932387 on Build GRCh38
rs1043968141 Chr.16: 28932723-28932723 on Build GRCh38
rs765745106 Chr.16: 28932599-28932599 on Build GRCh38
rs755195471 Chr.16: 28932623-28932623 on Build GRCh38
rs548605519 Chr.16: 28932665-28932665 on Build GRCh38
rs766624731 Chr.16: 28932536-28932536 on Build GRCh38
rs754293950 Chr.16: 28932562-28932562 on Build GRCh38
rs764575859 Chr.16: 28932302-28932303 on Build GRCh38
rs906908443 Chr.16: 28932664-28932664 on Build GRCh38
rs758662669 Chr.16: 28932417-28932417 on Build GRCh38
rs753975373 Chr.16: 28932303-28932303 on Build GRCh38
rs1023930462 Chr.16: 28932330-28932330 on Build GRCh38
rs185062613 Chr.16: 28932427-28932427 on Build GRCh38
rs376715546 Chr.16: 28932564-28932564 on Build GRCh38
rs1010611591 Chr.16: 28932435-28932435 on Build GRCh38
rs574802885 Chr.16: 28932701-28932701 on Build GRCh38
rs776903049 Chr.16: 28932580-28932580 on Build GRCh38
rs368196796 Chr.16: 28932554-28932554 on Build GRCh38
rs148732534 Chr.16: 28932309-28932309 on Build GRCh38
rs762651897 Chr.16: 28932470-28932470 on Build GRCh38
rs199665700 Chr.16: 28932428-28932428 on Build GRCh38
rs752560561 Chr.16: 28932629-28932629 on Build GRCh38
rs750625293 Chr.16: 28932415-28932415 on Build GRCh38
rs780897095 Chr.16: 28932551-28932551 on Build GRCh38
rs944597391 Chr.16: 28932508-28932508 on Build GRCh38
rs751783268 Chr.16: 28932622-28932622 on Build GRCh38
rs999692059 Chr.16: 28932485-28932485 on Build GRCh38
rs778340411 Chr.16: 28932663-28932663 on Build GRCh38
rs946954641 Chr.16: 28932717-28932717 on Build GRCh38
rs916476576 Chr.16: 28932246-28932246 on Build GRCh38
rs374321123 Chr.16: 28932553-28932553 on Build GRCh38

View Full Product Details