Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55785566-55786133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NLRP11
Entrez Gene ID
204801
Gene Name
NLR family pyrin domain containing 11
Gene Aliases
CLR19.6, NALP11, NOD17, PAN10, PYPAF6, PYPAF7
UniGene
Hs.375039
Gene Chromosome Location
Chr.19: 55785397-55836762 on Build GRCh38
Transcripts
NM_001297743.1 NM_145007.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2969485 c.3046C>T p.P1016S Chr.19: 55785681-55785681 on Build GRCh38
1712868 c.3025G>A p.D1009N Chr.19: 55785702-55785702 on Build GRCh38
2969487 c.2995C>T p.P999S Chr.19: 55785732-55785732 on Build GRCh38
1712869 c.2959G>A p.E987K Chr.19: 55785768-55785768 on Build GRCh38
3735557 c.3047C>T p.P1016L Chr.19: 55785680-55785680 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11671248 Chr.19: 55785653-55785653 on Build GRCh38
rs746468121 Chr.19: 55785639-55785639 on Build GRCh38
rs750399264 Chr.19: 55785796-55785796 on Build GRCh38
rs531029933 Chr.19: 55785968-55785968 on Build GRCh38
rs897042930 Chr.19: 55786067-55786067 on Build GRCh38
rs561422172 Chr.19: 55786029-55786029 on Build GRCh38
rs755811062 Chr.19: 55785884-55785885 on Build GRCh38
rs752650898 Chr.19: 55786038-55786038 on Build GRCh38
rs925645925 Chr.19: 55785662-55785662 on Build GRCh38
rs759879184 Chr.19: 55785864-55785864 on Build GRCh38
rs769714987 Chr.19: 55785660-55785660 on Build GRCh38
rs868013577 Chr.19: 55785799-55785799 on Build GRCh38
rs778715243 Chr.19: 55785909-55785909 on Build GRCh38
rs761693639 Chr.19: 55785822-55785822 on Build GRCh38
rs773290992 Chr.19: 55785828-55785828 on Build GRCh38
rs757992975 Chr.19: 55785635-55785635 on Build GRCh38
rs375884712 Chr.19: 55785894-55785894 on Build GRCh38
rs777371539 Chr.19: 55785637-55785637 on Build GRCh38
rs979018745 Chr.19: 55785929-55785929 on Build GRCh38
rs772062539 Chr.19: 55785823-55785823 on Build GRCh38
rs754736781 Chr.19: 55785903-55785903 on Build GRCh38
rs982205774 Chr.19: 55785833-55785833 on Build GRCh38
rs763871559 Chr.19: 55785874-55785874 on Build GRCh38
rs373504408 Chr.19: 55786000-55786000 on Build GRCh38
rs1016511812 Chr.19: 55785679-55785679 on Build GRCh38
rs777026964 Chr.19: 55785863-55785863 on Build GRCh38
rs752355787 Chr.19: 55785630-55785630 on Build GRCh38
rs748344703 Chr.19: 55785915-55785915 on Build GRCh38
rs763628855 Chr.19: 55785774-55785774 on Build GRCh38
rs993539839 Chr.19: 55786112-55786112 on Build GRCh38
rs377418854 Chr.19: 55785628-55785628 on Build GRCh38
rs59238812 Chr.19: 55785883-55785883 on Build GRCh38
rs751137056 Chr.19: 55785777-55785777 on Build GRCh38
rs535635252 Chr.19: 55785613-55785613 on Build GRCh38
rs116820715 Chr.19: 55785798-55785798 on Build GRCh38
rs764263811 Chr.19: 55785884-55785887 on Build GRCh38
rs761856496 Chr.19: 55786004-55786004 on Build GRCh38
rs144741765 Chr.19: 55785885-55785885 on Build GRCh38
rs779002503 Chr.19: 55785820-55785820 on Build GRCh38
rs751788635 Chr.19: 55785883-55785884 on Build GRCh38
rs1005211191 Chr.19: 55785902-55785902 on Build GRCh38
rs1044084908 Chr.19: 55786028-55786028 on Build GRCh38
rs368312156 Chr.19: 55785693-55785693 on Build GRCh38
rs144909019 Chr.19: 55785717-55785717 on Build GRCh38
rs934383363 Chr.19: 55785947-55785947 on Build GRCh38
rs866074331 Chr.19: 55785768-55785768 on Build GRCh38
rs761117440 Chr.19: 55785743-55785743 on Build GRCh38
rs529108694 Chr.19: 55786039-55786039 on Build GRCh38
rs768139801 Chr.19: 55785714-55785714 on Build GRCh38
rs761789117 Chr.19: 55785710-55785710 on Build GRCh38
rs775377221 Chr.19: 55785661-55785661 on Build GRCh38
rs866064963 Chr.19: 55786016-55786016 on Build GRCh38
rs917034927 Chr.19: 55785976-55785976 on Build GRCh38
rs989508457 Chr.19: 55786107-55786107 on Build GRCh38
rs972671262 Chr.19: 55785753-55785753 on Build GRCh38
rs373925447 Chr.19: 55785848-55785848 on Build GRCh38
rs193180010 Chr.19: 55785656-55785656 on Build GRCh38
rs749071062 Chr.19: 55785807-55785807 on Build GRCh38
rs372343459 Chr.19: 55785948-55785948 on Build GRCh38
rs780686846 Chr.19: 55785792-55785792 on Build GRCh38
rs749921411 Chr.19: 55785896-55785896 on Build GRCh38
rs1052909714 Chr.19: 55785698-55785698 on Build GRCh38
rs73933365 Chr.19: 55785923-55785923 on Build GRCh38
rs756170921 Chr.19: 55785897-55785897 on Build GRCh38
rs758498121 Chr.19: 55785600-55785600 on Build GRCh38
rs910740816 Chr.19: 55785905-55785905 on Build GRCh38
rs774255180 Chr.19: 55785589-55785589 on Build GRCh38
rs145199542 Chr.19: 55785587-55785587 on Build GRCh38
rs1014066502 Chr.19: 55785783-55785783 on Build GRCh38
rs762424014 Chr.19: 55785595-55785595 on Build GRCh38
rs926576214 Chr.19: 55785849-55785849 on Build GRCh38
rs766748570 Chr.19: 55785744-55785744 on Build GRCh38
rs987617261 Chr.19: 55785622-55785622 on Build GRCh38
rs868582143 Chr.19: 55785755-55785755 on Build GRCh38
rs768970824 Chr.19: 55785682-55785682 on Build GRCh38
rs758084844 Chr.19: 55785760-55785760 on Build GRCh38
rs530396765 Chr.19: 55786032-55786032 on Build GRCh38
rs919332181 Chr.19: 55785791-55785791 on Build GRCh38
rs926192376 Chr.19: 55785932-55785932 on Build GRCh38
rs115847229 Chr.19: 55785668-55785668 on Build GRCh38
rs752753934 Chr.19: 55785598-55785601 on Build GRCh38
rs753740562 Chr.19: 55785815-55785815 on Build GRCh38
rs370557210 Chr.19: 55785787-55785787 on Build GRCh38
rs549137337 Chr.19: 55786087-55786087 on Build GRCh38
rs780035413 Chr.19: 55785801-55785801 on Build GRCh38
rs766968565 Chr.19: 55785895-55785895 on Build GRCh38
rs767889163 Chr.19: 55785611-55785611 on Build GRCh38
rs370823183 Chr.19: 55785655-55785655 on Build GRCh38
rs368951328 Chr.19: 55785898-55785898 on Build GRCh38
rs756470699 Chr.19: 55785614-55785614 on Build GRCh38
rs754157491 Chr.19: 55785757-55785757 on Build GRCh38
rs184634768 Chr.19: 55785954-55785954 on Build GRCh38
rs573456824 Chr.19: 55785944-55785944 on Build GRCh38
rs748337702 Chr.19: 55785821-55785821 on Build GRCh38
rs536704098 Chr.19: 55785862-55785862 on Build GRCh38

View Full Product Details