Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 16783932-16784443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSPAN13
Entrez Gene ID
27075
Gene Name
tetraspanin 13
Gene Aliases
NET-6, NET6, TM4SF13
UniGene
Hs.364544
Gene Chromosome Location
Chr.7: 16753726-16784536 on Build GRCh38
Transcripts
NM_014399.3

SNP Detail

SNP ID Chromosome Location
rs551083668 Chr.7: 16784047-16784047 on Build GRCh38
rs957699692 Chr.7: 16784366-16784366 on Build GRCh38
rs1001856505 Chr.7: 16784055-16784055 on Build GRCh38
rs987009867 Chr.7: 16784390-16784390 on Build GRCh38
rs115362407 Chr.7: 16784288-16784288 on Build GRCh38
rs1020980670 Chr.7: 16783982-16783985 on Build GRCh38
rs915385418 Chr.7: 16784230-16784230 on Build GRCh38
rs537558366 Chr.7: 16784286-16784286 on Build GRCh38
rs555524422 Chr.7: 16784108-16784108 on Build GRCh38
rs533607135 Chr.7: 16784072-16784072 on Build GRCh38
rs373091532 Chr.7: 16784227-16784227 on Build GRCh38
rs1009801372 Chr.7: 16783956-16783957 on Build GRCh38
rs967533459 Chr.7: 16784185-16784185 on Build GRCh38
rs950653603 Chr.7: 16784115-16784115 on Build GRCh38
rs571577744 Chr.7: 16784280-16784280 on Build GRCh38
rs911584859 Chr.7: 16784141-16784141 on Build GRCh38
rs7728 Chr.7: 16784380-16784380 on Build GRCh38
rs557871160 Chr.7: 16784222-16784222 on Build GRCh38
rs3840575 Chr.7: 16783982-16783983 on Build GRCh38
rs942928602 Chr.7: 16784238-16784238 on Build GRCh38
rs535325060 Chr.7: 16783969-16783969 on Build GRCh38
rs1013389645 Chr.7: 16784294-16784294 on Build GRCh38
rs973268208 Chr.7: 16784075-16784075 on Build GRCh38
rs1003539085 Chr.7: 16784278-16784278 on Build GRCh38
rs77270635 Chr.7: 16783957-16783957 on Build GRCh38
rs759595953 Chr.7: 16784148-16784148 on Build GRCh38
rs1024857006 Chr.7: 16784028-16784028 on Build GRCh38
rs1032003714 Chr.7: 16784362-16784362 on Build GRCh38
rs906567153 Chr.7: 16784275-16784275 on Build GRCh38
rs567827117 Chr.7: 16784157-16784157 on Build GRCh38
rs983300451 Chr.7: 16784121-16784121 on Build GRCh38
rs969471199 Chr.7: 16784048-16784048 on Build GRCh38
rs553832552 Chr.7: 16784340-16784340 on Build GRCh38
rs1009275743 Chr.7: 16784368-16784368 on Build GRCh38
rs6616 Chr.7: 16784353-16784353 on Build GRCh38
rs987844163 Chr.7: 16784088-16784088 on Build GRCh38
rs111699315 Chr.7: 16784341-16784341 on Build GRCh38
rs191600592 Chr.7: 16784186-16784186 on Build GRCh38
rs1047136301 Chr.7: 16784287-16784287 on Build GRCh38
rs143475712 Chr.7: 16784251-16784251 on Build GRCh38
rs931423096 Chr.7: 16784285-16784285 on Build GRCh38
rs987467561 Chr.7: 16784203-16784203 on Build GRCh38
rs148850575 Chr.7: 16784064-16784064 on Build GRCh38
rs1046764504 Chr.7: 16784297-16784297 on Build GRCh38
rs912785096 Chr.7: 16784228-16784228 on Build GRCh38
rs184463584 Chr.7: 16784369-16784369 on Build GRCh38
rs765244340 Chr.7: 16784150-16784150 on Build GRCh38
rs935711230 Chr.7: 16784209-16784209 on Build GRCh38
rs967963684 Chr.7: 16784374-16784374 on Build GRCh38
rs764521960 Chr.7: 16784258-16784258 on Build GRCh38
rs768062274 Chr.7: 16783983-16783986 on Build GRCh38
rs1031951746 Chr.7: 16784062-16784062 on Build GRCh38
rs913443412 Chr.7: 16784171-16784171 on Build GRCh38

View Full Product Details