Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 43067574-43067760 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRCA1
Entrez Gene ID
672
Gene Name
BRCA1, DNA repair associated
Gene Aliases
BRCAI, BRCC1, BROVCA1, FANCS, IRIS, PNCA4, PPP1R53, PSCP, RNF53
UniGene
Hs.194143
Gene Chromosome Location
Chr.17: 43044295-43125483 on Build GRCh38
Transcripts
NM_007294.3 NM_007297.3 NM_007298.3 NM_007299.3 NM_007300.3 NR_027676.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4427610 c.5083A>T p.I1695F Chr.17: 43067662-43067662 on Build GRCh38
1610297 c.5023A>G p.T1675A Chr.17: 43067659-43067659 on Build GRCh38
1610298 c.5000A>C p.K1667T Chr.17: 43067682-43067682 on Build GRCh38
6974300 c.5038A>G p.I1680V Chr.17: 43067644-43067644 on Build GRCh38
7343721 c.5005G>T p.A1669S Chr.17: 43067677-43067677 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs80357580 Chr.17: 43067652-43067655 on Build GRCh38
rs431825411 Chr.17: 43067603-43067603 on Build GRCh38
rs879254050 Chr.17: 43067624-43067625 on Build GRCh38
rs876659314 Chr.17: 43067641-43067641 on Build GRCh38
rs367645566 Chr.17: 43067728-43067728 on Build GRCh38
rs754256578 Chr.17: 43067645-43067645 on Build GRCh38
rs397509218 Chr.17: 43067624-43067624 on Build GRCh38
rs886040264 Chr.17: 43067640-43067641 on Build GRCh38
rs886040261 Chr.17: 43067655-43067656 on Build GRCh38
rs748807039 Chr.17: 43067693-43067693 on Build GRCh38
rs397509214 Chr.17: 43067700-43067700 on Build GRCh38
rs80357894 Chr.17: 43067646-43067656 on Build GRCh38
rs80357974 Chr.17: 43067625-43067626 on Build GRCh38
rs898511378 Chr.17: 43067648-43067648 on Build GRCh38
rs766784305 Chr.17: 43067640-43067640 on Build GRCh38
rs80357043 Chr.17: 43067628-43067628 on Build GRCh38
rs773821829 Chr.17: 43067711-43067711 on Build GRCh38
rs587781477 Chr.17: 43067668-43067668 on Build GRCh38
rs80358053 Chr.17: 43067607-43067607 on Build GRCh38
rs886040266 Chr.17: 43067634-43067638 on Build GRCh38
rs80357673 Chr.17: 43067642-43067642 on Build GRCh38
rs397509215 Chr.17: 43067686-43067686 on Build GRCh38
rs80357061 Chr.17: 43067616-43067616 on Build GRCh38
rs80357623 Chr.17: 43067643-43067647 on Build GRCh38
rs80358170 Chr.17: 43067706-43067706 on Build GRCh38
rs80357672 Chr.17: 43067610-43067611 on Build GRCh38
rs80357034 Chr.17: 43067610-43067610 on Build GRCh38
rs397509216 Chr.17: 43067685-43067685 on Build GRCh38
rs397509217 Chr.17: 43067654-43067655 on Build GRCh38
rs777828258 Chr.17: 43067672-43067672 on Build GRCh38
rs879930398 Chr.17: 43067713-43067713 on Build GRCh38
rs879255491 Chr.17: 43067632-43067632 on Build GRCh38
rs142459158 Chr.17: 43067690-43067690 on Build GRCh38
rs80356853 Chr.17: 43067609-43067609 on Build GRCh38
rs80356890 Chr.17: 43067629-43067629 on Build GRCh38
rs80358344 Chr.17: 43067618-43067620 on Build GRCh38
rs150729791 Chr.17: 43067658-43067658 on Build GRCh38
rs80357924 Chr.17: 43067651-43067654 on Build GRCh38
rs80357314 Chr.17: 43067691-43067691 on Build GRCh38
rs730882166 Chr.17: 43067625-43067625 on Build GRCh38
rs776992322 Chr.17: 43067731-43067731 on Build GRCh38
rs886040268 Chr.17: 43067616-43067617 on Build GRCh38
rs751856943 Chr.17: 43067675-43067675 on Build GRCh38
rs80358343 Chr.17: 43067663-43067665 on Build GRCh38
rs80358032 Chr.17: 43067602-43067602 on Build GRCh38
rs397509212 Chr.17: 43067697-43067697 on Build GRCh38
rs886038039 Chr.17: 43067651-43067655 on Build GRCh38
rs876660263 Chr.17: 43067652-43067652 on Build GRCh38
rs886040260 Chr.17: 43067663-43067663 on Build GRCh38
rs80358056 Chr.17: 43067735-43067735 on Build GRCh38
rs397509219 Chr.17: 43067611-43067611 on Build GRCh38
rs730882165 Chr.17: 43067684-43067684 on Build GRCh38
rs483353097 Chr.17: 43067674-43067675 on Build GRCh38
rs80357938 Chr.17: 43067677-43067677 on Build GRCh38
rs80358181 Chr.17: 43067605-43067605 on Build GRCh38
rs397509213 Chr.17: 43067698-43067698 on Build GRCh38
rs886040262 Chr.17: 43067655-43067655 on Build GRCh38
rs876658947 Chr.17: 43067684-43067685 on Build GRCh38
rs889937720 Chr.17: 43067681-43067681 on Build GRCh38
rs760038328 Chr.17: 43067646-43067646 on Build GRCh38
rs1025136518 Chr.17: 43067702-43067702 on Build GRCh38
rs760922019 Chr.17: 43067630-43067630 on Build GRCh38
rs886040265 Chr.17: 43067640-43067640 on Build GRCh38
rs759950362 Chr.17: 43067733-43067733 on Build GRCh38
rs80356958 Chr.17: 43067638-43067638 on Build GRCh38
rs80187739 Chr.17: 43067608-43067608 on Build GRCh38
rs786203868 Chr.17: 43067660-43067660 on Build GRCh38
rs80357087 Chr.17: 43067677-43067677 on Build GRCh38
rs1057519495 Chr.17: 43067682-43067682 on Build GRCh38
rs80358089 Chr.17: 43067606-43067606 on Build GRCh38
rs994608035 Chr.17: 43067724-43067724 on Build GRCh38
rs886040267 Chr.17: 43067623-43067624 on Build GRCh38
rs80358035 Chr.17: 43067715-43067715 on Build GRCh38
rs80357204 Chr.17: 43067683-43067683 on Build GRCh38
rs879255283 Chr.17: 43067631-43067632 on Build GRCh38
rs758539222 Chr.17: 43067665-43067665 on Build GRCh38
rs765241963 Chr.17: 43067651-43067651 on Build GRCh38
rs934248073 Chr.17: 43067647-43067647 on Build GRCh38
rs1057518640 Chr.17: 43067676-43067676 on Build GRCh38
rs483353098 Chr.17: 43067648-43067649 on Build GRCh38
rs80357117 Chr.17: 43067695-43067695 on Build GRCh38
rs397507239 Chr.17: 43067614-43067614 on Build GRCh38
rs431825410 Chr.17: 43067652-43067656 on Build GRCh38
rs876658226 Chr.17: 43067657-43067657 on Build GRCh38
rs886040912 Chr.17: 43067695-43067695 on Build GRCh38
rs80357896 Chr.17: 43067647-43067647 on Build GRCh38
rs886040259 Chr.17: 43067668-43067669 on Build GRCh38
rs80357265 Chr.17: 43067637-43067637 on Build GRCh38
rs786203754 Chr.17: 43067655-43067655 on Build GRCh38
rs587781472 Chr.17: 43067680-43067680 on Build GRCh38
rs730881495 Chr.17: 43067696-43067696 on Build GRCh38
rs774452090 Chr.17: 43067659-43067659 on Build GRCh38
rs80357862 Chr.17: 43067649-43067652 on Build GRCh38
rs1050371477 Chr.17: 43067662-43067662 on Build GRCh38
rs80356879 Chr.17: 43067635-43067635 on Build GRCh38
rs80357169 Chr.17: 43067689-43067689 on Build GRCh38
rs80357205 Chr.17: 43067694-43067694 on Build GRCh38

View Full Product Details