Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27987862-27988420 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRE
Entrez Gene ID
9577
Gene Name
brain and reproductive organ-expressed (TNFRSF1A modulator)
Gene Aliases
BRCC4, BRCC45
UniGene
Hs.258314
Gene Chromosome Location
Chr.2: 27890615-28338901 on Build GRCh38
Transcripts
NM_001261840.1 NM_004899.4 NM_199191.2 NM_199192.2 NM_199193.2 NM_199194.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7136451 c.247G>T p.D83Y Chr.2: 27988034-27988034 on Build GRCh38
1153781 c.243T>C p.P81P Chr.2: 27988030-27988030 on Build GRCh38
7136449 c.247G>T p.D83Y Chr.2: 27988034-27988034 on Build GRCh38
3580753 c.259G>T p.G87* Chr.2: 27988046-27988046 on Build GRCh38
6158072 c.247G>A p.D83N Chr.2: 27988034-27988034 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550220860 Chr.2: 27988248-27988248 on Build GRCh38
rs878995653 Chr.2: 27988391-27988391 on Build GRCh38
rs541192918 Chr.2: 27987909-27987909 on Build GRCh38
rs571880325 Chr.2: 27988253-27988253 on Build GRCh38
rs770769036 Chr.2: 27988018-27988018 on Build GRCh38
rs532074070 Chr.2: 27988182-27988182 on Build GRCh38
rs772560433 Chr.2: 27988022-27988022 on Build GRCh38
rs762256443 Chr.2: 27988346-27988346 on Build GRCh38
rs184735129 Chr.2: 27988094-27988094 on Build GRCh38
rs996557921 Chr.2: 27988163-27988163 on Build GRCh38
rs140802379 Chr.2: 27988073-27988073 on Build GRCh38
rs967023834 Chr.2: 27988326-27988326 on Build GRCh38
rs373014128 Chr.2: 27988228-27988228 on Build GRCh38
rs557932684 Chr.2: 27988341-27988341 on Build GRCh38
rs1020639640 Chr.2: 27988288-27988288 on Build GRCh38
rs748135959 Chr.2: 27988015-27988015 on Build GRCh38
rs768506215 Chr.2: 27987987-27987987 on Build GRCh38
rs145549560 Chr.2: 27988066-27988066 on Build GRCh38
rs780337550 Chr.2: 27987971-27987971 on Build GRCh38
rs1009525175 Chr.2: 27988358-27988358 on Build GRCh38
rs778908413 Chr.2: 27987968-27987968 on Build GRCh38
rs1040067303 Chr.2: 27988332-27988332 on Build GRCh38
rs560571611 Chr.2: 27987989-27987989 on Build GRCh38
rs759345889 Chr.2: 27988139-27988139 on Build GRCh38
rs888844464 Chr.2: 27988268-27988268 on Build GRCh38
rs754373625 Chr.2: 27988077-27988077 on Build GRCh38
rs761898286 Chr.2: 27987951-27987954 on Build GRCh38
rs745980349 Chr.2: 27987969-27987969 on Build GRCh38
rs1021281897 Chr.2: 27988387-27988387 on Build GRCh38
rs1037020979 Chr.2: 27988153-27988153 on Build GRCh38
rs765887686 Chr.2: 27988033-27988033 on Build GRCh38
rs1040555855 Chr.2: 27988093-27988093 on Build GRCh38
rs967850379 Chr.2: 27987939-27987939 on Build GRCh38
rs750692921 Chr.2: 27987958-27987961 on Build GRCh38
rs1020808124 Chr.2: 27987921-27987921 on Build GRCh38
rs769796449 Chr.2: 27988021-27988021 on Build GRCh38
rs538858980 Chr.2: 27988299-27988299 on Build GRCh38
rs757788978 Chr.2: 27988091-27988091 on Build GRCh38
rs762526215 Chr.2: 27988027-27988027 on Build GRCh38
rs926002077 Chr.2: 27988001-27988001 on Build GRCh38
rs1034785389 Chr.2: 27988345-27988345 on Build GRCh38
rs750414615 Chr.2: 27988136-27988136 on Build GRCh38
rs758484958 Chr.2: 27987970-27987970 on Build GRCh38
rs764593436 Chr.2: 27988075-27988075 on Build GRCh38
rs765254261 Chr.2: 27987958-27987958 on Build GRCh38
rs774107402 Chr.2: 27988046-27988046 on Build GRCh38
rs369134360 Chr.2: 27987973-27987973 on Build GRCh38
rs567664165 Chr.2: 27988102-27988102 on Build GRCh38
rs901484818 Chr.2: 27988354-27988354 on Build GRCh38
rs1009883076 Chr.2: 27987901-27987901 on Build GRCh38
rs769590619 Chr.2: 27988172-27988172 on Build GRCh38

View Full Product Details