Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18877995-18878510 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CERS1
Entrez Gene ID
10715
Gene Name
ceramide synthase 1
Gene Aliases
EPM8, LAG1, LASS1, UOG1
UniGene
Hs.412355
Gene Chromosome Location
Chr.19: 18868546-18896727 on Build GRCh38
Transcripts
NM_001290265.1 NM_021267.4 NM_198207.2 NM_001492.5

Gene Symbol
GDF1
Entrez Gene ID
2657
Gene Name
growth differentiation factor 1
Gene Aliases
DORV, DTGA3, RAI
UniGene
Hs.412355
Gene Chromosome Location
Chr.19: 18868546-18896144 on Build GRCh38
Transcripts
NM_001290265.1 NM_021267.4 NM_198207.2 NM_001492.5

SNP Detail

SNP ID Chromosome Location
rs1030287490 Chr.19: 18878279-18878279 on Build GRCh38
rs564002546 Chr.19: 18878333-18878333 on Build GRCh38
rs559657143 Chr.19: 18878324-18878324 on Build GRCh38
rs890670309 Chr.19: 18878325-18878325 on Build GRCh38
rs1011586623 Chr.19: 18878379-18878379 on Build GRCh38
rs947769869 Chr.19: 18878197-18878197 on Build GRCh38
rs551798078 Chr.19: 18878482-18878482 on Build GRCh38
rs113080020 Chr.19: 18878037-18878037 on Build GRCh38
rs891161198 Chr.19: 18878295-18878295 on Build GRCh38
rs77593844 Chr.19: 18878109-18878109 on Build GRCh38
rs1009159170 Chr.19: 18878339-18878339 on Build GRCh38
rs1029277481 Chr.19: 18878067-18878067 on Build GRCh38
rs1055584786 Chr.19: 18878223-18878223 on Build GRCh38
rs188033775 Chr.19: 18878442-18878442 on Build GRCh38
rs765067887 Chr.19: 18878060-18878060 on Build GRCh38
rs1005717275 Chr.19: 18878019-18878019 on Build GRCh38
rs193060861 Chr.19: 18878457-18878457 on Build GRCh38
rs946112134 Chr.19: 18878272-18878272 on Build GRCh38
rs540900923 Chr.19: 18878392-18878392 on Build GRCh38
rs994977151 Chr.19: 18878430-18878430 on Build GRCh38
rs759864943 Chr.19: 18878126-18878126 on Build GRCh38
rs989922717 Chr.19: 18878117-18878117 on Build GRCh38
rs1010284824 Chr.19: 18878027-18878027 on Build GRCh38
rs1038534561 Chr.19: 18878233-18878233 on Build GRCh38
rs115623466 Chr.19: 18878479-18878479 on Build GRCh38
rs149125950 Chr.19: 18878052-18878052 on Build GRCh38
rs562624889 Chr.19: 18878163-18878163 on Build GRCh38
rs1029677818 Chr.19: 18878059-18878059 on Build GRCh38
rs572083411 Chr.19: 18878299-18878299 on Build GRCh38
rs1052186354 Chr.19: 18878232-18878232 on Build GRCh38
rs556414544 Chr.19: 18878220-18878220 on Build GRCh38
rs1043171742 Chr.19: 18878275-18878275 on Build GRCh38
rs967142683 Chr.19: 18878401-18878401 on Build GRCh38
rs768519231 Chr.19: 18878483-18878483 on Build GRCh38
rs927919222 Chr.19: 18878225-18878225 on Build GRCh38
rs962221165 Chr.19: 18878417-18878417 on Build GRCh38
rs955532939 Chr.19: 18878071-18878071 on Build GRCh38
rs771335059 Chr.19: 18878076-18878076 on Build GRCh38
rs897300152 Chr.19: 18878234-18878234 on Build GRCh38
rs1017237969 Chr.19: 18878026-18878026 on Build GRCh38
rs904511296 Chr.19: 18878276-18878276 on Build GRCh38
rs928537506 Chr.19: 18878420-18878420 on Build GRCh38
rs4808869 Chr.19: 18878461-18878461 on Build GRCh38
rs887525491 Chr.19: 18878227-18878228 on Build GRCh38
rs183051735 Chr.19: 18878124-18878124 on Build GRCh38
rs913750924 Chr.19: 18878257-18878257 on Build GRCh38
rs960356267 Chr.19: 18878363-18878363 on Build GRCh38
rs10426520 Chr.19: 18878136-18878136 on Build GRCh38
rs981081884 Chr.19: 18878222-18878222 on Build GRCh38
rs913179458 Chr.19: 18878118-18878118 on Build GRCh38
rs183667458 Chr.19: 18878287-18878287 on Build GRCh38
rs561149924 Chr.19: 18878440-18878440 on Build GRCh38
rs987679562 Chr.19: 18878077-18878077 on Build GRCh38
rs187001958 Chr.19: 18878230-18878230 on Build GRCh38
rs973187815 Chr.19: 18878162-18878162 on Build GRCh38
rs191513907 Chr.19: 18878246-18878246 on Build GRCh38

View Full Product Details