Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 51354126-51354634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETFB
Entrez Gene ID
2109
Gene Name
electron transfer flavoprotein beta subunit
Gene Aliases
FP585, MADD
UniGene
Hs.348531
Gene Chromosome Location
Chr.19: 51345155-51366418 on Build GRCh38
Transcripts
NM_001014763.1 NM_001985.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1683709 c.12_13insCTGTGG p.W6_V7insLW Chr.19: 51354626-51354627 on Build GRCh38
7106380 c.234C>T p.V78V Chr.19: 51354405-51354405 on Build GRCh38
5511113 c.157delC p.L53fs*38 Chr.19: 51354482-51354482 on Build GRCh38
3743087 c.168G>T p.K56N Chr.19: 51354198-51354198 on Build GRCh38
349647 c.396C>G p.F132L Chr.19: 51354243-51354243 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547092773 Chr.19: 51354579-51354579 on Build GRCh38
rs751958348 Chr.19: 51354535-51354535 on Build GRCh38
rs768911920 Chr.19: 51354427-51354427 on Build GRCh38
rs564020088 Chr.19: 51354288-51354288 on Build GRCh38
rs146798901 Chr.19: 51354399-51354399 on Build GRCh38
rs374277927 Chr.19: 51354434-51354434 on Build GRCh38
rs762647069 Chr.19: 51354407-51354421 on Build GRCh38
rs140608276 Chr.19: 51354404-51354404 on Build GRCh38
rs199860339 Chr.19: 51354306-51354306 on Build GRCh38
rs375136201 Chr.19: 51354444-51354444 on Build GRCh38
rs757779883 Chr.19: 51354389-51354389 on Build GRCh38
rs371751519 Chr.19: 51354274-51354274 on Build GRCh38
rs765099692 Chr.19: 51354281-51354281 on Build GRCh38
rs772322071 Chr.19: 51354248-51354248 on Build GRCh38
rs775541180 Chr.19: 51354253-51354253 on Build GRCh38
rs368139326 Chr.19: 51354268-51354268 on Build GRCh38
rs779344556 Chr.19: 51354396-51354396 on Build GRCh38
rs752825481 Chr.19: 51354196-51354196 on Build GRCh38
rs770414295 Chr.19: 51354232-51354232 on Build GRCh38
rs986329798 Chr.19: 51354387-51354387 on Build GRCh38
rs772266930 Chr.19: 51354330-51354330 on Build GRCh38
rs767608889 Chr.19: 51354586-51354587 on Build GRCh38
rs989233154 Chr.19: 51354176-51354176 on Build GRCh38
rs760573811 Chr.19: 51354197-51354197 on Build GRCh38
rs750230877 Chr.19: 51354284-51354284 on Build GRCh38
rs74357706 Chr.19: 51354365-51354365 on Build GRCh38
rs781748603 Chr.19: 51354149-51354149 on Build GRCh38
rs374819445 Chr.19: 51354285-51354285 on Build GRCh38
rs775425395 Chr.19: 51354375-51354376 on Build GRCh38
rs764083482 Chr.19: 51354198-51354198 on Build GRCh38
rs769799163 Chr.19: 51354568-51354568 on Build GRCh38
rs151108898 Chr.19: 51354233-51354233 on Build GRCh38
rs957700187 Chr.19: 51354489-51354489 on Build GRCh38
rs760115478 Chr.19: 51354509-51354509 on Build GRCh38
rs759464497 Chr.19: 51354609-51354609 on Build GRCh38
rs780209598 Chr.19: 51354414-51354414 on Build GRCh38
rs746082442 Chr.19: 51354244-51354244 on Build GRCh38
rs183159891 Chr.19: 51354273-51354273 on Build GRCh38
rs755221189 Chr.19: 51354536-51354536 on Build GRCh38
rs139990490 Chr.19: 51354372-51354372 on Build GRCh38
rs745870211 Chr.19: 51354335-51354336 on Build GRCh38
rs748684435 Chr.19: 51354159-51354159 on Build GRCh38
rs796051954 Chr.19: 51354206-51354206 on Build GRCh38
rs1862495 Chr.19: 51354450-51354450 on Build GRCh38
rs372040033 Chr.19: 51354231-51354231 on Build GRCh38
rs780806600 Chr.19: 51354198-51354200 on Build GRCh38
rs774561602 Chr.19: 51354540-51354541 on Build GRCh38
rs377521762 Chr.19: 51354435-51354435 on Build GRCh38
rs138412625 Chr.19: 51354474-51354474 on Build GRCh38
rs759233806 Chr.19: 51354482-51354482 on Build GRCh38
rs754545072 Chr.19: 51354302-51354302 on Build GRCh38
rs759494324 Chr.19: 51354183-51354183 on Build GRCh38
rs766066977 Chr.19: 51354365-51354365 on Build GRCh38
rs537142189 Chr.19: 51354228-51354228 on Build GRCh38
rs774387920 Chr.19: 51354242-51354242 on Build GRCh38
rs762800887 Chr.19: 51354471-51354471 on Build GRCh38
rs142966954 Chr.19: 51354179-51354179 on Build GRCh38
rs202006668 Chr.19: 51354366-51354366 on Build GRCh38
rs756659613 Chr.19: 51354361-51354361 on Build GRCh38
rs993404483 Chr.19: 51354356-51354356 on Build GRCh38
rs750117869 Chr.19: 51354224-51354224 on Build GRCh38
rs551328958 Chr.19: 51354560-51354560 on Build GRCh38
rs746284138 Chr.19: 51354307-51354307 on Build GRCh38
rs143144671 Chr.19: 51354520-51354520 on Build GRCh38
rs141529162 Chr.19: 51354360-51354361 on Build GRCh38
rs760800809 Chr.19: 51354254-51354254 on Build GRCh38
rs144991327 Chr.19: 51354483-51354483 on Build GRCh38
rs369216610 Chr.19: 51354304-51354304 on Build GRCh38
rs747250184 Chr.19: 51354422-51354422 on Build GRCh38
rs143544475 Chr.19: 51354373-51354373 on Build GRCh38
rs199705168 Chr.19: 51354182-51354182 on Build GRCh38
rs978749081 Chr.19: 51354405-51354405 on Build GRCh38
rs79338777 Chr.19: 51354484-51354484 on Build GRCh38
rs747565560 Chr.19: 51354503-51354503 on Build GRCh38
rs768440463 Chr.19: 51354502-51354504 on Build GRCh38
rs780370754 Chr.19: 51354534-51354534 on Build GRCh38
rs770467292 Chr.19: 51354164-51354164 on Build GRCh38
rs374935848 Chr.19: 51354234-51354234 on Build GRCh38
rs368212620 Chr.19: 51354207-51354207 on Build GRCh38
rs776379865 Chr.19: 51354352-51354352 on Build GRCh38
rs761984459 Chr.19: 51354583-51354586 on Build GRCh38
rs267605609 Chr.19: 51354177-51354177 on Build GRCh38
rs765125722 Chr.19: 51354354-51354354 on Build GRCh38
rs767608788 Chr.19: 51354192-51354192 on Build GRCh38
rs775702541 Chr.19: 51354334-51354334 on Build GRCh38
rs774338112 Chr.19: 51354481-51354481 on Build GRCh38
rs770567402 Chr.19: 51354185-51354187 on Build GRCh38
rs141875365 Chr.19: 51354543-51354543 on Build GRCh38
rs578243774 Chr.19: 51354344-51354344 on Build GRCh38
rs752190895 Chr.19: 51354379-51354379 on Build GRCh38
rs201291204 Chr.19: 51354533-51354533 on Build GRCh38
rs374288379 Chr.19: 51354305-51354305 on Build GRCh38
rs1005896775 Chr.19: 51354291-51354291 on Build GRCh38
rs553721514 Chr.19: 51354335-51354335 on Build GRCh38
rs768750525 Chr.19: 51354350-51354350 on Build GRCh38
rs201015794 Chr.19: 51354580-51354580 on Build GRCh38
rs374583902 Chr.19: 51354180-51354180 on Build GRCh38
rs774285144 Chr.19: 51354583-51354583 on Build GRCh38
rs771033601 Chr.19: 51354578-51354578 on Build GRCh38
rs371201213 Chr.19: 51354297-51354297 on Build GRCh38
rs757088247 Chr.19: 51354200-51354200 on Build GRCh38
rs769716812 Chr.19: 51354449-51354449 on Build GRCh38
rs749216461 Chr.19: 51354576-51354576 on Build GRCh38
rs559251008 Chr.19: 51354526-51354526 on Build GRCh38

View Full Product Details