Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41801785-41802283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
P3H4
Entrez Gene ID
10609
Gene Name
prolyl 3-hydroxylase family member 4 (non-enzymatic)
Gene Aliases
LEPREL4, NO55, NOL55, SC65
UniGene
Hs.446459
Gene Chromosome Location
Chr.17: 41801952-41812617 on Build GRCh38
Transcripts
NM_006455.2 XM_006721640.3

SNP Detail

SNP ID Chromosome Location
rs1027629090 Chr.17: 41801985-41801985 on Build GRCh38
rs782455832 Chr.17: 41801897-41801897 on Build GRCh38
rs886429687 Chr.17: 41801989-41801989 on Build GRCh38
rs144721873 Chr.17: 41801849-41801849 on Build GRCh38
rs957593860 Chr.17: 41802126-41802126 on Build GRCh38
rs8068917 Chr.17: 41801828-41801828 on Build GRCh38
rs1004994006 Chr.17: 41802035-41802035 on Build GRCh38
rs994859444 Chr.17: 41801943-41801943 on Build GRCh38
rs1034607439 Chr.17: 41802093-41802093 on Build GRCh38
rs1024806639 Chr.17: 41802042-41802042 on Build GRCh38
rs1039256856 Chr.17: 41802222-41802222 on Build GRCh38
rs782163932 Chr.17: 41802099-41802099 on Build GRCh38
rs543887541 Chr.17: 41802140-41802140 on Build GRCh38
rs944560090 Chr.17: 41802197-41802197 on Build GRCh38
rs528201381 Chr.17: 41801935-41801935 on Build GRCh38
rs1061843 Chr.17: 41802092-41802092 on Build GRCh38
rs545307931 Chr.17: 41801908-41801908 on Build GRCh38
rs911597326 Chr.17: 41802181-41802181 on Build GRCh38
rs944529711 Chr.17: 41801934-41801934 on Build GRCh38
rs977903362 Chr.17: 41802162-41802162 on Build GRCh38
rs782236002 Chr.17: 41801847-41801848 on Build GRCh38
rs12602672 Chr.17: 41802137-41802137 on Build GRCh38
rs28500587 Chr.17: 41801927-41801927 on Build GRCh38
rs573602970 Chr.17: 41802015-41802015 on Build GRCh38
rs781833583 Chr.17: 41802049-41802049 on Build GRCh38
rs971621233 Chr.17: 41802052-41802052 on Build GRCh38
rs537925504 Chr.17: 41801815-41801815 on Build GRCh38
rs542844126 Chr.17: 41802059-41802059 on Build GRCh38
rs900381667 Chr.17: 41801941-41801941 on Build GRCh38
rs989117956 Chr.17: 41802132-41802132 on Build GRCh38
rs890335282 Chr.17: 41801930-41801930 on Build GRCh38
rs377025474 Chr.17: 41802173-41802173 on Build GRCh38

View Full Product Details