Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 139278253-139278800 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MATR3
Entrez Gene ID
9782
Gene Name
matrin 3
Gene Aliases
ALS21, MPD2, VCPDM
UniGene
Hs.268939
Gene Chromosome Location
Chr.5: 139273752-139331677 on Build GRCh38
Transcripts
NM_001194954.1 NM_001282278.1 NM_199189.2 NR_003141.3 NR_036536.1 NR_002915.1

Gene Symbol
SNHG4
Entrez Gene ID
724102
Gene Name
small nucleolar RNA host gene 4
Gene Aliases
NCRNA00059, U19H
UniGene
Hs.268939
Gene Chromosome Location
Chr.5: 139273752-139283184 on Build GRCh38
Transcripts
NM_001194954.1 NM_001282278.1 NM_199189.2 NR_003141.3 NR_036536.1 NR_002915.1

Gene Symbol
SNORA74A
Entrez Gene ID
26821
Gene Name
small nucleolar RNA, H/ACA box 74A
Gene Aliases
RNU19, U19
UniGene
Hs.684893
Gene Chromosome Location
Chr.5: 139278780-139278979 on Build GRCh38
Transcripts
NM_001194954.1 NM_001282278.1 NM_199189.2 NR_003141.3 NR_036536.1 NR_002915.1

SNP Detail

SNP ID Chromosome Location
rs762401917 Chr.5: 139278315-139278318 on Build GRCh38
rs983857433 Chr.5: 139278514-139278514 on Build GRCh38
rs776067427 Chr.5: 139278383-139278383 on Build GRCh38
rs1016723720 Chr.5: 139278704-139278704 on Build GRCh38
rs982269090 Chr.5: 139278665-139278665 on Build GRCh38
rs965590475 Chr.5: 139278377-139278377 on Build GRCh38
rs554662043 Chr.5: 139278708-139278708 on Build GRCh38
rs571432057 Chr.5: 139278573-139278573 on Build GRCh38
rs935001378 Chr.5: 139278695-139278695 on Build GRCh38
rs1039881626 Chr.5: 139278719-139278719 on Build GRCh38
rs958422331 Chr.5: 139278531-139278531 on Build GRCh38
rs1011047122 Chr.5: 139278358-139278358 on Build GRCh38
rs749404686 Chr.5: 139278772-139278772 on Build GRCh38
rs191189910 Chr.5: 139278435-139278435 on Build GRCh38
rs754790240 Chr.5: 139278382-139278382 on Build GRCh38
rs763609314 Chr.5: 139278745-139278745 on Build GRCh38
rs562289266 Chr.5: 139278328-139278328 on Build GRCh38
rs1040074012 Chr.5: 139278617-139278619 on Build GRCh38
rs576214782 Chr.5: 139278747-139278747 on Build GRCh38
rs752127962 Chr.5: 139278735-139278735 on Build GRCh38
rs998667842 Chr.5: 139278442-139278442 on Build GRCh38
rs530975907 Chr.5: 139278449-139278449 on Build GRCh38
rs756462888 Chr.5: 139278752-139278752 on Build GRCh38
rs142278625 Chr.5: 139278609-139278609 on Build GRCh38
rs576243316 Chr.5: 139278738-139278738 on Build GRCh38
rs1006113993 Chr.5: 139278690-139278690 on Build GRCh38
rs1025727692 Chr.5: 139278507-139278507 on Build GRCh38
rs780319113 Chr.5: 139278758-139278758 on Build GRCh38
rs938883784 Chr.5: 139278406-139278406 on Build GRCh38
rs1039828197 Chr.5: 139278413-139278413 on Build GRCh38
rs550781795 Chr.5: 139278571-139278571 on Build GRCh38
rs951150541 Chr.5: 139278513-139278513 on Build GRCh38
rs538960882 Chr.5: 139278576-139278576 on Build GRCh38
rs909609126 Chr.5: 139278515-139278515 on Build GRCh38
rs927699487 Chr.5: 139278688-139278688 on Build GRCh38
rs963268361 Chr.5: 139278587-139278587 on Build GRCh38
rs951462800 Chr.5: 139278458-139278458 on Build GRCh38
rs935808161 Chr.5: 139278584-139278584 on Build GRCh38
rs182568165 Chr.5: 139278604-139278604 on Build GRCh38
rs899940794 Chr.5: 139278440-139278440 on Build GRCh38
rs759086594 Chr.5: 139278405-139278405 on Build GRCh38
rs969199661 Chr.5: 139278657-139278657 on Build GRCh38
rs189174220 Chr.5: 139278751-139278751 on Build GRCh38
rs915515234 Chr.5: 139278713-139278713 on Build GRCh38
rs901434228 Chr.5: 139278665-139278665 on Build GRCh38
rs991560229 Chr.5: 139278540-139278540 on Build GRCh38
rs768639630 Chr.5: 139278718-139278718 on Build GRCh38
rs1007286256 Chr.5: 139278289-139278289 on Build GRCh38
rs536255935 Chr.5: 139278672-139278672 on Build GRCh38
rs757797235 Chr.5: 139278744-139278744 on Build GRCh38
rs991173993 Chr.5: 139278739-139278739 on Build GRCh38
rs11747498 Chr.5: 139278588-139278588 on Build GRCh38

View Full Product Details