Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30704612-30705116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MDC1
Entrez Gene ID
9656
Gene Name
mediator of DNA damage checkpoint 1
Gene Aliases
NFBD1
UniGene
Hs.653495
Gene Chromosome Location
Chr.6: 30699807-30717988 on Build GRCh38
Transcripts
NM_014641.2 XM_005249492.2 XM_005249493.3 XM_005249494.4 XM_005249497.3 XM_005249498.3 XM_011515001.2 XM_011515003.2 XM_011515004.2 XM_017011519.1 XM_017011520.1 XM_017011521.1 XM_017011522.1 NR_133647.1

Gene Symbol
MDC1-AS1
Entrez Gene ID
106478956
Gene Name
MDC1 antisense RNA 1
Gene Aliases
MDC1-AS
UniGene
Hs.708954
Gene Chromosome Location
Chr.6: 30703067-30713184 on Build GRCh38
Transcripts
NM_014641.2 XM_005249492.2 XM_005249493.3 XM_005249494.4 XM_005249497.3 XM_005249498.3 XM_011515001.2 XM_011515003.2 XM_011515004.2 XM_017011519.1 XM_017011520.1 XM_017011521.1 XM_017011522.1 NR_133647.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1311963 c.4406C>A p.P1469H Chr.6: 30704777-30704777 on Build GRCh38
6955957 c.4498G>A p.A1500T Chr.6: 30704685-30704685 on Build GRCh38
1647933 c.4373C>A p.S1458Y Chr.6: 30704810-30704810 on Build GRCh38
5423140 c.4243G>A p.E1415K Chr.6: 30704940-30704940 on Build GRCh38
4548562 c.4507G>A p.D1503N Chr.6: 30704676-30704676 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148215318 Chr.6: 30704685-30704685 on Build GRCh38
rs369032728 Chr.6: 30705073-30705073 on Build GRCh38
rs753435554 Chr.6: 30705047-30705047 on Build GRCh38
rs771424163 Chr.6: 30704643-30704643 on Build GRCh38
rs770005786 Chr.6: 30704741-30704741 on Build GRCh38
rs1033514189 Chr.6: 30704787-30704787 on Build GRCh38
rs74590481 Chr.6: 30704941-30704941 on Build GRCh38
rs973577778 Chr.6: 30704664-30704664 on Build GRCh38
rs769695123 Chr.6: 30704924-30704924 on Build GRCh38
rs762976754 Chr.6: 30704820-30704820 on Build GRCh38
rs373993459 Chr.6: 30704707-30704707 on Build GRCh38
rs915933612 Chr.6: 30704690-30704690 on Build GRCh38
rs547516755 Chr.6: 30704773-30704773 on Build GRCh38
rs28994870 Chr.6: 30704980-30704980 on Build GRCh38
rs765522705 Chr.6: 30705078-30705078 on Build GRCh38
rs766872395 Chr.6: 30704944-30704944 on Build GRCh38
rs775132681 Chr.6: 30704925-30704925 on Build GRCh38
rs752524195 Chr.6: 30704713-30704713 on Build GRCh38
rs756506466 Chr.6: 30704878-30704878 on Build GRCh38
rs768793223 Chr.6: 30704756-30704756 on Build GRCh38
rs758821594 Chr.6: 30705085-30705085 on Build GRCh38
rs781723109 Chr.6: 30704740-30704740 on Build GRCh38
rs527477295 Chr.6: 30704705-30704705 on Build GRCh38
rs796874911 Chr.6: 30704796-30704796 on Build GRCh38
rs764878361 Chr.6: 30704808-30704808 on Build GRCh38
rs550952069 Chr.6: 30705019-30705019 on Build GRCh38
rs749692698 Chr.6: 30704750-30704750 on Build GRCh38
rs142364168 Chr.6: 30704790-30704790 on Build GRCh38
rs751090753 Chr.6: 30704841-30704841 on Build GRCh38
rs748601560 Chr.6: 30704981-30704981 on Build GRCh38
rs774561643 Chr.6: 30704684-30704684 on Build GRCh38
rs767309014 Chr.6: 30704871-30704871 on Build GRCh38
rs768664894 Chr.6: 30704676-30704676 on Build GRCh38
rs749127132 Chr.6: 30705025-30705025 on Build GRCh38
rs754117712 Chr.6: 30704967-30704967 on Build GRCh38
rs200570441 Chr.6: 30704940-30704940 on Build GRCh38
rs868326132 Chr.6: 30704696-30704696 on Build GRCh38
rs750158627 Chr.6: 30704950-30704950 on Build GRCh38
rs765255281 Chr.6: 30704710-30704710 on Build GRCh38
rs777350430 Chr.6: 30704717-30704717 on Build GRCh38
rs1044529904 Chr.6: 30704747-30704747 on Build GRCh38
rs777713892 Chr.6: 30705011-30705011 on Build GRCh38
rs751321678 Chr.6: 30704723-30704723 on Build GRCh38
rs370120921 Chr.6: 30704646-30704646 on Build GRCh38
rs749866489 Chr.6: 30704890-30704890 on Build GRCh38
rs765975354 Chr.6: 30705045-30705045 on Build GRCh38
rs756955408 Chr.6: 30704728-30704728 on Build GRCh38
rs10947087 Chr.6: 30704686-30704686 on Build GRCh38
rs755458710 Chr.6: 30704894-30704894 on Build GRCh38
rs1010646827 Chr.6: 30704839-30704839 on Build GRCh38
rs755149297 Chr.6: 30704786-30704787 on Build GRCh38
rs950326503 Chr.6: 30704731-30704731 on Build GRCh38
rs370528787 Chr.6: 30704975-30704975 on Build GRCh38
rs868321115 Chr.6: 30704939-30704939 on Build GRCh38
rs773270002 Chr.6: 30704774-30704774 on Build GRCh38
rs763116546 Chr.6: 30704669-30704669 on Build GRCh38
rs748262427 Chr.6: 30704895-30704895 on Build GRCh38
rs148973631 Chr.6: 30704645-30704645 on Build GRCh38
rs766818773 Chr.6: 30704966-30704966 on Build GRCh38
rs200629873 Chr.6: 30704720-30704720 on Build GRCh38
rs779574818 Chr.6: 30704759-30704759 on Build GRCh38
rs145237068 Chr.6: 30704702-30704702 on Build GRCh38
rs761836754 Chr.6: 30705034-30705034 on Build GRCh38
rs150580476 Chr.6: 30704821-30704821 on Build GRCh38
rs780625550 Chr.6: 30704743-30704743 on Build GRCh38
rs747316218 Chr.6: 30705014-30705014 on Build GRCh38
rs749986214 Chr.6: 30704875-30704875 on Build GRCh38
rs768522998 Chr.6: 30705028-30705028 on Build GRCh38
rs753273267 Chr.6: 30705084-30705084 on Build GRCh38
rs763031886 Chr.6: 30704937-30704937 on Build GRCh38
rs377664869 Chr.6: 30704907-30704907 on Build GRCh38
rs758481782 Chr.6: 30704987-30704987 on Build GRCh38
rs3132589 Chr.6: 30704656-30704656 on Build GRCh38
rs144566920 Chr.6: 30705015-30705015 on Build GRCh38
rs774404458 Chr.6: 30704757-30704757 on Build GRCh38
rs774434419 Chr.6: 30705030-30705030 on Build GRCh38
rs747130577 Chr.6: 30704916-30704916 on Build GRCh38
rs1054604945 Chr.6: 30704869-30704869 on Build GRCh38
rs767111274 Chr.6: 30705039-30705039 on Build GRCh38
rs772572746 Chr.6: 30705042-30705042 on Build GRCh38
rs772094988 Chr.6: 30704904-30704904 on Build GRCh38
rs760413060 Chr.6: 30705044-30705044 on Build GRCh38
rs752681877 Chr.6: 30704816-30704816 on Build GRCh38
rs1000262648 Chr.6: 30704777-30704777 on Build GRCh38
rs770311711 Chr.6: 30704649-30704649 on Build GRCh38
rs757008913 Chr.6: 30704852-30704852 on Build GRCh38
rs780279566 Chr.6: 30704885-30704885 on Build GRCh38
rs755710749 Chr.6: 30704957-30704957 on Build GRCh38
rs201576197 Chr.6: 30704636-30704636 on Build GRCh38

View Full Product Details