Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 16352965-16353514 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MGST1
Entrez Gene ID
4257
Gene Name
microsomal glutathione S-transferase 1
Gene Aliases
GST12, MGST, MGST-I
UniGene
Hs.389700
Gene Chromosome Location
Chr.12: 16347142-16377189 on Build GRCh38
Transcripts
NM_001260511.1 NM_001260512.1 NM_001267598.1 NM_020300.4 NM_145764.2 NM_145791.2 NM_145792.2 NR_048545.1 NR_048547.1 XM_006719079.1 XM_011520673.1 XM_011520674.2

SNP Detail

SNP ID Chromosome Location
rs539764181 Chr.12: 16353324-16353324 on Build GRCh38
rs760827735 Chr.12: 16353014-16353014 on Build GRCh38
rs182561905 Chr.12: 16353335-16353335 on Build GRCh38
rs1049467175 Chr.12: 16353342-16353342 on Build GRCh38
rs888577234 Chr.12: 16353061-16353061 on Build GRCh38
rs1004174730 Chr.12: 16353471-16353471 on Build GRCh38
rs942365937 Chr.12: 16353276-16353276 on Build GRCh38
rs552448761 Chr.12: 16353402-16353402 on Build GRCh38
rs929010085 Chr.12: 16353162-16353162 on Build GRCh38
rs2975151 Chr.12: 16353047-16353047 on Build GRCh38
rs979247037 Chr.12: 16353107-16353107 on Build GRCh38
rs899895568 Chr.12: 16353034-16353034 on Build GRCh38
rs779481866 Chr.12: 16353269-16353269 on Build GRCh38
rs917581110 Chr.12: 16353141-16353141 on Build GRCh38
rs189539735 Chr.12: 16353244-16353244 on Build GRCh38
rs796203943 Chr.12: 16353089-16353089 on Build GRCh38
rs991863084 Chr.12: 16353285-16353285 on Build GRCh38
rs908886361 Chr.12: 16353326-16353326 on Build GRCh38
rs934595166 Chr.12: 16353312-16353312 on Build GRCh38
rs573719209 Chr.12: 16353425-16353425 on Build GRCh38
rs950523317 Chr.12: 16353311-16353311 on Build GRCh38
rs934334730 Chr.12: 16353047-16353047 on Build GRCh38
rs1005687918 Chr.12: 16353003-16353003 on Build GRCh38
rs991897402 Chr.12: 16353136-16353136 on Build GRCh38
rs563058032 Chr.12: 16353045-16353045 on Build GRCh38
rs900031501 Chr.12: 16353190-16353190 on Build GRCh38
rs1049478624 Chr.12: 16353056-16353056 on Build GRCh38
rs932703928 Chr.12: 16353191-16353191 on Build GRCh38
rs9332898 Chr.12: 16353341-16353341 on Build GRCh38
rs926473929 Chr.12: 16353119-16353119 on Build GRCh38
rs1009894729 Chr.12: 16353071-16353071 on Build GRCh38
rs996947490 Chr.12: 16353042-16353042 on Build GRCh38
rs552132682 Chr.12: 16353182-16353182 on Build GRCh38
rs891173617 Chr.12: 16353477-16353477 on Build GRCh38
rs1035286135 Chr.12: 16353473-16353473 on Build GRCh38
rs867836964 Chr.12: 16353352-16353352 on Build GRCh38
rs968376834 Chr.12: 16353094-16353094 on Build GRCh38
rs924536525 Chr.12: 16353260-16353260 on Build GRCh38
rs570534822 Chr.12: 16353194-16353194 on Build GRCh38
rs186733943 Chr.12: 16353201-16353201 on Build GRCh38
rs560328960 Chr.12: 16353197-16353197 on Build GRCh38
rs542455479 Chr.12: 16353064-16353064 on Build GRCh38
rs908757421 Chr.12: 16353178-16353178 on Build GRCh38
rs1005716620 Chr.12: 16353080-16353080 on Build GRCh38
rs1035409585 Chr.12: 16353144-16353144 on Build GRCh38
rs139595050 Chr.12: 16353422-16353422 on Build GRCh38
rs932838226 Chr.12: 16353407-16353407 on Build GRCh38
rs543272508 Chr.12: 16353234-16353234 on Build GRCh38
rs979035498 Chr.12: 16353232-16353232 on Build GRCh38
rs1001204078 Chr.12: 16353258-16353258 on Build GRCh38
rs531660551 Chr.12: 16353199-16353199 on Build GRCh38
rs553975645 Chr.12: 16353280-16353280 on Build GRCh38
rs560613227 Chr.12: 16353166-16353166 on Build GRCh38
rs543457709 Chr.12: 16353408-16353408 on Build GRCh38
rs897339951 Chr.12: 16353462-16353462 on Build GRCh38
rs758327737 Chr.12: 16353195-16353195 on Build GRCh38
rs765007054 Chr.12: 16353179-16353179 on Build GRCh38
rs1024647264 Chr.12: 16353303-16353303 on Build GRCh38
rs915718958 Chr.12: 16352991-16352991 on Build GRCh38
rs11056902 Chr.12: 16353371-16353371 on Build GRCh38
rs903744597 Chr.12: 16353245-16353245 on Build GRCh38
rs528013645 Chr.12: 16353167-16353167 on Build GRCh38
rs568204212 Chr.12: 16353223-16353223 on Build GRCh38
rs532271533 Chr.12: 16353284-16353284 on Build GRCh38
rs1038946065 Chr.12: 16353022-16353022 on Build GRCh38
rs968536823 Chr.12: 16353210-16353210 on Build GRCh38
rs959373978 Chr.12: 16353148-16353148 on Build GRCh38
rs565956028 Chr.12: 16353313-16353313 on Build GRCh38
rs959084940 Chr.12: 16353126-16353126 on Build GRCh38
rs932376684 Chr.12: 16353052-16353052 on Build GRCh38
rs76205046 Chr.12: 16353044-16353044 on Build GRCh38
rs768497418 Chr.12: 16353046-16353046 on Build GRCh38

View Full Product Details