Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4461563-4462060 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NMRAL1
Entrez Gene ID
57407
Gene Name
NmrA-like family domain containing 1
Gene Aliases
HSCARG, SDR48A1
UniGene
Hs.288969
Gene Chromosome Location
Chr.16: 4461677-4476320 on Build GRCh38
Transcripts
NM_001305141.1 NM_001305142.1 NM_020677.4 XM_006720905.3 XM_006720906.2 XM_017023485.1 XM_017023489.1 XM_017023490.1 XM_017023491.1 XM_017023492.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3420989 c.852G>A p.T284T Chr.16: 4461828-4461828 on Build GRCh38
327484 c.832C>A p.L278I Chr.16: 4461848-4461848 on Build GRCh38
3509644 c.746T>C p.L249P Chr.16: 4461934-4461934 on Build GRCh38
5740545 c.732G>T p.E244D Chr.16: 4461948-4461948 on Build GRCh38
3509643 c.858C>T p.D286D Chr.16: 4461822-4461822 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1020991640 Chr.16: 4461629-4461630 on Build GRCh38
rs777295138 Chr.16: 4461752-4461752 on Build GRCh38
rs912256777 Chr.16: 4461731-4461731 on Build GRCh38
rs367738608 Chr.16: 4461997-4461997 on Build GRCh38
rs543114262 Chr.16: 4461836-4461836 on Build GRCh38
rs757395113 Chr.16: 4461945-4461945 on Build GRCh38
rs368163218 Chr.16: 4461911-4461911 on Build GRCh38
rs765777318 Chr.16: 4461919-4461919 on Build GRCh38
rs936793425 Chr.16: 4462015-4462015 on Build GRCh38
rs1025516979 Chr.16: 4462028-4462028 on Build GRCh38
rs562879265 Chr.16: 4461838-4461838 on Build GRCh38
rs902557065 Chr.16: 4461821-4461821 on Build GRCh38
rs745809273 Chr.16: 4461767-4461767 on Build GRCh38
rs143114954 Chr.16: 4461808-4461808 on Build GRCh38
rs763676996 Chr.16: 4461790-4461790 on Build GRCh38
rs200609977 Chr.16: 4461990-4461990 on Build GRCh38
rs374250406 Chr.16: 4461863-4461863 on Build GRCh38
rs1019316617 Chr.16: 4462037-4462037 on Build GRCh38
rs770043309 Chr.16: 4461822-4461822 on Build GRCh38
rs568929481 Chr.16: 4461688-4461688 on Build GRCh38
rs767503025 Chr.16: 4461991-4461991 on Build GRCh38
rs1003391554 Chr.16: 4461601-4461601 on Build GRCh38
rs74005327 Chr.16: 4461987-4461987 on Build GRCh38
rs1037739699 Chr.16: 4461989-4461989 on Build GRCh38
rs553613931 Chr.16: 4462030-4462030 on Build GRCh38
rs771402910 Chr.16: 4461828-4461828 on Build GRCh38
rs774987013 Chr.16: 4461884-4461884 on Build GRCh38
rs747436162 Chr.16: 4461969-4461969 on Build GRCh38
rs200534634 Chr.16: 4461742-4461742 on Build GRCh38
rs141206910 Chr.16: 4461783-4461783 on Build GRCh38
rs776866806 Chr.16: 4461772-4461772 on Build GRCh38
rs182821752 Chr.16: 4461600-4461600 on Build GRCh38
rs148206252 Chr.16: 4461613-4461613 on Build GRCh38
rs764890767 Chr.16: 4461913-4461913 on Build GRCh38
rs368378735 Chr.16: 4461765-4461765 on Build GRCh38
rs778358522 Chr.16: 4461799-4461799 on Build GRCh38
rs374990585 Chr.16: 4462004-4462004 on Build GRCh38
rs141298817 Chr.16: 4461861-4461861 on Build GRCh38
rs769992191 Chr.16: 4461778-4461778 on Build GRCh38
rs534569403 Chr.16: 4461771-4461771 on Build GRCh38
rs148278703 Chr.16: 4461816-4461816 on Build GRCh38
rs1054279561 Chr.16: 4462027-4462027 on Build GRCh38
rs754915086 Chr.16: 4461924-4461924 on Build GRCh38
rs1043380 Chr.16: 4461891-4461891 on Build GRCh38
rs763295684 Chr.16: 4461747-4461747 on Build GRCh38
rs765702607 Chr.16: 4461915-4461921 on Build GRCh38
rs761429723 Chr.16: 4461847-4461847 on Build GRCh38
rs772470093 Chr.16: 4461895-4461895 on Build GRCh38
rs185503301 Chr.16: 4461981-4461981 on Build GRCh38
rs776816993 Chr.16: 4461735-4461735 on Build GRCh38
rs750142956 Chr.16: 4461879-4461879 on Build GRCh38
rs775849892 Chr.16: 4461968-4461968 on Build GRCh38
rs147175262 Chr.16: 4461917-4461917 on Build GRCh38
rs765784562 Chr.16: 4461740-4461740 on Build GRCh38
rs762417179 Chr.16: 4461736-4461736 on Build GRCh38
rs569724456 Chr.16: 4461632-4461632 on Build GRCh38
rs141564520 Chr.16: 4461867-4461867 on Build GRCh38
rs762043314 Chr.16: 4461773-4461773 on Build GRCh38
rs942864867 Chr.16: 4461757-4461757 on Build GRCh38
rs773633426 Chr.16: 4461779-4461779 on Build GRCh38
rs1035764047 Chr.16: 4461623-4461623 on Build GRCh38
rs567740340 Chr.16: 4462029-4462029 on Build GRCh38
rs903180286 Chr.16: 4461594-4461594 on Build GRCh38
rs778113907 Chr.16: 4461935-4461935 on Build GRCh38
rs775780185 Chr.16: 4461896-4461896 on Build GRCh38
rs1032676007 Chr.16: 4461693-4461693 on Build GRCh38
rs746763615 Chr.16: 4461811-4461811 on Build GRCh38
rs938690562 Chr.16: 4461980-4461980 on Build GRCh38
rs953959401 Chr.16: 4461723-4461723 on Build GRCh38
rs145081403 Chr.16: 4461789-4461789 on Build GRCh38
rs753903089 Chr.16: 4461777-4461790 on Build GRCh38
rs764688115 Chr.16: 4461848-4461848 on Build GRCh38
rs756363068 Chr.16: 4461876-4461876 on Build GRCh38
rs746341030 Chr.16: 4461892-4461892 on Build GRCh38
rs143394570 Chr.16: 4461916-4461916 on Build GRCh38
rs1032205486 Chr.16: 4461841-4461841 on Build GRCh38
rs773295242 Chr.16: 4461825-4461825 on Build GRCh38
rs3747582 Chr.16: 4461925-4461925 on Build GRCh38
rs779853896 Chr.16: 4461770-4461770 on Build GRCh38
rs746392355 Chr.16: 4461958-4461958 on Build GRCh38
rs751598351 Chr.16: 4461923-4461923 on Build GRCh38
rs771799219 Chr.16: 4461768-4461768 on Build GRCh38
rs761947746 Chr.16: 4461813-4461814 on Build GRCh38
rs752472708 Chr.16: 4461926-4461926 on Build GRCh38
rs750372088 Chr.16: 4461800-4461800 on Build GRCh38
rs760584712 Chr.16: 4462000-4462000 on Build GRCh38
rs756418093 Chr.16: 4461928-4461928 on Build GRCh38
rs781112929 Chr.16: 4461871-4461871 on Build GRCh38
rs149409840 Chr.16: 4461941-4461941 on Build GRCh38
rs772504761 Chr.16: 4461966-4461966 on Build GRCh38
rs774591050 Chr.16: 4461829-4461829 on Build GRCh38
rs986441759 Chr.16: 4461897-4461897 on Build GRCh38
rs748351440 Chr.16: 4461818-4461818 on Build GRCh38
rs973958424 Chr.16: 4461727-4461727 on Build GRCh38
rs754135251 Chr.16: 4462005-4462005 on Build GRCh38
rs752342281 Chr.16: 4461749-4461749 on Build GRCh38
rs758675629 Chr.16: 4461668-4461668 on Build GRCh38
rs7191458 Chr.16: 4461751-4461751 on Build GRCh38
rs765085922 Chr.16: 4461651-4461652 on Build GRCh38
rs990632336 Chr.16: 4461648-4461648 on Build GRCh38
rs569677625 Chr.16: 4462019-4462019 on Build GRCh38
rs778877498 Chr.16: 4461766-4461766 on Build GRCh38
rs770499447 Chr.16: 4461986-4461986 on Build GRCh38
rs202218846 Chr.16: 4461872-4461872 on Build GRCh38
rs758319453 Chr.16: 4461807-4461807 on Build GRCh38
rs973991204 Chr.16: 4461854-4461854 on Build GRCh38
rs960842584 Chr.16: 4461646-4461646 on Build GRCh38
rs763400756 Chr.16: 4461827-4461827 on Build GRCh38
rs370619031 Chr.16: 4461864-4461864 on Build GRCh38
rs144759756 Chr.16: 4461948-4461948 on Build GRCh38
rs777718263 Chr.16: 4461878-4461878 on Build GRCh38
rs377501302 Chr.16: 4461628-4461628 on Build GRCh38
rs369306379 Chr.16: 4461748-4461748 on Build GRCh38
rs1023984523 Chr.16: 4461667-4461667 on Build GRCh38
rs1042004045 Chr.16: 4461942-4461942 on Build GRCh38

View Full Product Details