Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 96986120-96986669 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OXGR1
Entrez Gene ID
27199
Gene Name
oxoglutarate receptor 1
Gene Aliases
GPR80, GPR99, P2RY15, P2Y15, aKGR
UniGene
Hs.352218
Gene Chromosome Location
Chr.13: 96985719-96995842 on Build GRCh38
Transcripts
NM_080818.3 XM_005254043.3 XM_005254044.3 XM_005254045.2 XM_005254046.2 XM_011521077.2

SNP Detail

SNP ID Chromosome Location
rs1025356440 Chr.13: 96986364-96986364 on Build GRCh38
rs1033885185 Chr.13: 96986354-96986354 on Build GRCh38
rs1021782553 Chr.13: 96986159-96986159 on Build GRCh38
rs1053844698 Chr.13: 96986628-96986628 on Build GRCh38
rs1020267741 Chr.13: 96986355-96986355 on Build GRCh38
rs981988606 Chr.13: 96986459-96986459 on Build GRCh38
rs893785606 Chr.13: 96986279-96986279 on Build GRCh38
rs943449051 Chr.13: 96986596-96986596 on Build GRCh38
rs973090369 Chr.13: 96986470-96986470 on Build GRCh38
rs986118246 Chr.13: 96986200-96986200 on Build GRCh38
rs907176114 Chr.13: 96986270-96986270 on Build GRCh38
rs146502547 Chr.13: 96986154-96986154 on Build GRCh38
rs774198882 Chr.13: 96986232-96986232 on Build GRCh38
rs965437404 Chr.13: 96986207-96986207 on Build GRCh38
rs913236927 Chr.13: 96986607-96986607 on Build GRCh38
rs933859407 Chr.13: 96986527-96986527 on Build GRCh38
rs889205227 Chr.13: 96986250-96986250 on Build GRCh38
rs947439639 Chr.13: 96986616-96986616 on Build GRCh38
rs1043552705 Chr.13: 96986617-96986617 on Build GRCh38
rs564543279 Chr.13: 96986223-96986223 on Build GRCh38
rs61975469 Chr.13: 96986149-96986149 on Build GRCh38
rs188156899 Chr.13: 96986212-96986212 on Build GRCh38
rs1021707605 Chr.13: 96986315-96986315 on Build GRCh38
rs945344600 Chr.13: 96986643-96986643 on Build GRCh38
rs552865865 Chr.13: 96986575-96986575 on Build GRCh38
rs61368531 Chr.13: 96986152-96986152 on Build GRCh38
rs939948347 Chr.13: 96986173-96986173 on Build GRCh38
rs1009839689 Chr.13: 96986281-96986281 on Build GRCh38
rs954316439 Chr.13: 96986172-96986172 on Build GRCh38
rs141110769 Chr.13: 96986231-96986231 on Build GRCh38
rs1053076139 Chr.13: 96986283-96986283 on Build GRCh38
rs985315495 Chr.13: 96986566-96986566 on Build GRCh38
rs889958712 Chr.13: 96986342-96986342 on Build GRCh38
rs1019250000 Chr.13: 96986202-96986202 on Build GRCh38
rs759106033 Chr.13: 96986239-96986239 on Build GRCh38
rs1021185951 Chr.13: 96986302-96986302 on Build GRCh38
rs891873488 Chr.13: 96986296-96986296 on Build GRCh38
rs927855692 Chr.13: 96986492-96986492 on Build GRCh38
rs1011656909 Chr.13: 96986298-96986298 on Build GRCh38
rs17301222 Chr.13: 96986372-96986372 on Build GRCh38
rs965686739 Chr.13: 96986357-96986357 on Build GRCh38
rs918328095 Chr.13: 96986215-96986215 on Build GRCh38
rs751914089 Chr.13: 96986543-96986543 on Build GRCh38
rs571738116 Chr.13: 96986299-96986299 on Build GRCh38
rs1002930030 Chr.13: 96986151-96986151 on Build GRCh38
rs922537767 Chr.13: 96986487-96986487 on Build GRCh38
rs192738081 Chr.13: 96986391-96986391 on Build GRCh38
rs551455919 Chr.13: 96986259-96986259 on Build GRCh38
rs184285545 Chr.13: 96986557-96986557 on Build GRCh38
rs1011686691 Chr.13: 96986157-96986157 on Build GRCh38
rs972386164 Chr.13: 96986362-96986362 on Build GRCh38
rs1053045451 Chr.13: 96986586-96986586 on Build GRCh38
rs756091534 Chr.13: 96986538-96986538 on Build GRCh38
rs983880697 Chr.13: 96986381-96986381 on Build GRCh38
rs931086515 Chr.13: 96986218-96986218 on Build GRCh38
rs906722226 Chr.13: 96986332-96986332 on Build GRCh38
rs138296283 Chr.13: 96986181-96986181 on Build GRCh38
rs529664015 Chr.13: 96986183-96986183 on Build GRCh38
rs554176377 Chr.13: 96986423-96986423 on Build GRCh38
rs1016567621 Chr.13: 96986454-96986454 on Build GRCh38
rs573026807 Chr.13: 96986629-96986629 on Build GRCh38
rs537131322 Chr.13: 96986350-96986350 on Build GRCh38
rs879207911 Chr.13: 96986356-96986356 on Build GRCh38
rs1044366053 Chr.13: 96986148-96986148 on Build GRCh38
rs911026917 Chr.13: 96986582-96986582 on Build GRCh38
rs767206946 Chr.13: 96986276-96986276 on Build GRCh38
rs935282973 Chr.13: 96986580-96986580 on Build GRCh38

View Full Product Details