Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 147642870-147643405 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRMT9
Entrez Gene ID
90826
Gene Name
protein arginine methyltransferase 9
Gene Aliases
PRMT10
UniGene
Hs.591692
Gene Chromosome Location
Chr.4: 147635626-147684230 on Build GRCh38
Transcripts
NM_001304458.1 NM_138364.3 XM_017008823.1 XM_017008824.1 XM_017008825.1 XM_017008826.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1169013 c.2083T>G p.Y695D Chr.4: 147642903-147642903 on Build GRCh38
1427429 c.2092A>C p.M698L Chr.4: 147642894-147642894 on Build GRCh38
1051852 c.2086G>A p.V696M Chr.4: 147642900-147642900 on Build GRCh38
447459 c.2080C>T p.Q694* Chr.4: 147642906-147642906 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs2654943 Chr.4: 147643034-147643034 on Build GRCh38
rs778552755 Chr.4: 147642991-147642991 on Build GRCh38
rs963877054 Chr.4: 147643364-147643364 on Build GRCh38
rs75306154 Chr.4: 147643210-147643210 on Build GRCh38
rs901142059 Chr.4: 147643283-147643283 on Build GRCh38
rs913332851 Chr.4: 147643103-147643103 on Build GRCh38
rs1018087447 Chr.4: 147643360-147643360 on Build GRCh38
rs759855439 Chr.4: 147642957-147642957 on Build GRCh38
rs767801803 Chr.4: 147642967-147642967 on Build GRCh38
rs112534940 Chr.4: 147642896-147642896 on Build GRCh38
rs746354757 Chr.4: 147643157-147643157 on Build GRCh38
rs374007128 Chr.4: 147642984-147642984 on Build GRCh38
rs911141837 Chr.4: 147643130-147643130 on Build GRCh38
rs2654944 Chr.4: 147643011-147643011 on Build GRCh38
rs553860562 Chr.4: 147643027-147643027 on Build GRCh38
rs891543468 Chr.4: 147643224-147643224 on Build GRCh38
rs978586338 Chr.4: 147643347-147643347 on Build GRCh38
rs2654942 Chr.4: 147643087-147643087 on Build GRCh38
rs76813592 Chr.4: 147643218-147643218 on Build GRCh38
rs564876398 Chr.4: 147643143-147643143 on Build GRCh38
rs942629690 Chr.4: 147643136-147643136 on Build GRCh38
rs929676756 Chr.4: 147643168-147643168 on Build GRCh38
rs994174633 Chr.4: 147643286-147643287 on Build GRCh38
rs1025744608 Chr.4: 147643031-147643031 on Build GRCh38
rs1025690845 Chr.4: 147643287-147643287 on Build GRCh38
rs773815889 Chr.4: 147642944-147642944 on Build GRCh38
rs900908475 Chr.4: 147642919-147642919 on Build GRCh38
rs544900211 Chr.4: 147643105-147643105 on Build GRCh38
rs533810028 Chr.4: 147642955-147642955 on Build GRCh38
rs766543084 Chr.4: 147642950-147642950 on Build GRCh38
rs922223079 Chr.4: 147643144-147643144 on Build GRCh38
rs750434508 Chr.4: 147642992-147642992 on Build GRCh38
rs765547898 Chr.4: 147643236-147643236 on Build GRCh38
rs775448359 Chr.4: 147642925-147642925 on Build GRCh38
rs965386962 Chr.4: 147643000-147643000 on Build GRCh38
rs189470384 Chr.4: 147643273-147643273 on Build GRCh38
rs779562930 Chr.4: 147642930-147642931 on Build GRCh38
rs1007194338 Chr.4: 147643275-147643279 on Build GRCh38
rs763351937 Chr.4: 147642946-147642946 on Build GRCh38
rs191683189 Chr.4: 147643374-147643374 on Build GRCh38
rs762123829 Chr.4: 147642917-147642917 on Build GRCh38
rs2718459 Chr.4: 147643071-147643071 on Build GRCh38
rs978617618 Chr.4: 147643111-147643111 on Build GRCh38
rs552080117 Chr.4: 147643239-147643239 on Build GRCh38
rs750874827 Chr.4: 147643376-147643376 on Build GRCh38
rs888232529 Chr.4: 147643211-147643211 on Build GRCh38
rs935623228 Chr.4: 147643381-147643381 on Build GRCh38
rs948293066 Chr.4: 147643126-147643126 on Build GRCh38
rs549490871 Chr.4: 147643303-147643303 on Build GRCh38
rs746731362 Chr.4: 147642892-147642892 on Build GRCh38
rs1020771684 Chr.4: 147643297-147643297 on Build GRCh38
rs765484865 Chr.4: 147642929-147642929 on Build GRCh38
rs768239073 Chr.4: 147642908-147642908 on Build GRCh38
rs368384406 Chr.4: 147643078-147643078 on Build GRCh38
rs1046829210 Chr.4: 147642943-147642943 on Build GRCh38
rs2654945 Chr.4: 147643009-147643009 on Build GRCh38
rs149106006 Chr.4: 147643032-147643032 on Build GRCh38
rs1037872962 Chr.4: 147643223-147643223 on Build GRCh38
rs968872729 Chr.4: 147643329-147643329 on Build GRCh38
rs895779474 Chr.4: 147643286-147643286 on Build GRCh38
rs947190804 Chr.4: 147643229-147643229 on Build GRCh38
rs1019554693 Chr.4: 147643282-147643286 on Build GRCh38
rs143157060 Chr.4: 147643081-147643081 on Build GRCh38
rs377560741 Chr.4: 147642985-147642985 on Build GRCh38
rs867241324 Chr.4: 147643152-147643152 on Build GRCh38
rs1016462901 Chr.4: 147642995-147642995 on Build GRCh38
rs969602193 Chr.4: 147643343-147643343 on Build GRCh38
rs138660469 Chr.4: 147643295-147643295 on Build GRCh38
rs370967368 Chr.4: 147642915-147642915 on Build GRCh38
rs536414884 Chr.4: 147643066-147643066 on Build GRCh38
rs1035904894 Chr.4: 147643281-147643281 on Build GRCh38
rs906115971 Chr.4: 147643259-147643259 on Build GRCh38

View Full Product Details