Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 38824144-38824689 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TLR6
Entrez Gene ID
10333
Gene Name
toll like receptor 6
Gene Aliases
CD286
UniGene
Hs.575090
Gene Chromosome Location
Chr.4: 38822897-38857767 on Build GRCh38
Transcripts
NM_006068.4 XM_005262637.4 XM_011513612.2 XM_011513613.2 XM_011513614.2

SNP Detail

SNP ID Chromosome Location
rs375725711 Chr.4: 38824201-38824201 on Build GRCh38
rs574861725 Chr.4: 38824436-38824436 on Build GRCh38
rs73808601 Chr.4: 38824623-38824623 on Build GRCh38
rs765831085 Chr.4: 38824352-38824352 on Build GRCh38
rs974978653 Chr.4: 38824555-38824555 on Build GRCh38
rs959977089 Chr.4: 38824453-38824453 on Build GRCh38
rs527552150 Chr.4: 38824566-38824566 on Build GRCh38
rs905677863 Chr.4: 38824381-38824381 on Build GRCh38
rs151312242 Chr.4: 38824562-38824563 on Build GRCh38
rs376254476 Chr.4: 38824591-38824591 on Build GRCh38
rs943515625 Chr.4: 38824518-38824518 on Build GRCh38
rs943562453 Chr.4: 38824169-38824169 on Build GRCh38
rs970089982 Chr.4: 38824376-38824376 on Build GRCh38
rs113972196 Chr.4: 38824284-38824284 on Build GRCh38
rs1016368872 Chr.4: 38824322-38824322 on Build GRCh38
rs1001765620 Chr.4: 38824631-38824631 on Build GRCh38
rs183697322 Chr.4: 38824514-38824514 on Build GRCh38
rs562579840 Chr.4: 38824444-38824444 on Build GRCh38
rs942050695 Chr.4: 38824595-38824595 on Build GRCh38
rs543892924 Chr.4: 38824440-38824440 on Build GRCh38
rs547451646 Chr.4: 38824584-38824584 on Build GRCh38
rs142339799 Chr.4: 38824222-38824222 on Build GRCh38
rs144408528 Chr.4: 38824508-38824508 on Build GRCh38
rs759607780 Chr.4: 38824261-38824291 on Build GRCh38
rs933111578 Chr.4: 38824208-38824208 on Build GRCh38
rs1053800875 Chr.4: 38824267-38824267 on Build GRCh38
rs6822503 Chr.4: 38824455-38824455 on Build GRCh38
rs902117360 Chr.4: 38824180-38824180 on Build GRCh38
rs572376647 Chr.4: 38824217-38824217 on Build GRCh38
rs139542236 Chr.4: 38824303-38824304 on Build GRCh38
rs933542846 Chr.4: 38824569-38824569 on Build GRCh38
rs543939613 Chr.4: 38824291-38824292 on Build GRCh38
rs921677136 Chr.4: 38824198-38824198 on Build GRCh38
rs1039682246 Chr.4: 38824179-38824179 on Build GRCh38
rs1019922376 Chr.4: 38824474-38824474 on Build GRCh38
rs201615840 Chr.4: 38824563-38824563 on Build GRCh38
rs202032371 Chr.4: 38824588-38824588 on Build GRCh38
rs886420754 Chr.4: 38824586-38824586 on Build GRCh38
rs565208201 Chr.4: 38824534-38824534 on Build GRCh38
rs34095756 Chr.4: 38824544-38824545 on Build GRCh38
rs1011573210 Chr.4: 38824289-38824289 on Build GRCh38
rs188620339 Chr.4: 38824606-38824606 on Build GRCh38
rs905985330 Chr.4: 38824624-38824624 on Build GRCh38
rs978477031 Chr.4: 38824511-38824511 on Build GRCh38
rs1003375382 Chr.4: 38824385-38824385 on Build GRCh38
rs1051963347 Chr.4: 38824586-38824588 on Build GRCh38
rs768363953 Chr.4: 38824191-38824191 on Build GRCh38
rs764987392 Chr.4: 38824554-38824555 on Build GRCh38
rs73236630 Chr.4: 38824434-38824434 on Build GRCh38
rs772660457 Chr.4: 38824519-38824519 on Build GRCh38
rs999087841 Chr.4: 38824250-38824250 on Build GRCh38
rs778727707 Chr.4: 38824410-38824410 on Build GRCh38
rs1001397900 Chr.4: 38824382-38824382 on Build GRCh38
rs1035493060 Chr.4: 38824661-38824661 on Build GRCh38
rs1006225328 Chr.4: 38824298-38824298 on Build GRCh38

View Full Product Details