Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143650019-143650520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF623
Entrez Gene ID
9831
Gene Name
zinc finger protein 623
Gene Aliases
-
UniGene
Hs.43133
Gene Chromosome Location
Chr.8: 143636013-143653730 on Build GRCh38
Transcripts
NM_001082480.2 NM_001261843.1 NM_014789.3 XM_006716708.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
324527 c.474A>T p.K158N Chr.8: 143650346-143650346 on Build GRCh38
4338690 c.328_330delCTT p.L113delL Chr.8: 143650200-143650202 on Build GRCh38
3367236 c.625T>A p.C209S Chr.8: 143650497-143650497 on Build GRCh38
5788067 c.580C>G p.L194V Chr.8: 143650452-143650452 on Build GRCh38
7121032 c.637G>T p.G213W Chr.8: 143650509-143650509 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769530838 Chr.8: 143650082-143650082 on Build GRCh38
rs776317051 Chr.8: 143650328-143650328 on Build GRCh38
rs137936747 Chr.8: 143650164-143650164 on Build GRCh38
rs139397709 Chr.8: 143650185-143650185 on Build GRCh38
rs868205639 Chr.8: 143650373-143650373 on Build GRCh38
rs1022885143 Chr.8: 143650232-143650232 on Build GRCh38
rs966855865 Chr.8: 143650242-143650242 on Build GRCh38
rs781570540 Chr.8: 143650441-143650441 on Build GRCh38
rs141901810 Chr.8: 143650149-143650149 on Build GRCh38
rs754755147 Chr.8: 143650372-143650372 on Build GRCh38
rs530519666 Chr.8: 143650271-143650271 on Build GRCh38
rs558228017 Chr.8: 143650059-143650059 on Build GRCh38
rs756731274 Chr.8: 143650404-143650405 on Build GRCh38
rs577628297 Chr.8: 143650468-143650468 on Build GRCh38
rs768217570 Chr.8: 143650088-143650088 on Build GRCh38
rs752765590 Chr.8: 143650136-143650136 on Build GRCh38
rs760976001 Chr.8: 143650208-143650208 on Build GRCh38
rs770007205 Chr.8: 143650327-143650327 on Build GRCh38
rs867834005 Chr.8: 143650353-143650353 on Build GRCh38
rs752033972 Chr.8: 143650286-143650286 on Build GRCh38
rs1037578505 Chr.8: 143650304-143650304 on Build GRCh38
rs898915922 Chr.8: 143650068-143650068 on Build GRCh38
rs752107502 Chr.8: 143650098-143650099 on Build GRCh38
rs766269967 Chr.8: 143650363-143650363 on Build GRCh38
rs760669056 Chr.8: 143650341-143650341 on Build GRCh38
rs144172822 Chr.8: 143650332-143650332 on Build GRCh38
rs200227957 Chr.8: 143650090-143650090 on Build GRCh38
rs372484982 Chr.8: 143650200-143650200 on Build GRCh38
rs374072601 Chr.8: 143650102-143650102 on Build GRCh38
rs776915019 Chr.8: 143650213-143650213 on Build GRCh38
rs773538441 Chr.8: 143650104-143650104 on Build GRCh38
rs888389550 Chr.8: 143650391-143650391 on Build GRCh38
rs759779396 Chr.8: 143650225-143650225 on Build GRCh38
rs561105190 Chr.8: 143650292-143650292 on Build GRCh38
rs747584247 Chr.8: 143650460-143650460 on Build GRCh38
rs144210785 Chr.8: 143650501-143650501 on Build GRCh38
rs942035655 Chr.8: 143650436-143650436 on Build GRCh38
rs4875017 Chr.8: 143650193-143650193 on Build GRCh38
rs773734672 Chr.8: 143650206-143650206 on Build GRCh38
rs771046141 Chr.8: 143650047-143650047 on Build GRCh38
rs1036200252 Chr.8: 143650497-143650497 on Build GRCh38
rs367972475 Chr.8: 143650189-143650189 on Build GRCh38
rs201943513 Chr.8: 143650283-143650283 on Build GRCh38
rs746559684 Chr.8: 143650427-143650427 on Build GRCh38
rs779506484 Chr.8: 143650168-143650168 on Build GRCh38
rs867324263 Chr.8: 143650405-143650405 on Build GRCh38
rs766293687 Chr.8: 143650484-143650484 on Build GRCh38
rs755218294 Chr.8: 143650127-143650127 on Build GRCh38
rs377222268 Chr.8: 143650470-143650470 on Build GRCh38
rs752780381 Chr.8: 143650247-143650247 on Build GRCh38
rs758101079 Chr.8: 143650418-143650418 on Build GRCh38
rs139398523 Chr.8: 143650331-143650331 on Build GRCh38
rs929812413 Chr.8: 143650368-143650368 on Build GRCh38
rs989051355 Chr.8: 143650266-143650266 on Build GRCh38
rs777239594 Chr.8: 143650421-143650421 on Build GRCh38
rs200418048 Chr.8: 143650335-143650335 on Build GRCh38
rs770057782 Chr.8: 143650456-143650456 on Build GRCh38
rs753036328 Chr.8: 143650255-143650255 on Build GRCh38
rs148516981 Chr.8: 143650469-143650469 on Build GRCh38
rs765343191 Chr.8: 143650133-143650133 on Build GRCh38
rs763251196 Chr.8: 143650142-143650144 on Build GRCh38
rs764537153 Chr.8: 143650200-143650202 on Build GRCh38
rs3750204 Chr.8: 143650280-143650280 on Build GRCh38
rs757227153 Chr.8: 143650148-143650148 on Build GRCh38
rs766543889 Chr.8: 143650113-143650113 on Build GRCh38
rs534934074 Chr.8: 143650321-143650321 on Build GRCh38
rs762370655 Chr.8: 143650097-143650097 on Build GRCh38
rs761062554 Chr.8: 143650109-143650109 on Build GRCh38
rs746185206 Chr.8: 143650444-143650444 on Build GRCh38
rs778573982 Chr.8: 143650409-143650409 on Build GRCh38
rs753631510 Chr.8: 143650371-143650371 on Build GRCh38
rs768252678 Chr.8: 143650183-143650183 on Build GRCh38
rs993650640 Chr.8: 143650074-143650074 on Build GRCh38
rs146930318 Chr.8: 143650160-143650160 on Build GRCh38
rs771388232 Chr.8: 143650313-143650313 on Build GRCh38
rs748689928 Chr.8: 143650085-143650085 on Build GRCh38
rs150073213 Chr.8: 143650318-143650318 on Build GRCh38
rs200935028 Chr.8: 143650163-143650163 on Build GRCh38
rs201124540 Chr.8: 143650056-143650056 on Build GRCh38
rs372174669 Chr.8: 143650467-143650467 on Build GRCh38
rs146294757 Chr.8: 143650410-143650410 on Build GRCh38
rs4874084 Chr.8: 143650248-143650248 on Build GRCh38
rs779834650 Chr.8: 143650296-143650296 on Build GRCh38
rs375460182 Chr.8: 143650270-143650270 on Build GRCh38
rs768063442 Chr.8: 143650098-143650098 on Build GRCh38
rs776064066 Chr.8: 143650464-143650464 on Build GRCh38
rs746594770 Chr.8: 143650309-143650309 on Build GRCh38
rs750391222 Chr.8: 143650250-143650250 on Build GRCh38
rs1003443504 Chr.8: 143650178-143650178 on Build GRCh38
rs766721710 Chr.8: 143650210-143650210 on Build GRCh38
rs748966963 Chr.8: 143650172-143650172 on Build GRCh38
rs138574970 Chr.8: 143650279-143650279 on Build GRCh38
rs771427452 Chr.8: 143650440-143650440 on Build GRCh38
rs746958559 Chr.8: 143650043-143650043 on Build GRCh38
rs747498050 Chr.8: 143650297-143650297 on Build GRCh38
rs201415801 Chr.8: 143650238-143650238 on Build GRCh38
rs370100274 Chr.8: 143650360-143650360 on Build GRCh38
rs754090581 Chr.8: 143650124-143650124 on Build GRCh38

View Full Product Details