Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 170151070-170151573 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
METTL11B
Entrez Gene ID
149281
Gene Name
methyltransferase like 11B
Gene Aliases
C1orf184, HOMT1B, NTM1B
UniGene
Hs.553612
Gene Chromosome Location
Chr.1: 170146047-170168841 on Build GRCh38
Transcripts
NM_001136107.1 XM_011509232.2 XM_011509233.2 NR_036064.1 NR_036065.1

Gene Symbol
MIR3119-1
Entrez Gene ID
100422839
Gene Name
microRNA 3119-1
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.1: 170151378-170151462 on Build GRCh38
Transcripts
NM_001136107.1 XM_011509232.2 XM_011509233.2 NR_036064.1 NR_036065.1

Gene Symbol
MIR3119-2
Entrez Gene ID
100423010
Gene Name
microRNA 3119-2
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.1: 170151378-170151462 on Build GRCh38
Transcripts
NM_001136107.1 XM_011509232.2 XM_011509233.2 NR_036064.1 NR_036065.1

SNP Detail

SNP ID Chromosome Location
rs762956001 Chr.1: 170151169-170151169 on Build GRCh38
rs58602811 Chr.1: 170151434-170151434 on Build GRCh38
rs1003501265 Chr.1: 170151310-170151310 on Build GRCh38
rs959056941 Chr.1: 170151320-170151320 on Build GRCh38
rs1001999766 Chr.1: 170151336-170151336 on Build GRCh38
rs145826669 Chr.1: 170151319-170151319 on Build GRCh38
rs369864391 Chr.1: 170151177-170151177 on Build GRCh38
rs532260117 Chr.1: 170151160-170151160 on Build GRCh38
rs754589297 Chr.1: 170151332-170151332 on Build GRCh38
rs759046951 Chr.1: 170151358-170151358 on Build GRCh38
rs531031268 Chr.1: 170151159-170151159 on Build GRCh38
rs976695651 Chr.1: 170151454-170151454 on Build GRCh38
rs924732934 Chr.1: 170151386-170151386 on Build GRCh38
rs989130523 Chr.1: 170151338-170151338 on Build GRCh38
rs751132456 Chr.1: 170151298-170151298 on Build GRCh38
rs915017306 Chr.1: 170151339-170151339 on Build GRCh38
rs147804112 Chr.1: 170151500-170151500 on Build GRCh38
rs1015475182 Chr.1: 170151413-170151413 on Build GRCh38
rs966835685 Chr.1: 170151350-170151350 on Build GRCh38
rs76123652 Chr.1: 170151307-170151307 on Build GRCh38
rs1045693178 Chr.1: 170151276-170151276 on Build GRCh38
rs950345400 Chr.1: 170151216-170151216 on Build GRCh38
rs546053768 Chr.1: 170151330-170151330 on Build GRCh38
rs556174849 Chr.1: 170151481-170151481 on Build GRCh38
rs538527 Chr.1: 170151376-170151376 on Build GRCh38
rs537840374 Chr.1: 170151453-170151453 on Build GRCh38
rs372828964 Chr.1: 170151225-170151225 on Build GRCh38
rs767136312 Chr.1: 170151477-170151477 on Build GRCh38
rs1029304623 Chr.1: 170151465-170151465 on Build GRCh38
rs892041671 Chr.1: 170151196-170151196 on Build GRCh38
rs1026537378 Chr.1: 170151170-170151170 on Build GRCh38
rs977628513 Chr.1: 170151381-170151381 on Build GRCh38
rs765752812 Chr.1: 170151228-170151228 on Build GRCh38
rs888241732 Chr.1: 170151359-170151359 on Build GRCh38
rs529908933 Chr.1: 170151340-170151340 on Build GRCh38
rs765106040 Chr.1: 170151107-170151107 on Build GRCh38
rs185710757 Chr.1: 170151425-170151425 on Build GRCh38
rs889153326 Chr.1: 170151112-170151112 on Build GRCh38
rs1053747958 Chr.1: 170151164-170151164 on Build GRCh38
rs933485949 Chr.1: 170151428-170151428 on Build GRCh38
rs1007506346 Chr.1: 170151134-170151134 on Build GRCh38

View Full Product Details