Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 117488432-117488944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMX2OS
Entrez Gene ID
196047
Gene Name
EMX2 opposite strand/antisense RNA
Gene Aliases
EMX2-AS1, NCRNA00045
UniGene
Hs.312592
Gene Chromosome Location
Chr.10: 117484293-117545068 on Build GRCh38
Transcripts
NR_002791.2

SNP Detail

SNP ID Chromosome Location
rs939725346 Chr.10: 117488621-117488621 on Build GRCh38
rs947674844 Chr.10: 117488743-117488743 on Build GRCh38
rs1011514529 Chr.10: 117488462-117488462 on Build GRCh38
rs547491948 Chr.10: 117488640-117488640 on Build GRCh38
rs540856686 Chr.10: 117488830-117488830 on Build GRCh38
rs908245044 Chr.10: 117488604-117488604 on Build GRCh38
rs768865256 Chr.10: 117488616-117488616 on Build GRCh38
rs776661487 Chr.10: 117488773-117488773 on Build GRCh38
rs183983123 Chr.10: 117488744-117488744 on Build GRCh38
rs2768328 Chr.10: 117488559-117488559 on Build GRCh38
rs967859269 Chr.10: 117488551-117488551 on Build GRCh38
rs578126298 Chr.10: 117488592-117488592 on Build GRCh38
rs1004255819 Chr.10: 117488705-117488705 on Build GRCh38
rs11198012 Chr.10: 117488599-117488599 on Build GRCh38
rs368319006 Chr.10: 117488647-117488647 on Build GRCh38
rs188657449 Chr.10: 117488795-117488795 on Build GRCh38
rs903188172 Chr.10: 117488766-117488766 on Build GRCh38
rs758438836 Chr.10: 117488582-117488582 on Build GRCh38
rs776126377 Chr.10: 117488861-117488861 on Build GRCh38
rs530016327 Chr.10: 117488476-117488476 on Build GRCh38
rs769762354 Chr.10: 117488782-117488782 on Build GRCh38
rs567401909 Chr.10: 117488642-117488642 on Build GRCh38
rs547225034 Chr.10: 117488776-117488776 on Build GRCh38
rs569807220 Chr.10: 117488700-117488700 on Build GRCh38
rs552453964 Chr.10: 117488589-117488589 on Build GRCh38
rs988803118 Chr.10: 117488602-117488602 on Build GRCh38
rs969083000 Chr.10: 117488879-117488879 on Build GRCh38
rs140014707 Chr.10: 117488779-117488779 on Build GRCh38
rs543834671 Chr.10: 117488496-117488496 on Build GRCh38
rs2768329 Chr.10: 117488600-117488600 on Build GRCh38
rs545005126 Chr.10: 117488583-117488583 on Build GRCh38
rs934075172 Chr.10: 117488687-117488687 on Build GRCh38
rs535864093 Chr.10: 117488653-117488653 on Build GRCh38
rs1023328325 Chr.10: 117488524-117488524 on Build GRCh38
rs542280187 Chr.10: 117488909-117488909 on Build GRCh38
rs192188774 Chr.10: 117488696-117488696 on Build GRCh38
rs1003256296 Chr.10: 117488882-117488882 on Build GRCh38
rs952602005 Chr.10: 117488908-117488908 on Build GRCh38
rs777513313 Chr.10: 117488622-117488622 on Build GRCh38
rs946859189 Chr.10: 117488638-117488638 on Build GRCh38
rs1022837274 Chr.10: 117488842-117488842 on Build GRCh38
rs915536820 Chr.10: 117488603-117488603 on Build GRCh38
rs11198011 Chr.10: 117488578-117488578 on Build GRCh38
rs1035893509 Chr.10: 117488560-117488560 on Build GRCh38
rs899112012 Chr.10: 117488786-117488786 on Build GRCh38
rs889352536 Chr.10: 117488732-117488732 on Build GRCh38
rs959910229 Chr.10: 117488574-117488574 on Build GRCh38

View Full Product Details