Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 2134034-2134542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGF2
Entrez Gene ID
3481
Gene Name
insulin like growth factor 2
Gene Aliases
C11orf43, GRDF, IGF-II, PP9974
UniGene
Hs.272259
Gene Chromosome Location
Chr.11: 2129112-2149603 on Build GRCh38
Transcripts
NM_000612.5 NM_001007139.5 NM_001127598.2 NM_001291861.2 NM_001291862.2 NR_003512.3 NR_030158.1

Gene Symbol
INS-IGF2
Entrez Gene ID
723961
Gene Name
INS-IGF2 readthrough
Gene Aliases
INSIGF
UniGene
Hs.272259
Gene Chromosome Location
Chr.11: 2129112-2161209 on Build GRCh38
Transcripts
NM_000612.5 NM_001007139.5 NM_001127598.2 NM_001291861.2 NM_001291862.2 NR_003512.3 NR_030158.1

Gene Symbol
MIR483
Entrez Gene ID
619552
Gene Name
microRNA 483
Gene Aliases
MIRN483, hsa-mir-483, mir-483
UniGene
Gene Chromosome Location
Chr.11: 2134134-2134209 on Build GRCh38
Transcripts
NM_000612.5 NM_001007139.5 NM_001127598.2 NM_001291861.2 NM_001291862.2 NR_003512.3 NR_030158.1

SNP Detail

SNP ID Chromosome Location
rs951556830 Chr.11: 2134388-2134388 on Build GRCh38
rs991446702 Chr.11: 2134469-2134469 on Build GRCh38
rs558885880 Chr.11: 2134179-2134179 on Build GRCh38
rs911465882 Chr.11: 2134054-2134054 on Build GRCh38
rs1018580420 Chr.11: 2134333-2134333 on Build GRCh38
rs1008432521 Chr.11: 2134268-2134274 on Build GRCh38
rs1027084150 Chr.11: 2134296-2134296 on Build GRCh38
rs1041825767 Chr.11: 2134512-2134512 on Build GRCh38
rs1007215988 Chr.11: 2134318-2134318 on Build GRCh38
rs983285265 Chr.11: 2134396-2134396 on Build GRCh38
rs1030246534 Chr.11: 2134386-2134386 on Build GRCh38
rs187500191 Chr.11: 2134315-2134315 on Build GRCh38
rs950014277 Chr.11: 2134480-2134480 on Build GRCh38
rs557358105 Chr.11: 2134371-2134371 on Build GRCh38
rs796271646 Chr.11: 2134292-2134292 on Build GRCh38
rs775156935 Chr.11: 2134351-2134351 on Build GRCh38
rs757065698 Chr.11: 2134107-2134107 on Build GRCh38
rs541936322 Chr.11: 2134317-2134317 on Build GRCh38
rs763106708 Chr.11: 2134096-2134096 on Build GRCh38
rs748692358 Chr.11: 2134169-2134169 on Build GRCh38
rs964639957 Chr.11: 2134368-2134368 on Build GRCh38
rs1010741739 Chr.11: 2134207-2134207 on Build GRCh38
rs1054425167 Chr.11: 2134392-2134392 on Build GRCh38
rs914566930 Chr.11: 2134452-2134452 on Build GRCh38
rs747575116 Chr.11: 2134237-2134237 on Build GRCh38
rs769944284 Chr.11: 2134246-2134246 on Build GRCh38
rs775784989 Chr.11: 2134252-2134252 on Build GRCh38
rs894379067 Chr.11: 2134250-2134250 on Build GRCh38
rs915863325 Chr.11: 2134479-2134479 on Build GRCh38
rs776669618 Chr.11: 2134084-2134084 on Build GRCh38
rs902980285 Chr.11: 2134284-2134284 on Build GRCh38
rs781109245 Chr.11: 2134146-2134146 on Build GRCh38
rs371179917 Chr.11: 2134205-2134205 on Build GRCh38
rs776212373 Chr.11: 2134380-2134380 on Build GRCh38
rs763868632 Chr.11: 2134102-2134102 on Build GRCh38
rs537385465 Chr.11: 2134149-2134149 on Build GRCh38
rs367649889 Chr.11: 2134147-2134147 on Build GRCh38
rs941387920 Chr.11: 2134455-2134455 on Build GRCh38
rs909956562 Chr.11: 2134410-2134410 on Build GRCh38
rs1014127420 Chr.11: 2134268-2134268 on Build GRCh38
rs779817434 Chr.11: 2134154-2134154 on Build GRCh38
rs1045964492 Chr.11: 2134275-2134275 on Build GRCh38
rs184629388 Chr.11: 2134105-2134105 on Build GRCh38
rs11564729 Chr.11: 2134291-2134291 on Build GRCh38
rs777922227 Chr.11: 2134208-2134208 on Build GRCh38
rs375588009 Chr.11: 2134238-2134238 on Build GRCh38
rs891276800 Chr.11: 2134513-2134513 on Build GRCh38
rs980663624 Chr.11: 2134379-2134379 on Build GRCh38
rs893198544 Chr.11: 2134189-2134189 on Build GRCh38
rs115976511 Chr.11: 2134245-2134245 on Build GRCh38
rs368924870 Chr.11: 2134456-2134456 on Build GRCh38
rs964036791 Chr.11: 2134283-2134283 on Build GRCh38
rs768219457 Chr.11: 2134197-2134197 on Build GRCh38

View Full Product Details