Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 32439761-32440279 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WT1-AS
Entrez Gene ID
51352
Gene Name
WT1 antisense RNA
Gene Aliases
WIT-1, WIT1, WT1-AS1, WT1AS
UniGene
Hs.567499
Gene Chromosome Location
Chr.11: 32435518-32458769 on Build GRCh38
Transcripts
NR_120548.1

SNP Detail

SNP ID Chromosome Location
rs905440540 Chr.11: 32439892-32439892 on Build GRCh38
rs775225249 Chr.11: 32440189-32440189 on Build GRCh38
rs11600630 Chr.11: 32439942-32439942 on Build GRCh38
rs75039315 Chr.11: 32440239-32440239 on Build GRCh38
rs767039833 Chr.11: 32440112-32440112 on Build GRCh38
rs866211667 Chr.11: 32440105-32440105 on Build GRCh38
rs10835910 Chr.11: 32440063-32440063 on Build GRCh38
rs773172939 Chr.11: 32440107-32440107 on Build GRCh38
rs990401047 Chr.11: 32439891-32439891 on Build GRCh38
rs10835909 Chr.11: 32439840-32439840 on Build GRCh38
rs72893548 Chr.11: 32440129-32440129 on Build GRCh38
rs1004598982 Chr.11: 32440230-32440230 on Build GRCh38
rs780885190 Chr.11: 32440110-32440118 on Build GRCh38
rs895292057 Chr.11: 32440138-32440138 on Build GRCh38
rs758305234 Chr.11: 32440103-32440104 on Build GRCh38
rs573219108 Chr.11: 32439871-32439871 on Build GRCh38
rs749583533 Chr.11: 32440077-32440077 on Build GRCh38
rs577833472 Chr.11: 32440111-32440111 on Build GRCh38
rs765995443 Chr.11: 32440108-32440108 on Build GRCh38
rs755201541 Chr.11: 32440073-32440090 on Build GRCh38
rs71063728 Chr.11: 32440105-32440106 on Build GRCh38
rs1014538443 Chr.11: 32440233-32440233 on Build GRCh38
rs766596113 Chr.11: 32440073-32440087 on Build GRCh38
rs542326783 Chr.11: 32439945-32439945 on Build GRCh38
rs760262310 Chr.11: 32440192-32440192 on Build GRCh38
rs1014546679 Chr.11: 32439785-32439785 on Build GRCh38
rs761855211 Chr.11: 32440089-32440089 on Build GRCh38
rs544079703 Chr.11: 32440095-32440105 on Build GRCh38
rs978053007 Chr.11: 32439968-32439968 on Build GRCh38
rs752280934 Chr.11: 32440072-32440073 on Build GRCh38
rs11600653 Chr.11: 32440067-32440067 on Build GRCh38
rs542311420 Chr.11: 32440058-32440058 on Build GRCh38
rs561416904 Chr.11: 32440099-32440105 on Build GRCh38
rs1045239073 Chr.11: 32439855-32439855 on Build GRCh38
rs762345485 Chr.11: 32440049-32440050 on Build GRCh38
rs115320154 Chr.11: 32440015-32440015 on Build GRCh38
rs948218060 Chr.11: 32440185-32440185 on Build GRCh38
rs923972037 Chr.11: 32439973-32439973 on Build GRCh38
rs753846950 Chr.11: 32440255-32440255 on Build GRCh38
rs751191897 Chr.11: 32440110-32440110 on Build GRCh38
rs1009255877 Chr.11: 32440010-32440011 on Build GRCh38
rs761771050 Chr.11: 32439881-32439881 on Build GRCh38
rs139719053 Chr.11: 32440041-32440041 on Build GRCh38
rs72893546 Chr.11: 32440050-32440050 on Build GRCh38
rs765200572 Chr.11: 32440106-32440106 on Build GRCh38
rs771170958 Chr.11: 32440078-32440078 on Build GRCh38
rs763989212 Chr.11: 32439870-32439870 on Build GRCh38
rs774580733 Chr.11: 32440079-32440079 on Build GRCh38
rs939653300 Chr.11: 32439914-32439915 on Build GRCh38
rs558926734 Chr.11: 32440073-32440073 on Build GRCh38
rs758128489 Chr.11: 32440003-32440003 on Build GRCh38
rs776494995 Chr.11: 32440088-32440088 on Build GRCh38
rs972097054 Chr.11: 32439842-32439842 on Build GRCh38
rs752801389 Chr.11: 32440084-32440085 on Build GRCh38
rs180985730 Chr.11: 32439849-32439849 on Build GRCh38
rs778119365 Chr.11: 32440076-32440076 on Build GRCh38
rs965902878 Chr.11: 32439808-32439808 on Build GRCh38
rs35962451 Chr.11: 32440039-32440039 on Build GRCh38
rs746788140 Chr.11: 32440106-32440118 on Build GRCh38
rs755736041 Chr.11: 32439940-32439940 on Build GRCh38
rs772192094 Chr.11: 32440087-32440087 on Build GRCh38
rs545788660 Chr.11: 32439834-32439834 on Build GRCh38
rs562116418 Chr.11: 32439971-32439971 on Build GRCh38
rs773579019 Chr.11: 32440073-32440088 on Build GRCh38
rs920690215 Chr.11: 32439844-32439844 on Build GRCh38
rs767944020 Chr.11: 32440071-32440072 on Build GRCh38
rs754114244 Chr.11: 32440073-32440086 on Build GRCh38
rs760998169 Chr.11: 32440073-32440089 on Build GRCh38
rs11031786 Chr.11: 32440136-32440136 on Build GRCh38
rs563002248 Chr.11: 32440024-32440024 on Build GRCh38
rs887490301 Chr.11: 32440219-32440219 on Build GRCh38
rs953265645 Chr.11: 32439889-32439889 on Build GRCh38
rs559315401 Chr.11: 32440140-32440140 on Build GRCh38
rs189139584 Chr.11: 32439841-32439841 on Build GRCh38
rs35929634 Chr.11: 32440163-32440163 on Build GRCh38
rs765114831 Chr.11: 32439944-32439944 on Build GRCh38
rs777618070 Chr.11: 32440106-32440108 on Build GRCh38
rs996645264 Chr.11: 32440046-32440047 on Build GRCh38
rs759814476 Chr.11: 32439865-32439865 on Build GRCh38
rs986765034 Chr.11: 32440186-32440186 on Build GRCh38
rs745977770 Chr.11: 32440080-32440080 on Build GRCh38
rs745460557 Chr.11: 32440115-32440118 on Build GRCh38
rs528145547 Chr.11: 32440211-32440211 on Build GRCh38
rs756528044 Chr.11: 32440075-32440075 on Build GRCh38

View Full Product Details