Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 7108406-7108903 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1RL
Entrez Gene ID
51279
Gene Name
complement C1r subcomponent like
Gene Aliases
C1RL1, C1RLP, C1r-LP, CLSPa
UniGene
Hs.631730
Gene Chromosome Location
Chr.12: 7093662-7109278 on Build GRCh38
Transcripts
NM_001297640.1 NM_001297642.1 NM_001297643.1 NM_016546.3 XM_017019397.1 XM_017019398.1 XM_017019399.1 XM_017019400.1 XM_017019401.1 XM_017019402.1 XM_017019403.1 XM_017019404.1 NR_026947.1

Gene Symbol
C1RL-AS1
Entrez Gene ID
283314
Gene Name
C1RL antisense RNA 1
Gene Aliases
MATL2963
UniGene
Hs.744212
Gene Chromosome Location
Chr.12: 7108308-7121851 on Build GRCh38
Transcripts
NM_001297640.1 NM_001297642.1 NM_001297643.1 NM_016546.3 XM_017019397.1 XM_017019398.1 XM_017019399.1 XM_017019400.1 XM_017019401.1 XM_017019402.1 XM_017019403.1 XM_017019404.1 NR_026947.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1747233 c.141G>A p.E47E Chr.12: 7108410-7108410 on Build GRCh38
5483809 c.113C>A p.T38N Chr.12: 7108438-7108438 on Build GRCh38
1198523 c.85C>A p.L29I Chr.12: 7108466-7108466 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs904990850 Chr.12: 7108604-7108604 on Build GRCh38
rs775012431 Chr.12: 7108525-7108525 on Build GRCh38
rs562844835 Chr.12: 7108577-7108577 on Build GRCh38
rs1008564412 Chr.12: 7108733-7108733 on Build GRCh38
rs776366859 Chr.12: 7108793-7108793 on Build GRCh38
rs756297003 Chr.12: 7108445-7108453 on Build GRCh38
rs112130470 Chr.12: 7108621-7108621 on Build GRCh38
rs774685383 Chr.12: 7108711-7108711 on Build GRCh38
rs781446607 Chr.12: 7108427-7108427 on Build GRCh38
rs956476195 Chr.12: 7108769-7108769 on Build GRCh38
rs139354233 Chr.12: 7108464-7108464 on Build GRCh38
rs750984839 Chr.12: 7108476-7108476 on Build GRCh38
rs372793243 Chr.12: 7108487-7108487 on Build GRCh38
rs777932477 Chr.12: 7108488-7108489 on Build GRCh38
rs888785335 Chr.12: 7108617-7108617 on Build GRCh38
rs763634294 Chr.12: 7108474-7108474 on Build GRCh38
rs778874669 Chr.12: 7108521-7108521 on Build GRCh38
rs989095220 Chr.12: 7108770-7108770 on Build GRCh38
rs745696237 Chr.12: 7108524-7108524 on Build GRCh38
rs775183495 Chr.12: 7108472-7108472 on Build GRCh38
rs1036224613 Chr.12: 7108554-7108555 on Build GRCh38
rs997305459 Chr.12: 7108579-7108579 on Build GRCh38
rs1044450108 Chr.12: 7108798-7108798 on Build GRCh38
rs1034086643 Chr.12: 7108558-7108558 on Build GRCh38
rs1021103013 Chr.12: 7108701-7108701 on Build GRCh38
rs200463923 Chr.12: 7108499-7108499 on Build GRCh38
rs184096175 Chr.12: 7108741-7108741 on Build GRCh38
rs950037966 Chr.12: 7108780-7108780 on Build GRCh38
rs1009679948 Chr.12: 7108689-7108689 on Build GRCh38
rs1033028981 Chr.12: 7108633-7108633 on Build GRCh38
rs748529187 Chr.12: 7108766-7108766 on Build GRCh38
rs1032475794 Chr.12: 7108436-7108436 on Build GRCh38
rs1017317871 Chr.12: 7108737-7108737 on Build GRCh38
rs760349937 Chr.12: 7108530-7108530 on Build GRCh38
rs111395419 Chr.12: 7108490-7108490 on Build GRCh38
rs888766735 Chr.12: 7108869-7108869 on Build GRCh38
rs558755840 Chr.12: 7108771-7108771 on Build GRCh38
rs970785511 Chr.12: 7108736-7108736 on Build GRCh38
rs771659963 Chr.12: 7108469-7108469 on Build GRCh38
rs993455966 Chr.12: 7108637-7108637 on Build GRCh38
rs773619193 Chr.12: 7108431-7108431 on Build GRCh38
rs760178586 Chr.12: 7108679-7108679 on Build GRCh38
rs1037577132 Chr.12: 7108613-7108613 on Build GRCh38
rs749808942 Chr.12: 7108658-7108658 on Build GRCh38
rs773162127 Chr.12: 7108438-7108438 on Build GRCh38
rs752874709 Chr.12: 7108494-7108494 on Build GRCh38
rs763498414 Chr.12: 7108478-7108478 on Build GRCh38
rs377690456 Chr.12: 7108458-7108458 on Build GRCh38
rs749684850 Chr.12: 7108508-7108509 on Build GRCh38
rs1037924138 Chr.12: 7108825-7108825 on Build GRCh38
rs1002029498 Chr.12: 7108608-7108608 on Build GRCh38
rs199806858 Chr.12: 7108549-7108550 on Build GRCh38
rs991393094 Chr.12: 7108764-7108764 on Build GRCh38
rs774071238 Chr.12: 7108463-7108463 on Build GRCh38
rs770725490 Chr.12: 7108460-7108460 on Build GRCh38
rs745890228 Chr.12: 7108550-7108550 on Build GRCh38
rs150463201 Chr.12: 7108444-7108444 on Build GRCh38
rs763616202 Chr.12: 7108485-7108485 on Build GRCh38
rs1026373604 Chr.12: 7108641-7108641 on Build GRCh38
rs750091838 Chr.12: 7108801-7108801 on Build GRCh38
rs749356445 Chr.12: 7108510-7108510 on Build GRCh38
rs748226344 Chr.12: 7108432-7108432 on Build GRCh38
rs909108767 Chr.12: 7108767-7108767 on Build GRCh38
rs770836348 Chr.12: 7108520-7108520 on Build GRCh38
rs1053511486 Chr.12: 7108543-7108543 on Build GRCh38
rs941476706 Chr.12: 7108820-7108820 on Build GRCh38
rs781294369 Chr.12: 7108493-7108493 on Build GRCh38
rs934969841 Chr.12: 7108535-7108535 on Build GRCh38
rs752234367 Chr.12: 7108516-7108516 on Build GRCh38
rs919124912 Chr.12: 7108789-7108789 on Build GRCh38
rs979338166 Chr.12: 7108755-7108755 on Build GRCh38

View Full Product Details