Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 126869385-126869897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC440117
Entrez Gene ID
440117
Gene Name
uncharacterized LOC440117
Gene Aliases
-
UniGene
Hs.127100
Gene Chromosome Location
Chr.12: 126869494-126874690 on Build GRCh38
Transcripts
NR_033970.1

SNP Detail

SNP ID Chromosome Location
rs554393985 Chr.12: 126869728-126869728 on Build GRCh38
rs937074641 Chr.12: 126869609-126869609 on Build GRCh38
rs1039655535 Chr.12: 126869760-126869760 on Build GRCh38
rs1166230 Chr.12: 126869744-126869744 on Build GRCh38
rs181330000 Chr.12: 126869676-126869676 on Build GRCh38
rs956063597 Chr.12: 126869704-126869704 on Build GRCh38
rs1056805800 Chr.12: 126869833-126869833 on Build GRCh38
rs138545130 Chr.12: 126869797-126869797 on Build GRCh38
rs995269826 Chr.12: 126869458-126869458 on Build GRCh38
rs902537375 Chr.12: 126869700-126869700 on Build GRCh38
rs920791906 Chr.12: 126869763-126869763 on Build GRCh38
rs767397334 Chr.12: 126869778-126869778 on Build GRCh38
rs377456366 Chr.12: 126869424-126869424 on Build GRCh38
rs964803135 Chr.12: 126869484-126869484 on Build GRCh38
rs924686268 Chr.12: 126869679-126869679 on Build GRCh38
rs899296144 Chr.12: 126869447-126869447 on Build GRCh38
rs933600131 Chr.12: 126869504-126869504 on Build GRCh38
rs908140935 Chr.12: 126869839-126869839 on Build GRCh38
rs1035632735 Chr.12: 126869518-126869518 on Build GRCh38
rs895653222 Chr.12: 126869652-126869652 on Build GRCh38
rs373522587 Chr.12: 126869549-126869549 on Build GRCh38
rs747599622 Chr.12: 126869448-126869448 on Build GRCh38
rs386767551 Chr.12: 126869849-126869851 on Build GRCh38
rs771424794 Chr.12: 126869450-126869450 on Build GRCh38
rs545734288 Chr.12: 126869849-126869849 on Build GRCh38
rs968930810 Chr.12: 126869635-126869635 on Build GRCh38
rs770011598 Chr.12: 126869654-126869654 on Build GRCh38
rs983695774 Chr.12: 126869539-126869539 on Build GRCh38
rs111927524 Chr.12: 126869641-126869641 on Build GRCh38
rs886841576 Chr.12: 126869820-126869820 on Build GRCh38
rs571253134 Chr.12: 126869486-126869486 on Build GRCh38
rs966675078 Chr.12: 126869421-126869421 on Build GRCh38
rs563845354 Chr.12: 126869851-126869851 on Build GRCh38
rs905620608 Chr.12: 126869603-126869603 on Build GRCh38
rs941671677 Chr.12: 126869568-126869568 on Build GRCh38
rs370856734 Chr.12: 126869498-126869498 on Build GRCh38
rs943642877 Chr.12: 126869748-126869748 on Build GRCh38
rs1048148884 Chr.12: 126869472-126869472 on Build GRCh38
rs1010142711 Chr.12: 126869667-126869667 on Build GRCh38
rs1022798843 Chr.12: 126869611-126869611 on Build GRCh38
rs954843727 Chr.12: 126869802-126869802 on Build GRCh38
rs775745874 Chr.12: 126869720-126869720 on Build GRCh38
rs943571401 Chr.12: 126869413-126869413 on Build GRCh38
rs140063360 Chr.12: 126869680-126869680 on Build GRCh38
rs895429834 Chr.12: 126869848-126869848 on Build GRCh38
rs201082681 Chr.12: 126869727-126869728 on Build GRCh38
rs776750501 Chr.12: 126869475-126869475 on Build GRCh38
rs74411337 Chr.12: 126869578-126869578 on Build GRCh38
rs373850818 Chr.12: 126869828-126869828 on Build GRCh38
rs578240429 Chr.12: 126869841-126869841 on Build GRCh38
rs1056897922 Chr.12: 126869638-126869638 on Build GRCh38

View Full Product Details