Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 40196254-40196750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00548
Entrez Gene ID
400123
Gene Name
long intergenic non-protein coding RNA 548
Gene Aliases
-
UniGene
Hs.577960
Gene Chromosome Location
Chr.13: 40194509-40220502 on Build GRCh38
Transcripts
NR_033877.1

SNP Detail

SNP ID Chromosome Location
rs899019456 Chr.13: 40196535-40196535 on Build GRCh38
rs745842242 Chr.13: 40196290-40196290 on Build GRCh38
rs149517692 Chr.13: 40196373-40196373 on Build GRCh38
rs529387451 Chr.13: 40196724-40196724 on Build GRCh38
rs989617451 Chr.13: 40196666-40196666 on Build GRCh38
rs368933606 Chr.13: 40196567-40196567 on Build GRCh38
rs981013558 Chr.13: 40196641-40196641 on Build GRCh38
rs1023939266 Chr.13: 40196336-40196336 on Build GRCh38
rs918872868 Chr.13: 40196393-40196393 on Build GRCh38
rs76881476 Chr.13: 40196640-40196640 on Build GRCh38
rs553322296 Chr.13: 40196362-40196362 on Build GRCh38
rs1018403069 Chr.13: 40196335-40196335 on Build GRCh38
rs552537241 Chr.13: 40196721-40196721 on Build GRCh38
rs544919746 Chr.13: 40196418-40196418 on Build GRCh38
rs1038954931 Chr.13: 40196445-40196445 on Build GRCh38
rs943314253 Chr.13: 40196435-40196435 on Build GRCh38
rs958993361 Chr.13: 40196727-40196727 on Build GRCh38
rs9576956 Chr.13: 40196650-40196650 on Build GRCh38
rs936823667 Chr.13: 40196402-40196402 on Build GRCh38
rs1026048466 Chr.13: 40196287-40196287 on Build GRCh38
rs994614988 Chr.13: 40196281-40196281 on Build GRCh38
rs544273635 Chr.13: 40196549-40196549 on Build GRCh38
rs994688609 Chr.13: 40196553-40196553 on Build GRCh38
rs773372839 Chr.13: 40196702-40196702 on Build GRCh38
rs980982617 Chr.13: 40196368-40196368 on Build GRCh38
rs911908228 Chr.13: 40196725-40196725 on Build GRCh38
rs530551438 Chr.13: 40196506-40196506 on Build GRCh38
rs974732 Chr.13: 40196433-40196433 on Build GRCh38
rs1025888437 Chr.13: 40196649-40196649 on Build GRCh38
rs903138716 Chr.13: 40196610-40196610 on Build GRCh38
rs1052203998 Chr.13: 40196432-40196432 on Build GRCh38
rs949963924 Chr.13: 40196587-40196587 on Build GRCh38
rs967237634 Chr.13: 40196690-40196690 on Build GRCh38
rs1013965364 Chr.13: 40196591-40196591 on Build GRCh38
rs986300011 Chr.13: 40196311-40196311 on Build GRCh38
rs546146184 Chr.13: 40196644-40196644 on Build GRCh38
rs971389181 Chr.13: 40196339-40196339 on Build GRCh38
rs532838954 Chr.13: 40196676-40196676 on Build GRCh38
rs546139515 Chr.13: 40196386-40196386 on Build GRCh38
rs1000083748 Chr.13: 40196615-40196615 on Build GRCh38
rs888782676 Chr.13: 40196631-40196631 on Build GRCh38
rs987564043 Chr.13: 40196713-40196713 on Build GRCh38
rs895643584 Chr.13: 40196586-40196586 on Build GRCh38
rs965934643 Chr.13: 40196355-40196355 on Build GRCh38
rs1026123345 Chr.13: 40196572-40196572 on Build GRCh38
rs1032618781 Chr.13: 40196626-40196626 on Build GRCh38
rs1024426060 Chr.13: 40196605-40196605 on Build GRCh38
rs191269586 Chr.13: 40196648-40196648 on Build GRCh38
rs992916264 Chr.13: 40196310-40196310 on Build GRCh38
rs909896777 Chr.13: 40196465-40196465 on Build GRCh38
rs779966300 Chr.13: 40196431-40196431 on Build GRCh38
rs560621938 Chr.13: 40196577-40196577 on Build GRCh38
rs989584525 Chr.13: 40196410-40196410 on Build GRCh38
rs909850811 Chr.13: 40196693-40196693 on Build GRCh38

View Full Product Details