Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 24579825-24580356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PWRN1
Entrez Gene ID
791114
Gene Name
Prader-Willi region non-protein coding RNA 1
Gene Aliases
NCRNA00198
UniGene
Hs.720900
Gene Chromosome Location
Chr.15: 24558157-24587780 on Build GRCh38
Transcripts
NR_026646.1

SNP Detail

SNP ID Chromosome Location
rs925728528 Chr.15: 24579994-24579994 on Build GRCh38
rs1017667849 Chr.15: 24579909-24579909 on Build GRCh38
rs764273946 Chr.15: 24579907-24579907 on Build GRCh38
rs993127486 Chr.15: 24579932-24579932 on Build GRCh38
rs7180095 Chr.15: 24580220-24580220 on Build GRCh38
rs1031103776 Chr.15: 24580146-24580146 on Build GRCh38
rs559539205 Chr.15: 24580006-24580006 on Build GRCh38
rs892728493 Chr.15: 24579888-24579888 on Build GRCh38
rs944413217 Chr.15: 24580042-24580042 on Build GRCh38
rs930417668 Chr.15: 24580320-24580320 on Build GRCh38
rs572472869 Chr.15: 24580332-24580332 on Build GRCh38
rs912178172 Chr.15: 24580239-24580239 on Build GRCh38
rs747779055 Chr.15: 24580108-24580108 on Build GRCh38
rs74793630 Chr.15: 24579891-24579891 on Build GRCh38
rs758305104 Chr.15: 24580026-24580026 on Build GRCh38
rs111698967 Chr.15: 24579847-24579847 on Build GRCh38
rs1010683962 Chr.15: 24580164-24580164 on Build GRCh38
rs948394306 Chr.15: 24579978-24579978 on Build GRCh38
rs1052475232 Chr.15: 24580045-24580045 on Build GRCh38
rs1048584783 Chr.15: 24580326-24580326 on Build GRCh38
rs113966894 Chr.15: 24579931-24579931 on Build GRCh38
rs906543505 Chr.15: 24580134-24580134 on Build GRCh38
rs994943092 Chr.15: 24580307-24580307 on Build GRCh38
rs1004008059 Chr.15: 24580142-24580142 on Build GRCh38
rs762394258 Chr.15: 24580189-24580189 on Build GRCh38
rs971211297 Chr.15: 24579917-24579917 on Build GRCh38
rs112755532 Chr.15: 24580049-24580049 on Build GRCh38
rs557342633 Chr.15: 24580317-24580317 on Build GRCh38
rs150780460 Chr.15: 24580260-24580260 on Build GRCh38
rs568282050 Chr.15: 24580202-24580202 on Build GRCh38
rs112659785 Chr.15: 24580093-24580093 on Build GRCh38
rs953295560 Chr.15: 24580219-24580219 on Build GRCh38
rs956580552 Chr.15: 24579960-24579960 on Build GRCh38
rs768639456 Chr.15: 24580055-24580055 on Build GRCh38
rs139134323 Chr.15: 24580306-24580306 on Build GRCh38
rs539902691 Chr.15: 24580001-24580001 on Build GRCh38
rs1045049407 Chr.15: 24580130-24580130 on Build GRCh38
rs963427255 Chr.15: 24579910-24579910 on Build GRCh38
rs144885028 Chr.15: 24579906-24579906 on Build GRCh38
rs557190841 Chr.15: 24579918-24579918 on Build GRCh38
rs78971374 Chr.15: 24580149-24580149 on Build GRCh38
rs79039375 Chr.15: 24579995-24579995 on Build GRCh38
rs1051935935 Chr.15: 24580234-24580234 on Build GRCh38
rs148265197 Chr.15: 24579982-24579982 on Build GRCh38
rs972197732 Chr.15: 24580268-24580268 on Build GRCh38
rs965751271 Chr.15: 24580264-24580264 on Build GRCh38
rs79791765 Chr.15: 24579996-24579996 on Build GRCh38
rs899262387 Chr.15: 24580096-24580096 on Build GRCh38
rs994842493 Chr.15: 24580100-24580100 on Build GRCh38
rs745962907 Chr.15: 24580009-24580009 on Build GRCh38
rs912968211 Chr.15: 24580212-24580212 on Build GRCh38
rs1014073554 Chr.15: 24580119-24580119 on Build GRCh38
rs113463337 Chr.15: 24580113-24580113 on Build GRCh38
rs142309782 Chr.15: 24579892-24579892 on Build GRCh38
rs34011405 Chr.15: 24579898-24579898 on Build GRCh38
rs551205858 Chr.15: 24580018-24580018 on Build GRCh38
rs113307334 Chr.15: 24579871-24579871 on Build GRCh38
rs111588038 Chr.15: 24580031-24580031 on Build GRCh38
rs979774576 Chr.15: 24580151-24580151 on Build GRCh38
rs989781102 Chr.15: 24579970-24579970 on Build GRCh38
rs547080943 Chr.15: 24580188-24580188 on Build GRCh38
rs34394178 Chr.15: 24580304-24580304 on Build GRCh38
rs925572694 Chr.15: 24580177-24580177 on Build GRCh38
rs141266730 Chr.15: 24580109-24580109 on Build GRCh38

View Full Product Details