Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 14304229-14304757 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HS3ST3B1
Entrez Gene ID
9953
Gene Name
heparan sulfate-glucosamine 3-sulfotransferase 3B1
Gene Aliases
3-OST-3B, 3OST3B1, h3-OST-3B
UniGene
Hs.48384
Gene Chromosome Location
Chr.17: 14301050-14349404 on Build GRCh38
Transcripts
NM_006041.2 NR_130138.1 NR_026880.1

Gene Symbol
MGC12916
Entrez Gene ID
84815
Gene Name
uncharacterized protein MGC12916
Gene Aliases
-
UniGene
Hs.597458
Gene Chromosome Location
Chr.17: 14303740-14305745 on Build GRCh38
Transcripts
NM_006041.2 NR_130138.1 NR_026880.1

SNP Detail

SNP ID Chromosome Location
rs764817225 Chr.17: 14304282-14304282 on Build GRCh38
rs941797291 Chr.17: 14304548-14304548 on Build GRCh38
rs140595960 Chr.17: 14304496-14304496 on Build GRCh38
rs201501599 Chr.17: 14304413-14304414 on Build GRCh38
rs530825416 Chr.17: 14304628-14304628 on Build GRCh38
rs1046633368 Chr.17: 14304643-14304643 on Build GRCh38
rs893788705 Chr.17: 14304702-14304702 on Build GRCh38
rs1053754854 Chr.17: 14304684-14304684 on Build GRCh38
rs908873659 Chr.17: 14304531-14304531 on Build GRCh38
rs144915174 Chr.17: 14304485-14304485 on Build GRCh38
rs1032896812 Chr.17: 14304298-14304298 on Build GRCh38
rs964564583 Chr.17: 14304472-14304472 on Build GRCh38
rs1014281160 Chr.17: 14304345-14304345 on Build GRCh38
rs571669286 Chr.17: 14304337-14304337 on Build GRCh38
rs902526211 Chr.17: 14304629-14304630 on Build GRCh38
rs144546323 Chr.17: 14304718-14304718 on Build GRCh38
rs999443135 Chr.17: 14304409-14304409 on Build GRCh38
rs118162930 Chr.17: 14304493-14304493 on Build GRCh38
rs1006973286 Chr.17: 14304462-14304462 on Build GRCh38
rs1001492777 Chr.17: 14304655-14304655 on Build GRCh38
rs538320773 Chr.17: 14304266-14304266 on Build GRCh38
rs35615961 Chr.17: 14304451-14304452 on Build GRCh38
rs866955079 Chr.17: 14304363-14304363 on Build GRCh38
rs1020942730 Chr.17: 14304263-14304263 on Build GRCh38
rs540630680 Chr.17: 14304420-14304420 on Build GRCh38
rs558248756 Chr.17: 14304277-14304277 on Build GRCh38
rs190659742 Chr.17: 14304680-14304680 on Build GRCh38
rs1032396646 Chr.17: 14304414-14304414 on Build GRCh38
rs902540301 Chr.17: 14304378-14304378 on Build GRCh38
rs975733808 Chr.17: 14304447-14304447 on Build GRCh38
rs866377258 Chr.17: 14304538-14304538 on Build GRCh38
rs918952327 Chr.17: 14304480-14304480 on Build GRCh38
rs942221629 Chr.17: 14304293-14304293 on Build GRCh38
rs958397792 Chr.17: 14304416-14304416 on Build GRCh38
rs902739473 Chr.17: 14304660-14304660 on Build GRCh38
rs1007415428 Chr.17: 14304708-14304708 on Build GRCh38
rs983083603 Chr.17: 14304499-14304499 on Build GRCh38
rs758932444 Chr.17: 14304556-14304556 on Build GRCh38
rs1055738606 Chr.17: 14304314-14304314 on Build GRCh38
rs902747427 Chr.17: 14304264-14304264 on Build GRCh38
rs1047532515 Chr.17: 14304253-14304253 on Build GRCh38
rs778219311 Chr.17: 14304617-14304617 on Build GRCh38
rs999724418 Chr.17: 14304668-14304668 on Build GRCh38
rs1017879989 Chr.17: 14304471-14304471 on Build GRCh38
rs563310054 Chr.17: 14304580-14304580 on Build GRCh38
rs1017363630 Chr.17: 14304385-14304385 on Build GRCh38

View Full Product Details