Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 77881781-77882272 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ45079
Entrez Gene ID
400624
Gene Name
FLJ45079 protein
Gene Aliases
-
UniGene
Hs.514521
Gene Chromosome Location
Chr.17: 77879001-77884087 on Build GRCh38
Transcripts
NR_028337.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5955707 c.321G>A p.V107V Chr.17: 77882070-77882070 on Build GRCh38
5962654 c.121C>T p.H41Y Chr.17: 77882270-77882270 on Build GRCh38
6465214 c.277T>C p.F93L Chr.17: 77882114-77882114 on Build GRCh38
1589223 c.204G>T p.Q68H Chr.17: 77882187-77882187 on Build GRCh38
7243035 c.383T>C p.V128A Chr.17: 77882008-77882008 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs573192075 Chr.17: 77881863-77881863 on Build GRCh38
rs958800203 Chr.17: 77882042-77882042 on Build GRCh38
rs780616065 Chr.17: 77882108-77882108 on Build GRCh38
rs529196772 Chr.17: 77882070-77882070 on Build GRCh38
rs774701040 Chr.17: 77882161-77882161 on Build GRCh38
rs780705954 Chr.17: 77881899-77881899 on Build GRCh38
rs945195932 Chr.17: 77882175-77882175 on Build GRCh38
rs555566207 Chr.17: 77882157-77882157 on Build GRCh38
rs1050987816 Chr.17: 77881814-77881814 on Build GRCh38
rs969342544 Chr.17: 77881893-77881893 on Build GRCh38
rs191081523 Chr.17: 77881974-77881974 on Build GRCh38
rs1027700691 Chr.17: 77882109-77882109 on Build GRCh38
rs772612399 Chr.17: 77881953-77881953 on Build GRCh38
rs912192486 Chr.17: 77882195-77882195 on Build GRCh38
rs746213612 Chr.17: 77882172-77882172 on Build GRCh38
rs746438839 Chr.17: 77881927-77881927 on Build GRCh38
rs750976165 Chr.17: 77882160-77882161 on Build GRCh38
rs373790757 Chr.17: 77882221-77882221 on Build GRCh38
rs377180535 Chr.17: 77882021-77882021 on Build GRCh38
rs549250319 Chr.17: 77882084-77882084 on Build GRCh38
rs372748711 Chr.17: 77882009-77882009 on Build GRCh38
rs368326355 Chr.17: 77882049-77882049 on Build GRCh38
rs568785837 Chr.17: 77882205-77882205 on Build GRCh38
rs769443367 Chr.17: 77881905-77881905 on Build GRCh38
rs376255340 Chr.17: 77881889-77881889 on Build GRCh38
rs939875754 Chr.17: 77882030-77882030 on Build GRCh38
rs962479870 Chr.17: 77882068-77882068 on Build GRCh38
rs202033433 Chr.17: 77881890-77881890 on Build GRCh38
rs972625418 Chr.17: 77882249-77882249 on Build GRCh38
rs781539841 Chr.17: 77881909-77881909 on Build GRCh38
rs755752497 Chr.17: 77882120-77882120 on Build GRCh38
rs368899895 Chr.17: 77882169-77882169 on Build GRCh38
rs370781068 Chr.17: 77882186-77882186 on Build GRCh38
rs1002051754 Chr.17: 77882181-77882182 on Build GRCh38
rs958892480 Chr.17: 77881818-77881818 on Build GRCh38
rs183846941 Chr.17: 77881996-77881996 on Build GRCh38
rs374335966 Chr.17: 77882001-77882001 on Build GRCh38
rs1035708727 Chr.17: 77882150-77882150 on Build GRCh38
rs751407201 Chr.17: 77882092-77882092 on Build GRCh38
rs979116790 Chr.17: 77881997-77881997 on Build GRCh38
rs1002201824 Chr.17: 77882118-77882118 on Build GRCh38
rs773262243 Chr.17: 77882207-77882207 on Build GRCh38
rs372224234 Chr.17: 77882053-77882053 on Build GRCh38
rs888852584 Chr.17: 77881906-77881906 on Build GRCh38
rs376337488 Chr.17: 77882181-77882181 on Build GRCh38
rs375792051 Chr.17: 77882143-77882143 on Build GRCh38
rs148379576 Chr.17: 77881964-77881964 on Build GRCh38
rs761277417 Chr.17: 77881979-77881979 on Build GRCh38
rs960969468 Chr.17: 77882156-77882156 on Build GRCh38
rs898301695 Chr.17: 77882098-77882098 on Build GRCh38
rs539802004 Chr.17: 77881845-77881845 on Build GRCh38
rs555545358 Chr.17: 77881921-77881921 on Build GRCh38
rs988313057 Chr.17: 77881966-77881966 on Build GRCh38
rs75027882 Chr.17: 77882008-77882008 on Build GRCh38
rs531506565 Chr.17: 77882105-77882105 on Build GRCh38
rs747744872 Chr.17: 77881901-77881901 on Build GRCh38
rs560475019 Chr.17: 77882046-77882046 on Build GRCh38
rs374026442 Chr.17: 77881928-77881928 on Build GRCh38
rs1021086715 Chr.17: 77882176-77882176 on Build GRCh38
rs191913439 Chr.17: 77882101-77882101 on Build GRCh38
rs370678402 Chr.17: 77882211-77882211 on Build GRCh38
rs183241192 Chr.17: 77882116-77882116 on Build GRCh38
rs757399868 Chr.17: 77882093-77882094 on Build GRCh38
rs754728766 Chr.17: 77882194-77882194 on Build GRCh38
rs1015453537 Chr.17: 77881854-77881854 on Build GRCh38
rs764107873 Chr.17: 77881839-77881839 on Build GRCh38
rs762694139 Chr.17: 77882031-77882031 on Build GRCh38
rs370046209 Chr.17: 77882135-77882135 on Build GRCh38
rs984905381 Chr.17: 77881823-77881826 on Build GRCh38
rs940963550 Chr.17: 77881861-77881861 on Build GRCh38
rs369420864 Chr.17: 77882060-77882060 on Build GRCh38
rs554179423 Chr.17: 77882245-77882245 on Build GRCh38
rs763671331 Chr.17: 77882014-77882015 on Build GRCh38
rs761499131 Chr.17: 77882179-77882179 on Build GRCh38
rs188338057 Chr.17: 77882010-77882010 on Build GRCh38
rs145895629 Chr.17: 77882115-77882115 on Build GRCh38
rs553426883 Chr.17: 77881853-77881853 on Build GRCh38
rs776836502 Chr.17: 77882190-77882190 on Build GRCh38
rs1036986321 Chr.17: 77881862-77881862 on Build GRCh38
rs373509129 Chr.17: 77882178-77882178 on Build GRCh38
rs373109531 Chr.17: 77882187-77882187 on Build GRCh38
rs376105233 Chr.17: 77882191-77882191 on Build GRCh38
rs187643707 Chr.17: 77882200-77882200 on Build GRCh38
rs1037319888 Chr.17: 77882231-77882231 on Build GRCh38
rs1036883011 Chr.17: 77882091-77882091 on Build GRCh38
rs897896226 Chr.17: 77881858-77881858 on Build GRCh38
rs985813879 Chr.17: 77882118-77882119 on Build GRCh38
rs898332826 Chr.17: 77882238-77882238 on Build GRCh38

View Full Product Details