Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 116713655-116714203 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TUSC7
Entrez Gene ID
285194
Gene Name
tumor suppressor candidate 7 (non-protein coding)
Gene Aliases
LINC00902, LSAMP-AS1, LSAMP-AS3, LSAMPAS3, NCRNA00295
UniGene
Hs.120364
Gene Chromosome Location
Chr.3: 116709788-116717040 on Build GRCh38
Transcripts
NR_015391.1

SNP Detail

SNP ID Chromosome Location
rs555361891 Chr.3: 116714091-116714091 on Build GRCh38
rs1055321847 Chr.3: 116713842-116713842 on Build GRCh38
rs1051483142 Chr.3: 116714120-116714120 on Build GRCh38
rs539624412 Chr.3: 116713732-116713732 on Build GRCh38
rs374732433 Chr.3: 116713982-116713982 on Build GRCh38
rs969328385 Chr.3: 116714066-116714066 on Build GRCh38
rs207463588 Chr.3: 116714138-116714138 on Build GRCh38
rs1015632185 Chr.3: 116713753-116713753 on Build GRCh38
rs115757572 Chr.3: 116713910-116713910 on Build GRCh38
rs557704841 Chr.3: 116713736-116713736 on Build GRCh38
rs912963704 Chr.3: 116713701-116713701 on Build GRCh38
rs888430734 Chr.3: 116713749-116713749 on Build GRCh38
rs769443278 Chr.3: 116713841-116713841 on Build GRCh38
rs957276638 Chr.3: 116713924-116713924 on Build GRCh38
rs988421063 Chr.3: 116713783-116713783 on Build GRCh38
rs949829875 Chr.3: 116713704-116713704 on Build GRCh38
rs1046550464 Chr.3: 116713716-116713716 on Build GRCh38
rs772296318 Chr.3: 116713698-116713698 on Build GRCh38
rs527807646 Chr.3: 116713843-116713843 on Build GRCh38
rs541859364 Chr.3: 116713801-116713801 on Build GRCh38
rs539557220 Chr.3: 116713975-116713975 on Build GRCh38
rs941556331 Chr.3: 116714072-116714072 on Build GRCh38
rs1017874989 Chr.3: 116713797-116713797 on Build GRCh38
rs371689538 Chr.3: 116713954-116713954 on Build GRCh38
rs1007445119 Chr.3: 116713791-116713791 on Build GRCh38
rs186591257 Chr.3: 116713925-116713925 on Build GRCh38
rs574498927 Chr.3: 116713788-116713788 on Build GRCh38
rs760026288 Chr.3: 116713764-116713764 on Build GRCh38
rs1055792640 Chr.3: 116713745-116713745 on Build GRCh38
rs1007007524 Chr.3: 116714150-116714150 on Build GRCh38
rs375910831 Chr.3: 116713912-116713912 on Build GRCh38
rs991389088 Chr.3: 116714042-116714042 on Build GRCh38
rs910092073 Chr.3: 116714054-116714054 on Build GRCh38
rs1030176189 Chr.3: 116713909-116713909 on Build GRCh38
rs71616365 Chr.3: 116714153-116714154 on Build GRCh38
rs894618282 Chr.3: 116713742-116713742 on Build GRCh38
rs963214970 Chr.3: 116713817-116713817 on Build GRCh38
rs66505970 Chr.3: 116713890-116713890 on Build GRCh38
rs191389680 Chr.3: 116713963-116713963 on Build GRCh38
rs34251226 Chr.3: 116714113-116714113 on Build GRCh38
rs971044669 Chr.3: 116713953-116713953 on Build GRCh38
rs557909535 Chr.3: 116713977-116713977 on Build GRCh38
rs924736309 Chr.3: 116714112-116714112 on Build GRCh38
rs7638310 Chr.3: 116713752-116713752 on Build GRCh38
rs546361352 Chr.3: 116713876-116713876 on Build GRCh38
rs557171447 Chr.3: 116714152-116714153 on Build GRCh38
rs974591850 Chr.3: 116713822-116713822 on Build GRCh38
rs1035665313 Chr.3: 116713996-116713996 on Build GRCh38
rs368419811 Chr.3: 116713891-116713891 on Build GRCh38
rs66550426 Chr.3: 116713694-116713694 on Build GRCh38
rs992172047 Chr.3: 116714012-116714012 on Build GRCh38
rs533457064 Chr.3: 116713703-116713703 on Build GRCh38
rs376990863 Chr.3: 116713848-116713848 on Build GRCh38
rs916091618 Chr.3: 116713889-116713889 on Build GRCh38
rs916164911 Chr.3: 116714045-116714045 on Build GRCh38
rs942998856 Chr.3: 116714126-116714126 on Build GRCh38
rs998248581 Chr.3: 116713893-116713893 on Build GRCh38
rs978962490 Chr.3: 116714069-116714069 on Build GRCh38
rs9878792 Chr.3: 116713824-116713824 on Build GRCh38
rs906732322 Chr.3: 116713724-116713724 on Build GRCh38
rs1037297091 Chr.3: 116714107-116714107 on Build GRCh38
rs1039397296 Chr.3: 116714160-116714160 on Build GRCh38
rs998312412 Chr.3: 116713758-116713758 on Build GRCh38
rs947135672 Chr.3: 116713743-116713743 on Build GRCh38
rs925878217 Chr.3: 116713773-116713773 on Build GRCh38
rs143151540 Chr.3: 116714067-116714067 on Build GRCh38
rs1051123825 Chr.3: 116714130-116714130 on Build GRCh38
rs556472136 Chr.3: 116713780-116713780 on Build GRCh38
rs949800606 Chr.3: 116713823-116713823 on Build GRCh38
rs750399261 Chr.3: 116713989-116713989 on Build GRCh38
rs928119201 Chr.3: 116713827-116713827 on Build GRCh38
rs541797939 Chr.3: 116714111-116714111 on Build GRCh38
rs1026532354 Chr.3: 116713972-116713976 on Build GRCh38
rs397754669 Chr.3: 116714162-116714163 on Build GRCh38
rs927017146 Chr.3: 116713985-116713985 on Build GRCh38
rs565501230 Chr.3: 116713763-116713763 on Build GRCh38
rs960016099 Chr.3: 116714003-116714003 on Build GRCh38
rs762547441 Chr.3: 116713833-116713833 on Build GRCh38
rs773727659 Chr.3: 116714148-116714148 on Build GRCh38
rs767131336 Chr.3: 116713951-116713951 on Build GRCh38
rs7615877 Chr.3: 116713683-116713683 on Build GRCh38
rs961446746 Chr.3: 116713755-116713755 on Build GRCh38
rs928882385 Chr.3: 116713715-116713715 on Build GRCh38
rs552268174 Chr.3: 116713874-116713874 on Build GRCh38
rs148214536 Chr.3: 116714093-116714093 on Build GRCh38
rs569756456 Chr.3: 116713986-116713986 on Build GRCh38
rs983033723 Chr.3: 116713707-116713707 on Build GRCh38
rs35256076 Chr.3: 116714122-116714122 on Build GRCh38
rs770161337 Chr.3: 116713766-116713766 on Build GRCh38
rs960297400 Chr.3: 116713864-116713864 on Build GRCh38
rs528893635 Chr.3: 116713921-116713921 on Build GRCh38
rs568811479 Chr.3: 116713915-116713915 on Build GRCh38
rs546002948 Chr.3: 116714135-116714135 on Build GRCh38
rs1005841221 Chr.3: 116713884-116713884 on Build GRCh38

View Full Product Details