Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 160404644-160405160 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR15B
Entrez Gene ID
406949
Gene Name
microRNA 15b
Gene Aliases
MIRN15B, hsa-mir-15b, miR-15b
UniGene
Gene Chromosome Location
Chr.3: 160404588-160404685 on Build GRCh38
Transcripts
NR_029663.1 NR_029525.1 NM_001002800.2 NM_001288753.1 NM_005496.3 XM_006713459.3 XM_011512311.2 XM_011512312.2 XM_017005491.1

Gene Symbol
MIR16-2
Entrez Gene ID
406951
Gene Name
microRNA 16-2
Gene Aliases
MIRN16-2, mir-16-2, mir-16-3
UniGene
Gene Chromosome Location
Chr.3: 160404745-160404825 on Build GRCh38
Transcripts
NR_029663.1 NR_029525.1 NM_001002800.2 NM_001288753.1 NM_005496.3 XM_006713459.3 XM_011512311.2 XM_011512312.2 XM_017005491.1

Gene Symbol
SMC4
Entrez Gene ID
10051
Gene Name
structural maintenance of chromosomes 4
Gene Aliases
CAP-C, CAPC, SMC-4, SMC4L1
UniGene
Hs.58992
Gene Chromosome Location
Chr.3: 160399304-160434962 on Build GRCh38
Transcripts
NR_029663.1 NR_029525.1 NM_001002800.2 NM_001288753.1 NM_005496.3 XM_006713459.3 XM_011512311.2 XM_011512312.2 XM_017005491.1

SNP Detail

SNP ID Chromosome Location
rs986248879 Chr.3: 160404729-160404729 on Build GRCh38
rs749488063 Chr.3: 160404784-160404784 on Build GRCh38
rs775368833 Chr.3: 160404800-160404800 on Build GRCh38
rs773474590 Chr.3: 160404675-160404675 on Build GRCh38
rs370054586 Chr.3: 160404716-160404716 on Build GRCh38
rs556548670 Chr.3: 160405030-160405030 on Build GRCh38
rs377188987 Chr.3: 160404707-160404707 on Build GRCh38
rs758719464 Chr.3: 160404708-160404708 on Build GRCh38
rs139924760 Chr.3: 160404854-160404854 on Build GRCh38
rs1042840744 Chr.3: 160405079-160405079 on Build GRCh38
rs112178044 Chr.3: 160404850-160404850 on Build GRCh38
rs963428940 Chr.3: 160404880-160404880 on Build GRCh38
rs530751911 Chr.3: 160404706-160404706 on Build GRCh38
rs577400561 Chr.3: 160405128-160405128 on Build GRCh38
rs776686572 Chr.3: 160404829-160404829 on Build GRCh38
rs759302076 Chr.3: 160404676-160404676 on Build GRCh38
rs143326955 Chr.3: 160404910-160404910 on Build GRCh38
rs771871417 Chr.3: 160404680-160404680 on Build GRCh38
rs1011316784 Chr.3: 160405044-160405044 on Build GRCh38
rs1041315444 Chr.3: 160404893-160404893 on Build GRCh38
rs537392474 Chr.3: 160405087-160405087 on Build GRCh38
rs781659399 Chr.3: 160404750-160404750 on Build GRCh38
rs954641752 Chr.3: 160405116-160405116 on Build GRCh38
rs749204218 Chr.3: 160404786-160404786 on Build GRCh38
rs746992239 Chr.3: 160404798-160404799 on Build GRCh38
rs184323459 Chr.3: 160404739-160404739 on Build GRCh38
rs1027837936 Chr.3: 160405114-160405114 on Build GRCh38
rs772759024 Chr.3: 160404852-160404852 on Build GRCh38
rs372801908 Chr.3: 160405053-160405053 on Build GRCh38
rs891477292 Chr.3: 160405038-160405038 on Build GRCh38
rs1047064341 Chr.3: 160404949-160404949 on Build GRCh38
rs935613087 Chr.3: 160405006-160405006 on Build GRCh38
rs900313040 Chr.3: 160405086-160405086 on Build GRCh38
rs768780684 Chr.3: 160404809-160404809 on Build GRCh38
rs748289135 Chr.3: 160404763-160404763 on Build GRCh38
rs962171714 Chr.3: 160405090-160405090 on Build GRCh38
rs755407815 Chr.3: 160404727-160404727 on Build GRCh38
rs756199981 Chr.3: 160404774-160404774 on Build GRCh38
rs928243547 Chr.3: 160404956-160404956 on Build GRCh38
rs1044827216 Chr.3: 160405052-160405052 on Build GRCh38
rs151181818 Chr.3: 160405035-160405035 on Build GRCh38
rs996065368 Chr.3: 160404734-160404734 on Build GRCh38
rs535132249 Chr.3: 160404916-160404916 on Build GRCh38
rs369598613 Chr.3: 160404670-160404670 on Build GRCh38
rs372765795 Chr.3: 160404696-160404696 on Build GRCh38
rs761268125 Chr.3: 160404830-160404830 on Build GRCh38
rs1003289383 Chr.3: 160405065-160405067 on Build GRCh38
rs753332612 Chr.3: 160404705-160404705 on Build GRCh38
rs996013047 Chr.3: 160405092-160405092 on Build GRCh38
rs778638535 Chr.3: 160405091-160405091 on Build GRCh38
rs766731886 Chr.3: 160404857-160404857 on Build GRCh38
rs766678915 Chr.3: 160404715-160404715 on Build GRCh38
rs771786973 Chr.3: 160404785-160404785 on Build GRCh38
rs373202341 Chr.3: 160404806-160404806 on Build GRCh38
rs1021448761 Chr.3: 160404722-160404722 on Build GRCh38
rs751984870 Chr.3: 160404873-160404873 on Build GRCh38
rs948239453 Chr.3: 160404928-160404928 on Build GRCh38
rs760642448 Chr.3: 160404692-160404692 on Build GRCh38
rs559184480 Chr.3: 160405115-160405115 on Build GRCh38
rs775601972 Chr.3: 160404691-160404691 on Build GRCh38
rs777900452 Chr.3: 160404775-160404775 on Build GRCh38
rs964568694 Chr.3: 160404730-160404730 on Build GRCh38

View Full Product Details