Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 9902451-9902958 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00599
Entrez Gene ID
157627
Gene Name
long intergenic non-protein coding RNA 599
Gene Aliases
Rncr3
UniGene
Hs.12513
Gene Chromosome Location
Chr.8: 9900064-9903329 on Build GRCh38
Transcripts
NR_024281.1

SNP Detail

SNP ID Chromosome Location
rs898267037 Chr.8: 9902551-9902551 on Build GRCh38
rs780410943 Chr.8: 9902925-9902925 on Build GRCh38
rs900686357 Chr.8: 9902651-9902651 on Build GRCh38
rs1047715763 Chr.8: 9902623-9902623 on Build GRCh38
rs1017772404 Chr.8: 9902479-9902479 on Build GRCh38
rs908131646 Chr.8: 9902721-9902721 on Build GRCh38
rs150172541 Chr.8: 9902549-9902549 on Build GRCh38
rs532160427 Chr.8: 9902922-9902922 on Build GRCh38
rs561397671 Chr.8: 9902482-9902482 on Build GRCh38
rs995280054 Chr.8: 9902557-9902557 on Build GRCh38
rs960168954 Chr.8: 9902909-9902909 on Build GRCh38
rs138685748 Chr.8: 9902675-9902675 on Build GRCh38
rs998074200 Chr.8: 9902667-9902667 on Build GRCh38
rs73530415 Chr.8: 9902597-9902597 on Build GRCh38
rs546341166 Chr.8: 9902489-9902489 on Build GRCh38
rs1034439575 Chr.8: 9902892-9902892 on Build GRCh38
rs561128366 Chr.8: 9902731-9902731 on Build GRCh38
rs1022077770 Chr.8: 9902864-9902864 on Build GRCh38
rs992275493 Chr.8: 9902604-9902604 on Build GRCh38
rs950885637 Chr.8: 9902793-9902793 on Build GRCh38
rs188274998 Chr.8: 9902508-9902508 on Build GRCh38
rs559186691 Chr.8: 9902652-9902652 on Build GRCh38
rs575861344 Chr.8: 9902653-9902653 on Build GRCh38
rs532222555 Chr.8: 9902520-9902520 on Build GRCh38
rs891730328 Chr.8: 9902857-9902857 on Build GRCh38
rs1042273189 Chr.8: 9902929-9902929 on Build GRCh38
rs116156600 Chr.8: 9902636-9902636 on Build GRCh38
rs592420 Chr.8: 9902910-9902910 on Build GRCh38
rs372813889 Chr.8: 9902606-9902606 on Build GRCh38
rs959261188 Chr.8: 9902743-9902743 on Build GRCh38
rs541434540 Chr.8: 9902483-9902483 on Build GRCh38
rs770081968 Chr.8: 9902915-9902915 on Build GRCh38
rs931097215 Chr.8: 9902789-9902789 on Build GRCh38
rs181834869 Chr.8: 9902523-9902523 on Build GRCh38
rs568102710 Chr.8: 9902558-9902558 on Build GRCh38
rs942312235 Chr.8: 9902815-9902815 on Build GRCh38
rs960200059 Chr.8: 9902703-9902703 on Build GRCh38
rs992390737 Chr.8: 9902763-9902763 on Build GRCh38
rs1049576618 Chr.8: 9902585-9902585 on Build GRCh38
rs968819712 Chr.8: 9902650-9902650 on Build GRCh38
rs537815143 Chr.8: 9902540-9902540 on Build GRCh38
rs569093350 Chr.8: 9902527-9902527 on Build GRCh38
rs889577381 Chr.8: 9902798-9902798 on Build GRCh38
rs1052283798 Chr.8: 9902919-9902919 on Build GRCh38
rs927487212 Chr.8: 9902684-9902684 on Build GRCh38
rs909457287 Chr.8: 9902807-9902807 on Build GRCh38
rs1040944074 Chr.8: 9902835-9902835 on Build GRCh38
rs1033993166 Chr.8: 9902732-9902732 on Build GRCh38
rs1001578214 Chr.8: 9902883-9902883 on Build GRCh38
rs7465669 Chr.8: 9902699-9902699 on Build GRCh38
rs370127580 Chr.8: 9902845-9902845 on Build GRCh38
rs979305112 Chr.8: 9902566-9902566 on Build GRCh38
rs1009276890 Chr.8: 9902704-9902704 on Build GRCh38
rs781755494 Chr.8: 9902792-9902792 on Build GRCh38
rs1049546891 Chr.8: 9902797-9902797 on Build GRCh38
rs574836006 Chr.8: 9902784-9902784 on Build GRCh38
rs1010223901 Chr.8: 9902860-9902860 on Build GRCh38
rs926485753 Chr.8: 9902580-9902580 on Build GRCh38
rs938951878 Chr.8: 9902518-9902518 on Build GRCh38
rs993062872 Chr.8: 9902708-9902708 on Build GRCh38
rs757888942 Chr.8: 9902517-9902517 on Build GRCh38
rs1019952634 Chr.8: 9902613-9902613 on Build GRCh38
rs573468920 Chr.8: 9902630-9902630 on Build GRCh38
rs921941564 Chr.8: 9902874-9902874 on Build GRCh38
rs904603361 Chr.8: 9902871-9902871 on Build GRCh38
rs1037127063 Chr.8: 9902750-9902750 on Build GRCh38
rs939688042 Chr.8: 9902546-9902546 on Build GRCh38
rs927519031 Chr.8: 9902500-9902500 on Build GRCh38
rs540320371 Chr.8: 9902834-9902834 on Build GRCh38
rs186131714 Chr.8: 9902624-9902624 on Build GRCh38
rs956037199 Chr.8: 9902616-9902616 on Build GRCh38
rs919662769 Chr.8: 9902772-9902772 on Build GRCh38

View Full Product Details