Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35860349-35860846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00950
Entrez Gene ID
92973
Gene Name
long intergenic non-protein coding RNA 950
Gene Aliases
FP588
UniGene
Hs.12509
Gene Chromosome Location
Chr.9: 35860274-35865518 on Build GRCh38
Transcripts
NR_024006.1 XM_011517916.2

Gene Symbol
TMEM8B
Entrez Gene ID
51754
Gene Name
transmembrane protein 8B
Gene Aliases
C9orf127, NAG-5, NAG5, NGX6
UniGene
Hs.493808
Gene Chromosome Location
Chr.9: 35825817-35865515 on Build GRCh38
Transcripts
NR_024006.1 XM_011517916.2

SNP Detail

SNP ID Chromosome Location
rs199549443 Chr.9: 35860658-35860658 on Build GRCh38
rs200693877 Chr.9: 35860659-35860659 on Build GRCh38
rs919067663 Chr.9: 35860544-35860544 on Build GRCh38
rs1056759559 Chr.9: 35860542-35860542 on Build GRCh38
rs909121902 Chr.9: 35860569-35860569 on Build GRCh38
rs528826314 Chr.9: 35860444-35860444 on Build GRCh38
rs552981854 Chr.9: 35860579-35860579 on Build GRCh38
rs1017123599 Chr.9: 35860701-35860701 on Build GRCh38
rs1045062120 Chr.9: 35860590-35860590 on Build GRCh38
rs1035124913 Chr.9: 35860379-35860379 on Build GRCh38
rs933629008 Chr.9: 35860413-35860413 on Build GRCh38
rs1005758081 Chr.9: 35860614-35860614 on Build GRCh38
rs186902023 Chr.9: 35860555-35860555 on Build GRCh38
rs766318680 Chr.9: 35860409-35860409 on Build GRCh38
rs189546934 Chr.9: 35860494-35860494 on Build GRCh38
rs913685581 Chr.9: 35860824-35860824 on Build GRCh38
rs66822942 Chr.9: 35860654-35860657 on Build GRCh38
rs776538256 Chr.9: 35860428-35860428 on Build GRCh38
rs1009684773 Chr.9: 35860707-35860707 on Build GRCh38
rs1047202297 Chr.9: 35860578-35860578 on Build GRCh38
rs1043530780 Chr.9: 35860521-35860521 on Build GRCh38
rs955098431 Chr.9: 35860779-35860779 on Build GRCh38
rs758018360 Chr.9: 35860796-35860796 on Build GRCh38
rs367551110 Chr.9: 35860570-35860570 on Build GRCh38
rs904086902 Chr.9: 35860782-35860782 on Build GRCh38
rs1030300527 Chr.9: 35860777-35860777 on Build GRCh38
rs10814287 Chr.9: 35860486-35860486 on Build GRCh38
rs149024263 Chr.9: 35860702-35860702 on Build GRCh38
rs910317142 Chr.9: 35860469-35860469 on Build GRCh38
rs538659470 Chr.9: 35860658-35860661 on Build GRCh38
rs968153734 Chr.9: 35860442-35860442 on Build GRCh38
rs752487783 Chr.9: 35860749-35860749 on Build GRCh38
rs144952601 Chr.9: 35860448-35860448 on Build GRCh38
rs545358087 Chr.9: 35860689-35860689 on Build GRCh38
rs904998370 Chr.9: 35860528-35860528 on Build GRCh38
rs895827989 Chr.9: 35860549-35860549 on Build GRCh38
rs905291546 Chr.9: 35860602-35860602 on Build GRCh38
rs1021970129 Chr.9: 35860401-35860401 on Build GRCh38
rs573047140 Chr.9: 35860642-35860642 on Build GRCh38
rs544435985 Chr.9: 35860812-35860812 on Build GRCh38
rs566194671 Chr.9: 35860381-35860381 on Build GRCh38
rs199844812 Chr.9: 35860654-35860654 on Build GRCh38
rs182205424 Chr.9: 35860530-35860530 on Build GRCh38
rs1051783867 Chr.9: 35860481-35860481 on Build GRCh38
rs575483922 Chr.9: 35860786-35860786 on Build GRCh38
rs997127800 Chr.9: 35860723-35860723 on Build GRCh38
rs745657481 Chr.9: 35860397-35860400 on Build GRCh38
rs1023009598 Chr.9: 35860736-35860736 on Build GRCh38

View Full Product Details