Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136727889-136728386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNHG7
Entrez Gene ID
84973
Gene Name
small nucleolar RNA host gene 7
Gene Aliases
NCRNA00061
UniGene
Hs.495480
Gene Chromosome Location
Chr.9: 136724594-136728184 on Build GRCh38
Transcripts
NR_003672.2 NR_024542.1 NR_024543.1

SNP Detail

SNP ID Chromosome Location
rs1029656976 Chr.9: 136728142-136728142 on Build GRCh38
rs1007716715 Chr.9: 136728246-136728246 on Build GRCh38
rs931249226 Chr.9: 136728213-136728213 on Build GRCh38
rs992610649 Chr.9: 136728342-136728342 on Build GRCh38
rs992275127 Chr.9: 136728190-136728190 on Build GRCh38
rs889225746 Chr.9: 136728231-136728231 on Build GRCh38
rs968249723 Chr.9: 136728145-136728145 on Build GRCh38
rs1001476974 Chr.9: 136728116-136728116 on Build GRCh38
rs11545433 Chr.9: 136728147-136728147 on Build GRCh38
rs573010983 Chr.9: 136728287-136728287 on Build GRCh38
rs984232991 Chr.9: 136728294-136728294 on Build GRCh38
rs565019127 Chr.9: 136728097-136728097 on Build GRCh38
rs1046964174 Chr.9: 136728218-136728218 on Build GRCh38
rs376993900 Chr.9: 136727994-136727995 on Build GRCh38
rs2275158 Chr.9: 136727998-136727998 on Build GRCh38
rs58529385 Chr.9: 136727997-136727998 on Build GRCh38
rs542706466 Chr.9: 136727951-136727951 on Build GRCh38
rs369479948 Chr.9: 136728113-136728113 on Build GRCh38
rs867450795 Chr.9: 136728353-136728353 on Build GRCh38
rs1000640987 Chr.9: 136728204-136728204 on Build GRCh38
rs754039507 Chr.9: 136728131-136728131 on Build GRCh38
rs981145146 Chr.9: 136728195-136728195 on Build GRCh38
rs888329832 Chr.9: 136728115-136728115 on Build GRCh38
rs909658006 Chr.9: 136728180-136728180 on Build GRCh38
rs530643021 Chr.9: 136728100-136728101 on Build GRCh38
rs143034860 Chr.9: 136728350-136728350 on Build GRCh38
rs1043089696 Chr.9: 136728279-136728279 on Build GRCh38
rs980863703 Chr.9: 136728323-136728323 on Build GRCh38
rs9411221 Chr.9: 136728194-136728194 on Build GRCh38
rs565062816 Chr.9: 136728327-136728327 on Build GRCh38
rs1025806109 Chr.9: 136727923-136727923 on Build GRCh38
rs561302263 Chr.9: 136728135-136728135 on Build GRCh38
rs1034324318 Chr.9: 136727952-136727952 on Build GRCh38
rs896948863 Chr.9: 136728078-136728078 on Build GRCh38
rs536301241 Chr.9: 136728202-136728202 on Build GRCh38
rs943736845 Chr.9: 136728067-136728067 on Build GRCh38
rs1007869127 Chr.9: 136728128-136728128 on Build GRCh38
rs1019135439 Chr.9: 136728250-136728250 on Build GRCh38
rs193241184 Chr.9: 136728130-136728130 on Build GRCh38
rs538831931 Chr.9: 136728152-136728152 on Build GRCh38
rs963670471 Chr.9: 136728133-136728133 on Build GRCh38
rs2105078 Chr.9: 136727950-136727950 on Build GRCh38
rs150224514 Chr.9: 136728155-136728155 on Build GRCh38
rs7875525 Chr.9: 136727978-136727978 on Build GRCh38
rs1024845505 Chr.9: 136728013-136728014 on Build GRCh38
rs996430003 Chr.9: 136728177-136728177 on Build GRCh38
rs538494512 Chr.9: 136728179-136728179 on Build GRCh38
rs996540615 Chr.9: 136728264-136728264 on Build GRCh38
rs1031610144 Chr.9: 136728187-136728187 on Build GRCh38
rs4880137 Chr.9: 136727975-136727975 on Build GRCh38
rs993751459 Chr.9: 136728267-136728267 on Build GRCh38
rs1047314508 Chr.9: 136728158-136728158 on Build GRCh38
rs956751378 Chr.9: 136728148-136728148 on Build GRCh38
rs969439235 Chr.9: 136728313-136728313 on Build GRCh38
rs918001408 Chr.9: 136728193-136728193 on Build GRCh38
rs2275157 Chr.9: 136728062-136728062 on Build GRCh38
rs995292738 Chr.9: 136727918-136727918 on Build GRCh38
rs947975614 Chr.9: 136728088-136728088 on Build GRCh38
rs1045026702 Chr.9: 136728154-136728154 on Build GRCh38
rs951117498 Chr.9: 136727944-136727944 on Build GRCh38
rs111356893 Chr.9: 136727982-136727982 on Build GRCh38
rs950435687 Chr.9: 136728030-136728030 on Build GRCh38
rs571211242 Chr.9: 136728024-136728034 on Build GRCh38
rs1007964038 Chr.9: 136728166-136728166 on Build GRCh38
rs371819328 Chr.9: 136728149-136728149 on Build GRCh38
rs545267421 Chr.9: 136728021-136728021 on Build GRCh38
rs530038841 Chr.9: 136728146-136728146 on Build GRCh38
rs185732925 Chr.9: 136728169-136728169 on Build GRCh38
rs983145167 Chr.9: 136728328-136728328 on Build GRCh38
rs974949944 Chr.9: 136727967-136727967 on Build GRCh38
rs1013720656 Chr.9: 136728303-136728303 on Build GRCh38
rs530862580 Chr.9: 136728127-136728127 on Build GRCh38
rs772202055 Chr.9: 136728100-136728100 on Build GRCh38
rs899548442 Chr.9: 136728173-136728173 on Build GRCh38
rs113982899 Chr.9: 136728296-136728296 on Build GRCh38
rs1056449073 Chr.9: 136728049-136728049 on Build GRCh38
rs908419485 Chr.9: 136728219-136728219 on Build GRCh38
rs935326235 Chr.9: 136728024-136728024 on Build GRCh38
rs978977106 Chr.9: 136727984-136727984 on Build GRCh38
rs1016322493 Chr.9: 136728153-136728153 on Build GRCh38
rs1007773488 Chr.9: 136728162-136728162 on Build GRCh38
rs953719728 Chr.9: 136728143-136728143 on Build GRCh38
rs139012469 Chr.9: 136728157-136728157 on Build GRCh38
rs959985529 Chr.9: 136728330-136728330 on Build GRCh38
rs4880066 Chr.9: 136727972-136727972 on Build GRCh38
rs943871777 Chr.9: 136728226-136728226 on Build GRCh38
rs943883198 Chr.9: 136728355-136728355 on Build GRCh38
rs899558521 Chr.9: 136728245-136728245 on Build GRCh38
rs960089195 Chr.9: 136727953-136727953 on Build GRCh38
rs1031242752 Chr.9: 136728298-136728298 on Build GRCh38
rs896422781 Chr.9: 136728072-136728072 on Build GRCh38
rs1027431720 Chr.9: 136728326-136728326 on Build GRCh38
rs1055630505 Chr.9: 136728150-136728150 on Build GRCh38
rs937187324 Chr.9: 136728184-136728184 on Build GRCh38
rs994051967 Chr.9: 136727922-136727922 on Build GRCh38
rs554971361 Chr.9: 136728185-136728185 on Build GRCh38
rs966258534 Chr.9: 136728262-136728262 on Build GRCh38
rs898302285 Chr.9: 136727912-136727912 on Build GRCh38
rs1025393278 Chr.9: 136728120-136728120 on Build GRCh38
rs1040775207 Chr.9: 136728237-136728237 on Build GRCh38
rs992874164 Chr.9: 136727962-136727962 on Build GRCh38
rs1025070216 Chr.9: 136728308-136728308 on Build GRCh38
rs553097643 Chr.9: 136728281-136728281 on Build GRCh38
rs892675430 Chr.9: 136728300-136728300 on Build GRCh38
rs956844623 Chr.9: 136728188-136728188 on Build GRCh38
rs968674599 Chr.9: 136728182-136728182 on Build GRCh38
rs771437209 Chr.9: 136727975-136727976 on Build GRCh38
rs748236986 Chr.9: 136728093-136728093 on Build GRCh38
rs966246034 Chr.9: 136727957-136727957 on Build GRCh38
rs1040811346 Chr.9: 136728071-136728071 on Build GRCh38
rs886926403 Chr.9: 136728161-136728161 on Build GRCh38

View Full Product Details