Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35480095-35480644 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP8B5P
Entrez Gene ID
158381
Gene Name
ATPase phospholipid transporting 8B5, pseudogene
Gene Aliases
FetA
UniGene
Hs.575661
Gene Chromosome Location
Chr.9: 35406755-35483029 on Build GRCh38
Transcripts
NR_003581.2

SNP Detail

SNP ID Chromosome Location
rs765131282 Chr.9: 35480164-35480164 on Build GRCh38
rs138578397 Chr.9: 35480509-35480509 on Build GRCh38
rs1051606137 Chr.9: 35480463-35480463 on Build GRCh38
rs758259155 Chr.9: 35480183-35480183 on Build GRCh38
rs780525194 Chr.9: 35480464-35480464 on Build GRCh38
rs756811885 Chr.9: 35480333-35480333 on Build GRCh38
rs77887744 Chr.9: 35480176-35480176 on Build GRCh38
rs989215618 Chr.9: 35480282-35480282 on Build GRCh38
rs879676911 Chr.9: 35480530-35480530 on Build GRCh38
rs544029578 Chr.9: 35480583-35480583 on Build GRCh38
rs574348163 Chr.9: 35480535-35480535 on Build GRCh38
rs541395963 Chr.9: 35480405-35480405 on Build GRCh38
rs57100331 Chr.9: 35480304-35480304 on Build GRCh38
rs7040391 Chr.9: 35480567-35480567 on Build GRCh38
rs983129426 Chr.9: 35480366-35480366 on Build GRCh38
rs530701437 Chr.9: 35480208-35480208 on Build GRCh38
rs1001348717 Chr.9: 35480582-35480582 on Build GRCh38
rs1034259902 Chr.9: 35480234-35480234 on Build GRCh38
rs908929177 Chr.9: 35480381-35480381 on Build GRCh38
rs1034465259 Chr.9: 35480449-35480449 on Build GRCh38
rs921756062 Chr.9: 35480422-35480422 on Build GRCh38
rs572474266 Chr.9: 35480394-35480394 on Build GRCh38
rs933185279 Chr.9: 35480460-35480460 on Build GRCh38
rs376146356 Chr.9: 35480404-35480404 on Build GRCh38
rs570998345 Chr.9: 35480231-35480231 on Build GRCh38
rs573911667 Chr.9: 35480281-35480281 on Build GRCh38
rs1030852448 Chr.9: 35480540-35480540 on Build GRCh38
rs1020963846 Chr.9: 35480201-35480201 on Build GRCh38
rs900380264 Chr.9: 35480146-35480146 on Build GRCh38
rs987100753 Chr.9: 35480620-35480620 on Build GRCh38
rs941845831 Chr.9: 35480142-35480142 on Build GRCh38
rs989700283 Chr.9: 35480577-35480577 on Build GRCh38
rs890715501 Chr.9: 35480369-35480369 on Build GRCh38
rs953214203 Chr.9: 35480589-35480589 on Build GRCh38
rs970634671 Chr.9: 35480494-35480494 on Build GRCh38
rs1030242369 Chr.9: 35480165-35480165 on Build GRCh38
rs749805496 Chr.9: 35480503-35480503 on Build GRCh38
rs556853745 Chr.9: 35480280-35480280 on Build GRCh38
rs536889504 Chr.9: 35480279-35480279 on Build GRCh38
rs959786905 Chr.9: 35480594-35480594 on Build GRCh38
rs977925642 Chr.9: 35480275-35480275 on Build GRCh38
rs1034778294 Chr.9: 35480169-35480169 on Build GRCh38
rs968544684 Chr.9: 35480212-35480212 on Build GRCh38
rs1051371579 Chr.9: 35480358-35480358 on Build GRCh38
rs940908561 Chr.9: 35480403-35480403 on Build GRCh38
rs372856496 Chr.9: 35480427-35480427 on Build GRCh38
rs79328000 Chr.9: 35480300-35480300 on Build GRCh38
rs754928506 Chr.9: 35480144-35480144 on Build GRCh38
rs1024274832 Chr.9: 35480476-35480476 on Build GRCh38
rs959600437 Chr.9: 35480266-35480266 on Build GRCh38

View Full Product Details