Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 84364750-84365303 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UOX
Entrez Gene ID
391051
Gene Name
urate oxidase (pseudogene)
Gene Aliases
UOXP, URICASE
UniGene
Hs.647646
Gene Chromosome Location
Chr.1: 84364958-84384801 on Build GRCh38
Transcripts
NR_003927.2

SNP Detail

SNP ID Chromosome Location
rs143139877 Chr.1: 84364777-84364778 on Build GRCh38
rs887402936 Chr.1: 84364933-84364933 on Build GRCh38
rs35104349 Chr.1: 84365242-84365243 on Build GRCh38
rs71767028 Chr.1: 84364779-84364780 on Build GRCh38
rs1034117888 Chr.1: 84364935-84364935 on Build GRCh38
rs776681835 Chr.1: 84364828-84364828 on Build GRCh38
rs537169156 Chr.1: 84364887-84364887 on Build GRCh38
rs976928913 Chr.1: 84365123-84365123 on Build GRCh38
rs547316678 Chr.1: 84364910-84364910 on Build GRCh38
rs553237244 Chr.1: 84365028-84365028 on Build GRCh38
rs115729027 Chr.1: 84365265-84365265 on Build GRCh38
rs111590975 Chr.1: 84365022-84365022 on Build GRCh38
rs1024141870 Chr.1: 84364807-84364807 on Build GRCh38
rs200415161 Chr.1: 84364776-84364777 on Build GRCh38
rs985126684 Chr.1: 84365185-84365185 on Build GRCh38
rs572912808 Chr.1: 84365165-84365165 on Build GRCh38
rs191931442 Chr.1: 84365224-84365224 on Build GRCh38
rs113943590 Chr.1: 84364778-84364779 on Build GRCh38
rs949879000 Chr.1: 84365269-84365269 on Build GRCh38
rs544796669 Chr.1: 84365179-84365179 on Build GRCh38
rs571358849 Chr.1: 84364858-84364858 on Build GRCh38
rs184426593 Chr.1: 84365228-84365228 on Build GRCh38
rs200087272 Chr.1: 84364778-84364778 on Build GRCh38
rs187076820 Chr.1: 84365202-84365202 on Build GRCh38
rs989944686 Chr.1: 84364973-84364973 on Build GRCh38
rs1050522667 Chr.1: 84364913-84364913 on Build GRCh38
rs774161456 Chr.1: 84365045-84365045 on Build GRCh38
rs932928377 Chr.1: 84365168-84365168 on Build GRCh38
rs914322797 Chr.1: 84364967-84364967 on Build GRCh38
rs137966932 Chr.1: 84365233-84365233 on Build GRCh38
rs558377856 Chr.1: 84365192-84365192 on Build GRCh38
rs4001396 Chr.1: 84364780-84364781 on Build GRCh38
rs199699627 Chr.1: 84364776-84364776 on Build GRCh38
rs951299709 Chr.1: 84364866-84364866 on Build GRCh38
rs375670247 Chr.1: 84364966-84364966 on Build GRCh38
rs931401033 Chr.1: 84365278-84365279 on Build GRCh38
rs1003221934 Chr.1: 84364890-84364890 on Build GRCh38
rs1025907292 Chr.1: 84364806-84364806 on Build GRCh38
rs922841861 Chr.1: 84365130-84365130 on Build GRCh38
rs552044009 Chr.1: 84364804-84364804 on Build GRCh38
rs576457925 Chr.1: 84365188-84365188 on Build GRCh38
rs556978973 Chr.1: 84364900-84364900 on Build GRCh38
rs958351834 Chr.1: 84364912-84364912 on Build GRCh38

View Full Product Details