Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 112924152-112924711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKR7A2P1
Entrez Gene ID
246182
Gene Name
aldo-keto reductase family 7 member A2 pseudogene 1
Gene Aliases
AFARP1
UniGene
Hs.647691
Gene Chromosome Location
Chr.1: 112923350-112924673 on Build GRCh38
Transcripts
NR_002796.2 NM_001166496.1 NM_003051.3 XM_011542026.2 XM_011542027.2

Gene Symbol
SLC16A1
Entrez Gene ID
6566
Gene Name
solute carrier family 16 member 1
Gene Aliases
HHF7, MCT, MCT1, MCT1D
UniGene
Hs.75231
Gene Chromosome Location
Chr.1: 112911847-112956353 on Build GRCh38
Transcripts
NR_002796.2 NM_001166496.1 NM_003051.3 XM_011542026.2 XM_011542027.2

SNP Detail

SNP ID Chromosome Location
rs977661952 Chr.1: 112924214-112924214 on Build GRCh38
rs878992555 Chr.1: 112924603-112924606 on Build GRCh38
rs878963357 Chr.1: 112924200-112924200 on Build GRCh38
rs886742331 Chr.1: 112924453-112924453 on Build GRCh38
rs9429506 Chr.1: 112924574-112924574 on Build GRCh38
rs934126201 Chr.1: 112924598-112924598 on Build GRCh38
rs939674050 Chr.1: 112924647-112924647 on Build GRCh38
rs768620684 Chr.1: 112924437-112924437 on Build GRCh38
rs993276408 Chr.1: 112924191-112924191 on Build GRCh38
rs538143978 Chr.1: 112924544-112924544 on Build GRCh38
rs1065662 Chr.1: 112924476-112924476 on Build GRCh38
rs980919345 Chr.1: 112924185-112924185 on Build GRCh38
rs36069670 Chr.1: 112924461-112924461 on Build GRCh38
rs546366475 Chr.1: 112924667-112924667 on Build GRCh38
rs954830676 Chr.1: 112924600-112924600 on Build GRCh38
rs879213822 Chr.1: 112924231-112924231 on Build GRCh38
rs910786372 Chr.1: 112924639-112924639 on Build GRCh38
rs555273150 Chr.1: 112924553-112924553 on Build GRCh38
rs967354277 Chr.1: 112924542-112924542 on Build GRCh38
rs1025308729 Chr.1: 112924530-112924530 on Build GRCh38
rs937891821 Chr.1: 112924662-112924662 on Build GRCh38
rs181535539 Chr.1: 112924255-112924255 on Build GRCh38
rs531635418 Chr.1: 112924197-112924197 on Build GRCh38
rs1039505546 Chr.1: 112924324-112924324 on Build GRCh38
rs542141776 Chr.1: 112924198-112924198 on Build GRCh38
rs987779743 Chr.1: 112924635-112924635 on Build GRCh38
rs988154052 Chr.1: 112924578-112924578 on Build GRCh38
rs1046779192 Chr.1: 112924351-112924351 on Build GRCh38
rs569151354 Chr.1: 112924541-112924541 on Build GRCh38
rs921054803 Chr.1: 112924257-112924257 on Build GRCh38
rs561504729 Chr.1: 112924217-112924217 on Build GRCh38
rs761994656 Chr.1: 112924410-112924410 on Build GRCh38
rs976010804 Chr.1: 112924585-112924588 on Build GRCh38
rs58932372 Chr.1: 112924649-112924649 on Build GRCh38
rs186593737 Chr.1: 112924663-112924663 on Build GRCh38
rs1032414531 Chr.1: 112924565-112924565 on Build GRCh38
rs747043111 Chr.1: 112924384-112924385 on Build GRCh38
rs933600507 Chr.1: 112924279-112924279 on Build GRCh38
rs908246490 Chr.1: 112924625-112924625 on Build GRCh38
rs577365465 Chr.1: 112924659-112924659 on Build GRCh38
rs1024787350 Chr.1: 112924475-112924477 on Build GRCh38
rs776892131 Chr.1: 112924309-112924309 on Build GRCh38
rs532865087 Chr.1: 112924289-112924289 on Build GRCh38
rs567233349 Chr.1: 112924305-112924314 on Build GRCh38
rs866911339 Chr.1: 112924329-112924329 on Build GRCh38
rs878941984 Chr.1: 112924222-112924222 on Build GRCh38
rs933739155 Chr.1: 112924270-112924270 on Build GRCh38
rs959066353 Chr.1: 112924352-112924352 on Build GRCh38
rs761715554 Chr.1: 112924533-112924533 on Build GRCh38
rs968062324 Chr.1: 112924210-112924210 on Build GRCh38
rs958061618 Chr.1: 112924250-112924250 on Build GRCh38
rs190384888 Chr.1: 112924251-112924251 on Build GRCh38
rs397981286 Chr.1: 112924462-112924462 on Build GRCh38
rs1013508681 Chr.1: 112924473-112924473 on Build GRCh38
rs1033513780 Chr.1: 112924325-112924325 on Build GRCh38
rs570169870 Chr.1: 112924282-112924282 on Build GRCh38
rs552347500 Chr.1: 112924396-112924396 on Build GRCh38
rs898233248 Chr.1: 112924340-112924340 on Build GRCh38
rs913880194 Chr.1: 112924273-112924273 on Build GRCh38
rs995233259 Chr.1: 112924347-112924347 on Build GRCh38
rs769627794 Chr.1: 112924554-112924554 on Build GRCh38
rs752286261 Chr.1: 112924220-112924220 on Build GRCh38
rs534164451 Chr.1: 112924597-112924597 on Build GRCh38

View Full Product Details