Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 165701746-165702294 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC440700
Entrez Gene ID
440700
Gene Name
carbonic anhydrase 14 pseudogene
Gene Aliases
-
UniGene
Hs.568703
Gene Chromosome Location
Chr.1: 165698750-165709968 on Build GRCh38
Transcripts
NR_036683.1

SNP Detail

SNP ID Chromosome Location
rs1042584258 Chr.1: 165701937-165701937 on Build GRCh38
rs542224798 Chr.1: 165702200-165702201 on Build GRCh38
rs1002945527 Chr.1: 165702092-165702092 on Build GRCh38
rs565905588 Chr.1: 165702068-165702068 on Build GRCh38
rs1044963578 Chr.1: 165702031-165702031 on Build GRCh38
rs1032573155 Chr.1: 165702214-165702214 on Build GRCh38
rs1050825360 Chr.1: 165701963-165701963 on Build GRCh38
rs35909905 Chr.1: 165702099-165702099 on Build GRCh38
rs758462833 Chr.1: 165701851-165701851 on Build GRCh38
rs149865254 Chr.1: 165701895-165701895 on Build GRCh38
rs923805182 Chr.1: 165702164-165702164 on Build GRCh38
rs958722499 Chr.1: 165702096-165702096 on Build GRCh38
rs898750684 Chr.1: 165702207-165702207 on Build GRCh38
rs575004388 Chr.1: 165702190-165702191 on Build GRCh38
rs898714255 Chr.1: 165702060-165702060 on Build GRCh38
rs111947402 Chr.1: 165702070-165702070 on Build GRCh38
rs969256389 Chr.1: 165702123-165702123 on Build GRCh38
rs59284577 Chr.1: 165702007-165702007 on Build GRCh38
rs369360792 Chr.1: 165702019-165702020 on Build GRCh38
rs780036687 Chr.1: 165701899-165701899 on Build GRCh38
rs371892556 Chr.1: 165702163-165702163 on Build GRCh38
rs372304826 Chr.1: 165702017-165702017 on Build GRCh38
rs898701307 Chr.1: 165702017-165702017 on Build GRCh38
rs10800134 Chr.1: 165702082-165702082 on Build GRCh38
rs775768320 Chr.1: 165702091-165702091 on Build GRCh38
rs915173663 Chr.1: 165702104-165702104 on Build GRCh38
rs933811829 Chr.1: 165702193-165702193 on Build GRCh38
rs76155357 Chr.1: 165702237-165702237 on Build GRCh38
rs931942854 Chr.1: 165701825-165701825 on Build GRCh38
rs1025741727 Chr.1: 165701802-165701802 on Build GRCh38
rs536302340 Chr.1: 165701799-165701799 on Build GRCh38
rs923702117 Chr.1: 165701950-165701950 on Build GRCh38
rs12120652 Chr.1: 165702140-165702140 on Build GRCh38
rs1029175560 Chr.1: 165702154-165702154 on Build GRCh38
rs748054173 Chr.1: 165701915-165701915 on Build GRCh38
rs565227107 Chr.1: 165702270-165702270 on Build GRCh38
rs1036197527 Chr.1: 165701927-165701927 on Build GRCh38
rs537095113 Chr.1: 165702119-165702119 on Build GRCh38
rs938233331 Chr.1: 165702019-165702019 on Build GRCh38
rs942393852 Chr.1: 165702205-165702205 on Build GRCh38
rs79787559 Chr.1: 165702210-165702210 on Build GRCh38
rs139209810 Chr.1: 165701784-165701784 on Build GRCh38
rs576194250 Chr.1: 165701901-165701901 on Build GRCh38
rs771802843 Chr.1: 165702040-165702081 on Build GRCh38
rs535000771 Chr.1: 165702078-165702078 on Build GRCh38
rs527374438 Chr.1: 165702126-165702126 on Build GRCh38
rs1038422346 Chr.1: 165702206-165702206 on Build GRCh38
rs563752461 Chr.1: 165702202-165702203 on Build GRCh38
rs770121066 Chr.1: 165702055-165702055 on Build GRCh38
rs1006545415 Chr.1: 165702045-165702045 on Build GRCh38
rs981409874 Chr.1: 165701894-165701894 on Build GRCh38
rs971431272 Chr.1: 165701845-165701845 on Build GRCh38
rs994730781 Chr.1: 165701778-165701778 on Build GRCh38
rs558417996 Chr.1: 165702197-165702198 on Build GRCh38
rs1020441716 Chr.1: 165702109-165702109 on Build GRCh38
rs994366834 Chr.1: 165702076-165702076 on Build GRCh38
rs911966217 Chr.1: 165701864-165701864 on Build GRCh38
rs375553994 Chr.1: 165702019-165702019 on Build GRCh38
rs987531101 Chr.1: 165701830-165701830 on Build GRCh38
rs904877964 Chr.1: 165702056-165702056 on Build GRCh38
rs545202223 Chr.1: 165702269-165702269 on Build GRCh38
rs191306542 Chr.1: 165702175-165702175 on Build GRCh38
rs971461996 Chr.1: 165702086-165702086 on Build GRCh38
rs11282700 Chr.1: 165701806-165701807 on Build GRCh38
rs933695615 Chr.1: 165701951-165701951 on Build GRCh38
rs12043165 Chr.1: 165702189-165702189 on Build GRCh38
rs372011253 Chr.1: 165701938-165701938 on Build GRCh38
rs915123147 Chr.1: 165701910-165701910 on Build GRCh38
rs10918253 Chr.1: 165702018-165702018 on Build GRCh38
rs774365888 Chr.1: 165702183-165702183 on Build GRCh38
rs889497333 Chr.1: 165701986-165701986 on Build GRCh38
rs140406485 Chr.1: 165702017-165702019 on Build GRCh38
rs10157786 Chr.1: 165702190-165702190 on Build GRCh38
rs1034394413 Chr.1: 165702093-165702093 on Build GRCh38
rs1050857517 Chr.1: 165702196-165702196 on Build GRCh38
rs750630996 Chr.1: 165701820-165701820 on Build GRCh38
rs1003181555 Chr.1: 165702071-165702071 on Build GRCh38
rs553222339 Chr.1: 165702200-165702200 on Build GRCh38
rs369152969 Chr.1: 165702178-165702178 on Build GRCh38
rs1055399965 Chr.1: 165702051-165702051 on Build GRCh38
rs368503392 Chr.1: 165701826-165701826 on Build GRCh38

View Full Product Details