Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 171147502-171148078 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMO6P
Entrez Gene ID
388714
Gene Name
flavin containing monooxygenase 6 pseudogene
Gene Aliases
FMO6
UniGene
Hs.448988
Gene Chromosome Location
Chr.1: 171137740-171161563 on Build GRCh38
Transcripts
NR_002601.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3997069 c.485-5C>T p.? Chr.1: 171147621-171147621 on Build GRCh38
7283577 c.548_549insG p.K185fs*14 Chr.1: 171147689-171147690 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1030486814 Chr.1: 171147705-171147705 on Build GRCh38
rs935367227 Chr.1: 171147777-171147777 on Build GRCh38
rs369971821 Chr.1: 171147640-171147640 on Build GRCh38
rs934985793 Chr.1: 171147605-171147605 on Build GRCh38
rs533431061 Chr.1: 171147778-171147778 on Build GRCh38
rs953686480 Chr.1: 171148004-171148004 on Build GRCh38
rs1044801368 Chr.1: 171147867-171147867 on Build GRCh38
rs6608452 Chr.1: 171147974-171147974 on Build GRCh38
rs556151877 Chr.1: 171147551-171147551 on Build GRCh38
rs545336876 Chr.1: 171147892-171147892 on Build GRCh38
rs1011131911 Chr.1: 171147581-171147581 on Build GRCh38
rs760068984 Chr.1: 171147709-171147709 on Build GRCh38
rs890210163 Chr.1: 171147952-171147952 on Build GRCh38
rs1032559779 Chr.1: 171148028-171148028 on Build GRCh38
rs973272513 Chr.1: 171147725-171147725 on Build GRCh38
rs982527253 Chr.1: 171148037-171148037 on Build GRCh38
rs879117694 Chr.1: 171147641-171147641 on Build GRCh38
rs6608454 Chr.1: 171148031-171148031 on Build GRCh38
rs148136018 Chr.1: 171147542-171147542 on Build GRCh38
rs770270057 Chr.1: 171147683-171147683 on Build GRCh38
rs1052148856 Chr.1: 171147610-171147610 on Build GRCh38
rs571295133 Chr.1: 171148040-171148040 on Build GRCh38
rs949225927 Chr.1: 171147630-171147635 on Build GRCh38
rs114766203 Chr.1: 171148024-171148024 on Build GRCh38
rs879252690 Chr.1: 171147527-171147527 on Build GRCh38
rs750057351 Chr.1: 171147771-171147771 on Build GRCh38
rs780081120 Chr.1: 171147630-171147630 on Build GRCh38
rs1004893182 Chr.1: 171147949-171147949 on Build GRCh38
rs764255419 Chr.1: 171147680-171147680 on Build GRCh38
rs932261154 Chr.1: 171147546-171147546 on Build GRCh38
rs980123151 Chr.1: 171147745-171147745 on Build GRCh38
rs186117678 Chr.1: 171147816-171147816 on Build GRCh38
rs963165618 Chr.1: 171147992-171147992 on Build GRCh38
rs963278579 Chr.1: 171147712-171147712 on Build GRCh38
rs928039245 Chr.1: 171147767-171147767 on Build GRCh38
rs181949394 Chr.1: 171147662-171147662 on Build GRCh38
rs56118593 Chr.1: 171147921-171147921 on Build GRCh38
rs772327139 Chr.1: 171147665-171147665 on Build GRCh38
rs749257124 Chr.1: 171147631-171147631 on Build GRCh38
rs915386938 Chr.1: 171147853-171147853 on Build GRCh38
rs138902170 Chr.1: 171147755-171147755 on Build GRCh38
rs773377852 Chr.1: 171147683-171147683 on Build GRCh38
rs917674362 Chr.1: 171147812-171147812 on Build GRCh38
rs371421675 Chr.1: 171147532-171147532 on Build GRCh38
rs1040346887 Chr.1: 171147894-171147894 on Build GRCh38
rs995852513 Chr.1: 171147651-171147651 on Build GRCh38
rs1052565040 Chr.1: 171147536-171147536 on Build GRCh38
rs956792241 Chr.1: 171148049-171148049 on Build GRCh38
rs966083216 Chr.1: 171147716-171147716 on Build GRCh38
rs6608453 Chr.1: 171148001-171148001 on Build GRCh38
rs573910426 Chr.1: 171147737-171147737 on Build GRCh38
rs987729598 Chr.1: 171147811-171147811 on Build GRCh38
rs146089930 Chr.1: 171147726-171147726 on Build GRCh38
rs768813227 Chr.1: 171147657-171147657 on Build GRCh38
rs537162392 Chr.1: 171147950-171147950 on Build GRCh38
rs771299642 Chr.1: 171147704-171147704 on Build GRCh38
rs572307423 Chr.1: 171147697-171147697 on Build GRCh38
rs544609227 Chr.1: 171147738-171147738 on Build GRCh38
rs931129613 Chr.1: 171147899-171147899 on Build GRCh38
rs145653041 Chr.1: 171147569-171147569 on Build GRCh38
rs7063044 Chr.1: 171147621-171147621 on Build GRCh38
rs776883692 Chr.1: 171147893-171147893 on Build GRCh38
rs982253806 Chr.1: 171147754-171147754 on Build GRCh38
rs926478240 Chr.1: 171147871-171147871 on Build GRCh38
rs946975614 Chr.1: 171147891-171147891 on Build GRCh38
rs1014731039 Chr.1: 171147976-171147976 on Build GRCh38
rs998034838 Chr.1: 171147994-171147994 on Build GRCh38
rs949175847 Chr.1: 171147852-171147852 on Build GRCh38
rs1014677062 Chr.1: 171147710-171147710 on Build GRCh38
rs769010533 Chr.1: 171147572-171147572 on Build GRCh38
rs540724460 Chr.1: 171147706-171147706 on Build GRCh38
rs538960052 Chr.1: 171147661-171147661 on Build GRCh38
rs552263648 Chr.1: 171148020-171148020 on Build GRCh38
rs900198838 Chr.1: 171147906-171147906 on Build GRCh38

View Full Product Details