Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12112647-12113211 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF788
Entrez Gene ID
388507
Gene Name
zinc finger family member 788
Gene Aliases
-
UniGene
Hs.127473
Gene Chromosome Location
Chr.19: 12092263-12114678 on Build GRCh38
Transcripts
NR_027049.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5035969 c.1152C>T p.H384H Chr.19: 12112699-12112699 on Build GRCh38
991142 c.1379C>T p.S460F Chr.19: 12112926-12112926 on Build GRCh38
7223875 c.432T>C p.I144I Chr.19: 12113122-12113122 on Build GRCh38
5485328 c.1165C>T p.R389C Chr.19: 12112712-12112712 on Build GRCh38
991143 c.1385G>A p.R462Q Chr.19: 12112932-12112932 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745597448 Chr.19: 12112736-12112736 on Build GRCh38
rs555488002 Chr.19: 12113037-12113037 on Build GRCh38
rs904413916 Chr.19: 12112987-12112987 on Build GRCh38
rs868311640 Chr.19: 12112871-12112871 on Build GRCh38
rs933627815 Chr.19: 12112681-12112681 on Build GRCh38
rs762065972 Chr.19: 12113053-12113053 on Build GRCh38
rs533837678 Chr.19: 12113016-12113016 on Build GRCh38
rs769395893 Chr.19: 12113015-12113015 on Build GRCh38
rs770368559 Chr.19: 12113118-12113118 on Build GRCh38
rs757317915 Chr.19: 12112944-12112944 on Build GRCh38
rs757334456 Chr.19: 12112848-12112848 on Build GRCh38
rs753915584 Chr.19: 12112837-12112837 on Build GRCh38
rs747869860 Chr.19: 12112884-12112884 on Build GRCh38
rs372951804 Chr.19: 12112776-12112776 on Build GRCh38
rs867224351 Chr.19: 12112989-12112989 on Build GRCh38
rs994273662 Chr.19: 12112801-12112801 on Build GRCh38
rs922279822 Chr.19: 12112676-12112676 on Build GRCh38
rs761044412 Chr.19: 12113035-12113035 on Build GRCh38
rs760159973 Chr.19: 12112921-12112921 on Build GRCh38
rs374731727 Chr.19: 12113075-12113075 on Build GRCh38
rs750333437 Chr.19: 12112966-12112966 on Build GRCh38
rs867902580 Chr.19: 12112875-12112875 on Build GRCh38
rs772592026 Chr.19: 12113022-12113022 on Build GRCh38
rs747508700 Chr.19: 12113133-12113133 on Build GRCh38
rs528480360 Chr.19: 12113099-12113099 on Build GRCh38
rs774994144 Chr.19: 12112755-12112755 on Build GRCh38
rs775867022 Chr.19: 12113030-12113030 on Build GRCh38
rs370051429 Chr.19: 12112880-12112880 on Build GRCh38
rs774304486 Chr.19: 12112729-12112729 on Build GRCh38
rs756142880 Chr.19: 12112713-12112713 on Build GRCh38
rs752123086 Chr.19: 12112796-12112796 on Build GRCh38
rs957558797 Chr.19: 12113146-12113146 on Build GRCh38
rs573810304 Chr.19: 12113062-12113062 on Build GRCh38
rs200392853 Chr.19: 12112765-12112765 on Build GRCh38
rs767990629 Chr.19: 12112800-12112800 on Build GRCh38
rs753056329 Chr.19: 12112932-12112932 on Build GRCh38
rs201287382 Chr.19: 12112768-12112768 on Build GRCh38
rs1019693686 Chr.19: 12113064-12113064 on Build GRCh38
rs777572071 Chr.19: 12113106-12113106 on Build GRCh38
rs746626163 Chr.19: 12113072-12113073 on Build GRCh38
rs1001519197 Chr.19: 12112865-12112865 on Build GRCh38
rs745865505 Chr.19: 12112854-12112854 on Build GRCh38
rs749159053 Chr.19: 12112725-12112725 on Build GRCh38
rs890024185 Chr.19: 12112739-12112739 on Build GRCh38
rs1034347790 Chr.19: 12113138-12113138 on Build GRCh38
rs966912284 Chr.19: 12112891-12112891 on Build GRCh38
rs374288655 Chr.19: 12112684-12112685 on Build GRCh38
rs754057306 Chr.19: 12113039-12113039 on Build GRCh38
rs765372679 Chr.19: 12113167-12113167 on Build GRCh38
rs1024369489 Chr.19: 12112806-12112806 on Build GRCh38
rs764311786 Chr.19: 12112943-12112943 on Build GRCh38
rs769057711 Chr.19: 12113150-12113150 on Build GRCh38
rs980323727 Chr.19: 12113100-12113100 on Build GRCh38
rs972915508 Chr.19: 12112805-12112805 on Build GRCh38
rs966586831 Chr.19: 12113164-12113164 on Build GRCh38
rs779034722 Chr.19: 12112951-12112951 on Build GRCh38
rs771869965 Chr.19: 12112741-12112741 on Build GRCh38
rs544977013 Chr.19: 12113082-12113082 on Build GRCh38
rs748766488 Chr.19: 12113109-12113109 on Build GRCh38
rs777881348 Chr.19: 12112811-12112811 on Build GRCh38
rs74975302 Chr.19: 12112767-12112767 on Build GRCh38
rs369018506 Chr.19: 12112888-12112888 on Build GRCh38
rs865945644 Chr.19: 12112752-12112752 on Build GRCh38
rs765282697 Chr.19: 12113054-12113054 on Build GRCh38
rs758413949 Chr.19: 12112969-12112969 on Build GRCh38
rs904477494 Chr.19: 12112672-12112676 on Build GRCh38
rs757970971 Chr.19: 12113015-12113016 on Build GRCh38
rs577828318 Chr.19: 12113079-12113079 on Build GRCh38
rs146307695 Chr.19: 12113175-12113175 on Build GRCh38
rs761346138 Chr.19: 12112889-12112889 on Build GRCh38
rs779883067 Chr.19: 12112992-12112992 on Build GRCh38
rs771888240 Chr.19: 12113007-12113007 on Build GRCh38
rs922330831 Chr.19: 12112965-12112965 on Build GRCh38
rs778766952 Chr.19: 12112853-12112853 on Build GRCh38
rs575132436 Chr.19: 12112797-12112797 on Build GRCh38
rs777633120 Chr.19: 12113033-12113034 on Build GRCh38
rs776042223 Chr.19: 12113121-12113121 on Build GRCh38
rs747790748 Chr.19: 12113098-12113098 on Build GRCh38
rs543982425 Chr.19: 12112914-12112914 on Build GRCh38
rs766922818 Chr.19: 12112795-12112795 on Build GRCh38
rs769483921 Chr.19: 12112907-12112907 on Build GRCh38
rs564475398 Chr.19: 12113096-12113096 on Build GRCh38
rs768346677 Chr.19: 12113001-12113001 on Build GRCh38
rs746623671 Chr.19: 12112881-12112881 on Build GRCh38
rs1022604005 Chr.19: 12113158-12113158 on Build GRCh38
rs546621372 Chr.19: 12113151-12113151 on Build GRCh38
rs546708935 Chr.19: 12112712-12112712 on Build GRCh38
rs149343838 Chr.19: 12112716-12112716 on Build GRCh38
rs76324660 Chr.19: 12113066-12113066 on Build GRCh38
rs762175038 Chr.19: 12113166-12113166 on Build GRCh38
rs572899192 Chr.19: 12112931-12112931 on Build GRCh38
rs926916542 Chr.19: 12113145-12113145 on Build GRCh38
rs761436481 Chr.19: 12112786-12112786 on Build GRCh38
rs756408133 Chr.19: 12112807-12112807 on Build GRCh38
rs771853550 Chr.19: 12112879-12112879 on Build GRCh38
rs752830730 Chr.19: 12112804-12112804 on Build GRCh38
rs770807059 Chr.19: 12112726-12112726 on Build GRCh38
rs569525504 Chr.19: 12112785-12112785 on Build GRCh38

View Full Product Details