Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 52969040-52969590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF702P
Entrez Gene ID
79986
Gene Name
zinc finger protein 702, pseudogene
Gene Aliases
ZNF702
UniGene
Hs.714428
Gene Chromosome Location
Chr.19: 52968251-52993531 on Build GRCh38
Transcripts
NR_003578.1

SNP Detail

SNP ID Chromosome Location
rs969372423 Chr.19: 52969496-52969496 on Build GRCh38
rs150701168 Chr.19: 52969542-52969542 on Build GRCh38
rs555495330 Chr.19: 52969237-52969237 on Build GRCh38
rs918672713 Chr.19: 52969527-52969527 on Build GRCh38
rs183728980 Chr.19: 52969081-52969081 on Build GRCh38
rs190274557 Chr.19: 52969456-52969456 on Build GRCh38
rs1000301174 Chr.19: 52969327-52969327 on Build GRCh38
rs909047621 Chr.19: 52969176-52969176 on Build GRCh38
rs779663303 Chr.19: 52969358-52969359 on Build GRCh38
rs1053166996 Chr.19: 52969336-52969336 on Build GRCh38
rs1043954900 Chr.19: 52969259-52969259 on Build GRCh38
rs16984267 Chr.19: 52969304-52969304 on Build GRCh38
rs951810375 Chr.19: 52969093-52969093 on Build GRCh38
rs111348428 Chr.19: 52969429-52969429 on Build GRCh38
rs185332309 Chr.19: 52969444-52969444 on Build GRCh38
rs189581365 Chr.19: 52969136-52969136 on Build GRCh38
rs890049627 Chr.19: 52969229-52969229 on Build GRCh38
rs771120691 Chr.19: 52969214-52969214 on Build GRCh38
rs960376227 Chr.19: 52969071-52969071 on Build GRCh38
rs918641636 Chr.19: 52969208-52969208 on Build GRCh38
rs891644641 Chr.19: 52969352-52969352 on Build GRCh38
rs35986023 Chr.19: 52969426-52969429 on Build GRCh38
rs1016079600 Chr.19: 52969500-52969500 on Build GRCh38
rs576123018 Chr.19: 52969173-52969173 on Build GRCh38
rs961865852 Chr.19: 52969506-52969506 on Build GRCh38
rs983139765 Chr.19: 52969131-52969131 on Build GRCh38
rs1037064123 Chr.19: 52969183-52969183 on Build GRCh38
rs774760535 Chr.19: 52969283-52969283 on Build GRCh38
rs941472712 Chr.19: 52969182-52969182 on Build GRCh38
rs916506368 Chr.19: 52969080-52969080 on Build GRCh38
rs375920927 Chr.19: 52969497-52969497 on Build GRCh38
rs141992681 Chr.19: 52969213-52969213 on Build GRCh38
rs984675865 Chr.19: 52969082-52969082 on Build GRCh38
rs541291102 Chr.19: 52969318-52969318 on Build GRCh38
rs908728727 Chr.19: 52969174-52969174 on Build GRCh38
rs763672372 Chr.19: 52969545-52969545 on Build GRCh38
rs76680333 Chr.19: 52969478-52969478 on Build GRCh38
rs755865296 Chr.19: 52969166-52969166 on Build GRCh38
rs999668269 Chr.19: 52969421-52969421 on Build GRCh38
rs112572117 Chr.19: 52969428-52969428 on Build GRCh38
rs568924102 Chr.19: 52969348-52969348 on Build GRCh38
rs904630750 Chr.19: 52969271-52969271 on Build GRCh38
rs770633664 Chr.19: 52969425-52969426 on Build GRCh38
rs934024361 Chr.19: 52969210-52969210 on Build GRCh38
rs193250708 Chr.19: 52969415-52969415 on Build GRCh38
rs573766726 Chr.19: 52969294-52969294 on Build GRCh38
rs564358530 Chr.19: 52969145-52969145 on Build GRCh38
rs759940238 Chr.19: 52969397-52969397 on Build GRCh38
rs568605176 Chr.19: 52969520-52969520 on Build GRCh38
rs1013260707 Chr.19: 52969437-52969437 on Build GRCh38
rs1008696382 Chr.19: 52969365-52969365 on Build GRCh38
rs557765863 Chr.19: 52969166-52969166 on Build GRCh38
rs767977595 Chr.19: 52969426-52969426 on Build GRCh38
rs896186976 Chr.19: 52969432-52969432 on Build GRCh38
rs992212480 Chr.19: 52969072-52969072 on Build GRCh38
rs372140874 Chr.19: 52969428-52969431 on Build GRCh38
rs760128320 Chr.19: 52969526-52969526 on Build GRCh38

View Full Product Details